Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35201007-35201272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD44
Entrez Gene ID
960
Gene Name
CD44 molecule (Indian blood group)
Gene Aliases
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
UniGene
Hs.502328
Gene Chromosome Location
Chr.11: 35138870-35232402 on Build GRCh38
Transcripts
NM_000610.3 NM_001001389.1 NM_001001390.1 NM_001001391.1 NM_001001392.1 NM_001202555.1 NM_001202556.1 NM_001202557.1 XM_005253231.2 XM_005253232.2 XM_005253233.2 XM_005253234.2 XM_005253235.2 XM_005253238.2 XM_005253239.2 XM_005253240.2 XM_006718388.1 XM_006718390.3 XM_011520482.1 XM_011520483.1 XM_011520484.1 XM_011520485.1 XM_011520486.1 XM_011520487.2 XM_011520488.1 XM_011520489.2 XM_017018583.1 XM_017018584.1 XM_017018585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
377836 c.1001C>T p.P334L Chr.11: 35201160-35201160 on Build GRCh38
6869813 c.1000C>T p.P334S Chr.11: 35201159-35201159 on Build GRCh38
309970 c.1028G>T p.R343M Chr.11: 35201187-35201187 on Build GRCh38
1188206 c.976C>A p.Q326K Chr.11: 35201135-35201135 on Build GRCh38
84059 c.1017G>A p.Q339Q Chr.11: 35201176-35201176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751411160 Chr.11: 35201148-35201148 on Build GRCh38
rs375288503 Chr.11: 35201160-35201160 on Build GRCh38
rs758716190 Chr.11: 35201186-35201186 on Build GRCh38
rs372078003 Chr.11: 35201241-35201241 on Build GRCh38
rs747575162 Chr.11: 35201112-35201112 on Build GRCh38
rs774211572 Chr.11: 35201245-35201245 on Build GRCh38
rs773111146 Chr.11: 35201129-35201129 on Build GRCh38
rs915023065 Chr.11: 35201048-35201048 on Build GRCh38
rs1027074054 Chr.11: 35201227-35201227 on Build GRCh38
rs35356320 Chr.11: 35201134-35201134 on Build GRCh38
rs766295362 Chr.11: 35201139-35201139 on Build GRCh38
rs1013383967 Chr.11: 35201223-35201223 on Build GRCh38
rs747414676 Chr.11: 35201189-35201189 on Build GRCh38
rs759520256 Chr.11: 35201149-35201149 on Build GRCh38
rs772949914 Chr.11: 35201237-35201237 on Build GRCh38
rs371484317 Chr.11: 35201099-35201099 on Build GRCh38
rs766203326 Chr.11: 35201186-35201188 on Build GRCh38
rs777212802 Chr.11: 35201180-35201180 on Build GRCh38
rs776027428 Chr.11: 35201107-35201107 on Build GRCh38
rs74986198 Chr.11: 35201212-35201212 on Build GRCh38
rs776216352 Chr.11: 35201121-35201121 on Build GRCh38
rs779239765 Chr.11: 35201096-35201096 on Build GRCh38
rs765065561 Chr.11: 35201034-35201034 on Build GRCh38
rs768013679 Chr.11: 35201113-35201113 on Build GRCh38
rs758405003 Chr.11: 35201065-35201065 on Build GRCh38
rs753547249 Chr.11: 35201210-35201210 on Build GRCh38
rs754008748 Chr.11: 35201156-35201156 on Build GRCh38
rs769261800 Chr.11: 35201194-35201194 on Build GRCh38
rs1057152126 Chr.11: 35201210-35201210 on Build GRCh38
rs780430046 Chr.11: 35201187-35201187 on Build GRCh38
rs747729988 Chr.11: 35201216-35201216 on Build GRCh38
rs560004971 Chr.11: 35201126-35201126 on Build GRCh38
rs749406552 Chr.11: 35201087-35201087 on Build GRCh38
rs750301142 Chr.11: 35201063-35201063 on Build GRCh38
rs777938045 Chr.11: 35201079-35201079 on Build GRCh38
rs757517682 Chr.11: 35201093-35201093 on Build GRCh38
rs748597478 Chr.11: 35201133-35201133 on Build GRCh38
rs187512071 Chr.11: 35201030-35201030 on Build GRCh38
rs757467044 Chr.11: 35201158-35201158 on Build GRCh38
rs1013433910 Chr.11: 35201076-35201076 on Build GRCh38
rs750767970 Chr.11: 35201161-35201161 on Build GRCh38
rs769548167 Chr.11: 35201231-35201231 on Build GRCh38
rs754481709 Chr.11: 35201224-35201225 on Build GRCh38
rs367740427 Chr.11: 35201072-35201072 on Build GRCh38
rs746302323 Chr.11: 35201097-35201097 on Build GRCh38
rs764023818 Chr.11: 35201151-35201151 on Build GRCh38
rs766954190 Chr.11: 35201071-35201071 on Build GRCh38
rs769637367 Chr.11: 35201127-35201127 on Build GRCh38

View Full Product Details