Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29994982-29995255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIRIP3
Entrez Gene ID
8479
Gene Name
HIRA interacting protein 3
Gene Aliases
-
UniGene
Hs.567370
Gene Chromosome Location
Chr.16: 29992321-29996096 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7281668 c.187-7C>A p.? Chr.16: 29995224-29995224 on Build GRCh38
7100122 c.299C>T p.S100L Chr.16: 29995105-29995105 on Build GRCh38
5980962 c.231G>C p.K77N Chr.16: 29995173-29995173 on Build GRCh38
5495312 c.197C>A p.A66D Chr.16: 29995207-29995207 on Build GRCh38
5513182 c.264C>T p.D88D Chr.16: 29995140-29995140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747335921 Chr.16: 29995184-29995184 on Build GRCh38
rs772962130 Chr.16: 29995171-29995171 on Build GRCh38
rs772425545 Chr.16: 29995114-29995114 on Build GRCh38
rs770206676 Chr.16: 29995199-29995199 on Build GRCh38
rs763351617 Chr.16: 29995075-29995076 on Build GRCh38
rs145562651 Chr.16: 29995142-29995142 on Build GRCh38
rs879007282 Chr.16: 29995169-29995169 on Build GRCh38
rs746804249 Chr.16: 29995070-29995070 on Build GRCh38
rs770016466 Chr.16: 29995057-29995058 on Build GRCh38
rs751867660 Chr.16: 29995216-29995216 on Build GRCh38
rs374587572 Chr.16: 29995226-29995226 on Build GRCh38
rs907750312 Chr.16: 29995183-29995183 on Build GRCh38
rs377277376 Chr.16: 29995228-29995228 on Build GRCh38
rs879163555 Chr.16: 29995068-29995068 on Build GRCh38
rs756154606 Chr.16: 29995218-29995219 on Build GRCh38
rs62057683 Chr.16: 29995023-29995023 on Build GRCh38
rs781208594 Chr.16: 29995074-29995074 on Build GRCh38
rs576384905 Chr.16: 29995227-29995227 on Build GRCh38
rs763199720 Chr.16: 29995152-29995152 on Build GRCh38
rs557617885 Chr.16: 29995026-29995026 on Build GRCh38
rs751346333 Chr.16: 29995160-29995160 on Build GRCh38
rs375295069 Chr.16: 29995135-29995135 on Build GRCh38
rs942557541 Chr.16: 29995081-29995081 on Build GRCh38
rs377465758 Chr.16: 29995196-29995196 on Build GRCh38
rs780958050 Chr.16: 29995164-29995164 on Build GRCh38
rs749030526 Chr.16: 29995089-29995089 on Build GRCh38
rs371218681 Chr.16: 29995225-29995225 on Build GRCh38
rs1010526296 Chr.16: 29995047-29995047 on Build GRCh38
rs555295500 Chr.16: 29995119-29995119 on Build GRCh38
rs143383029 Chr.16: 29995177-29995177 on Build GRCh38
rs573565685 Chr.16: 29995120-29995120 on Build GRCh38
rs576190867 Chr.16: 29995037-29995037 on Build GRCh38
rs962956681 Chr.16: 29995223-29995223 on Build GRCh38
rs200521655 Chr.16: 29995122-29995122 on Build GRCh38
rs200237918 Chr.16: 29995175-29995175 on Build GRCh38
rs367843501 Chr.16: 29995104-29995104 on Build GRCh38
rs879127091 Chr.16: 29995107-29995107 on Build GRCh38
rs202024820 Chr.16: 29995077-29995077 on Build GRCh38
rs146248252 Chr.16: 29995147-29995147 on Build GRCh38
rs770403831 Chr.16: 29995186-29995186 on Build GRCh38
rs765338501 Chr.16: 29995052-29995052 on Build GRCh38
rs778560492 Chr.16: 29995181-29995181 on Build GRCh38
rs747568085 Chr.16: 29995075-29995075 on Build GRCh38
rs763144129 Chr.16: 29995207-29995207 on Build GRCh38
rs944536832 Chr.16: 29995185-29995186 on Build GRCh38
rs773285056 Chr.16: 29995204-29995204 on Build GRCh38
rs903809428 Chr.16: 29995125-29995125 on Build GRCh38
rs117241391 Chr.16: 29995071-29995071 on Build GRCh38
rs780061757 Chr.16: 29995065-29995065 on Build GRCh38
rs370819710 Chr.16: 29995233-29995233 on Build GRCh38
rs1040209006 Chr.16: 29995222-29995222 on Build GRCh38
rs376392068 Chr.16: 29995060-29995060 on Build GRCh38
rs148268876 Chr.16: 29995165-29995165 on Build GRCh38
rs769384431 Chr.16: 29995082-29995082 on Build GRCh38
rs974085770 Chr.16: 29995018-29995018 on Build GRCh38
rs761041939 Chr.16: 29995134-29995134 on Build GRCh38
rs765414116 Chr.16: 29995151-29995151 on Build GRCh38
rs749184448 Chr.16: 29995030-29995030 on Build GRCh38
rs759380787 Chr.16: 29995217-29995217 on Build GRCh38
rs539231939 Chr.16: 29995015-29995015 on Build GRCh38
rs777411438 Chr.16: 29995176-29995176 on Build GRCh38
rs776988811 Chr.16: 29995189-29995189 on Build GRCh38
rs777271103 Chr.16: 29995087-29995087 on Build GRCh38
rs758252848 Chr.16: 29995063-29995063 on Build GRCh38
rs766721352 Chr.16: 29995156-29995156 on Build GRCh38
rs754847685 Chr.16: 29995162-29995162 on Build GRCh38

View Full Product Details