Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5499155-5499408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALML5
Entrez Gene ID
51806
Gene Name
calmodulin like 5
Gene Aliases
CLSP
UniGene
Hs.180142
Gene Chromosome Location
Chr.10: 5498695-5499570 on Build GRCh38
Transcripts
NM_017422.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7113529 c.223A>G p.K75E Chr.10: 5499216-5499216 on Build GRCh38
1474658 c.103G>A p.A35T Chr.10: 5499336-5499336 on Build GRCh38
4014637 c.177C>T p.D59D Chr.10: 5499262-5499262 on Build GRCh38
4478931 c.227C>T p.A76V Chr.10: 5499212-5499212 on Build GRCh38
1561299 c.219G>A p.A73A Chr.10: 5499220-5499220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999235185 Chr.10: 5499177-5499177 on Build GRCh38
rs10904516 Chr.10: 5499218-5499218 on Build GRCh38
rs750445293 Chr.10: 5499334-5499334 on Build GRCh38
rs145226223 Chr.10: 5499276-5499276 on Build GRCh38
rs763347994 Chr.10: 5499325-5499325 on Build GRCh38
rs139732277 Chr.10: 5499175-5499175 on Build GRCh38
rs372864389 Chr.10: 5499309-5499309 on Build GRCh38
rs140538980 Chr.10: 5499332-5499332 on Build GRCh38
rs760264939 Chr.10: 5499263-5499263 on Build GRCh38
rs768558251 Chr.10: 5499253-5499253 on Build GRCh38
rs768112457 Chr.10: 5499174-5499174 on Build GRCh38
rs112999192 Chr.10: 5499305-5499305 on Build GRCh38
rs934990668 Chr.10: 5499381-5499381 on Build GRCh38
rs767518419 Chr.10: 5499277-5499277 on Build GRCh38
rs760605728 Chr.10: 5499206-5499250 on Build GRCh38
rs755891210 Chr.10: 5499237-5499237 on Build GRCh38
rs61164868 Chr.10: 5499363-5499363 on Build GRCh38
rs752116988 Chr.10: 5499275-5499275 on Build GRCh38
rs777826629 Chr.10: 5499373-5499373 on Build GRCh38
rs758946270 Chr.10: 5499213-5499213 on Build GRCh38
rs753604806 Chr.10: 5499266-5499267 on Build GRCh38
rs377606833 Chr.10: 5499385-5499385 on Build GRCh38
rs746579866 Chr.10: 5499319-5499319 on Build GRCh38
rs763116423 Chr.10: 5499267-5499268 on Build GRCh38
rs10904517 Chr.10: 5499220-5499220 on Build GRCh38
rs202221308 Chr.10: 5499220-5499220 on Build GRCh38
rs759875371 Chr.10: 5499192-5499192 on Build GRCh38
rs757347262 Chr.10: 5499221-5499221 on Build GRCh38
rs201275265 Chr.10: 5499369-5499369 on Build GRCh38
rs771484147 Chr.10: 5499261-5499261 on Build GRCh38
rs767184915 Chr.10: 5499181-5499181 on Build GRCh38
rs144222183 Chr.10: 5499314-5499314 on Build GRCh38
rs770478496 Chr.10: 5499320-5499320 on Build GRCh38
rs764697796 Chr.10: 5499274-5499274 on Build GRCh38
rs11546426 Chr.10: 5499267-5499267 on Build GRCh38
rs199855788 Chr.10: 5499224-5499224 on Build GRCh38
rs1008866162 Chr.10: 5499272-5499272 on Build GRCh38
rs780302997 Chr.10: 5499294-5499294 on Build GRCh38
rs761360982 Chr.10: 5499176-5499176 on Build GRCh38
rs779853119 Chr.10: 5499239-5499239 on Build GRCh38
rs376685895 Chr.10: 5499241-5499241 on Build GRCh38
rs747649025 Chr.10: 5499259-5499259 on Build GRCh38
rs778879510 Chr.10: 5499255-5499255 on Build GRCh38
rs893536813 Chr.10: 5499383-5499383 on Build GRCh38
rs935990248 Chr.10: 5499226-5499226 on Build GRCh38
rs141020784 Chr.10: 5499372-5499372 on Build GRCh38
rs753218442 Chr.10: 5499206-5499206 on Build GRCh38
rs373657626 Chr.10: 5499217-5499220 on Build GRCh38
rs139568157 Chr.10: 5499265-5499265 on Build GRCh38
rs549120790 Chr.10: 5499256-5499256 on Build GRCh38
rs776494109 Chr.10: 5499321-5499321 on Build GRCh38
rs373492621 Chr.10: 5499382-5499382 on Build GRCh38
rs745541869 Chr.10: 5499384-5499384 on Build GRCh38
rs893339664 Chr.10: 5499191-5499191 on Build GRCh38
rs149170647 Chr.10: 5499285-5499285 on Build GRCh38
rs112779065 Chr.10: 5499370-5499370 on Build GRCh38
rs386740368 Chr.10: 5499218-5499220 on Build GRCh38
rs760863630 Chr.10: 5499336-5499336 on Build GRCh38
rs766290977 Chr.10: 5499256-5499256 on Build GRCh38
rs540319277 Chr.10: 5499283-5499283 on Build GRCh38
rs781409942 Chr.10: 5499222-5499222 on Build GRCh38
rs755228272 Chr.10: 5499307-5499307 on Build GRCh38
rs113273825 Chr.10: 5499219-5499219 on Build GRCh38
rs771558621 Chr.10: 5499310-5499310 on Build GRCh38
rs200883143 Chr.10: 5499374-5499374 on Build GRCh38
rs753615918 Chr.10: 5499297-5499297 on Build GRCh38
rs145893521 Chr.10: 5499337-5499337 on Build GRCh38
rs778701515 Chr.10: 5499368-5499368 on Build GRCh38
rs765531522 Chr.10: 5499204-5499204 on Build GRCh38
rs768107163 Chr.10: 5499333-5499333 on Build GRCh38
rs772801950 Chr.10: 5499186-5499186 on Build GRCh38
rs116143222 Chr.10: 5499331-5499331 on Build GRCh38

View Full Product Details