Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 133920732-133920977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF9B
Entrez Gene ID
22997
Gene Name
immunoglobulin superfamily member 9B
Gene Aliases
-
UniGene
Hs.204121
Gene Chromosome Location
Chr.11: 133908564-133956960 on Build GRCh38
Transcripts
NM_001277285.1 XM_006718795.3 XM_011542691.2 XM_011542692.2 XM_011542694.2 XM_011542695.2 XM_017017393.1 XM_017017394.1 XM_017017395.1 XM_017017396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
320959 c.1330G>A p.V444M Chr.11: 133920763-133920763 on Build GRCh38
687197 c.1352T>A p.V451D Chr.11: 133920741-133920741 on Build GRCh38
6519769 c.1176G>T p.G392G Chr.11: 133920917-133920917 on Build GRCh38
6028109 c.1243delC p.R415fs*34 Chr.11: 133920850-133920850 on Build GRCh38
145511 c.1313C>T p.P438L Chr.11: 133920780-133920780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773909542 Chr.11: 133920756-133920756 on Build GRCh38
rs372000338 Chr.11: 133920779-133920779 on Build GRCh38
rs751732770 Chr.11: 133920793-133920793 on Build GRCh38
rs750701608 Chr.11: 133920952-133920952 on Build GRCh38
rs778574126 Chr.11: 133920845-133920845 on Build GRCh38
rs1039873023 Chr.11: 133920819-133920819 on Build GRCh38
rs765609202 Chr.11: 133920944-133920944 on Build GRCh38
rs369321654 Chr.11: 133920934-133920934 on Build GRCh38
rs998361553 Chr.11: 133920854-133920854 on Build GRCh38
rs756219238 Chr.11: 133920888-133920888 on Build GRCh38
rs866220109 Chr.11: 133920810-133920810 on Build GRCh38
rs749627499 Chr.11: 133920840-133920840 on Build GRCh38
rs1041715439 Chr.11: 133920767-133920767 on Build GRCh38
rs781306282 Chr.11: 133920817-133920817 on Build GRCh38
rs564750590 Chr.11: 133920856-133920856 on Build GRCh38
rs779872070 Chr.11: 133920913-133920913 on Build GRCh38
rs764955623 Chr.11: 133920864-133920864 on Build GRCh38
rs375067578 Chr.11: 133920780-133920780 on Build GRCh38
rs766581832 Chr.11: 133920786-133920786 on Build GRCh38
rs761600588 Chr.11: 133920863-133920863 on Build GRCh38
rs770140795 Chr.11: 133920750-133920750 on Build GRCh38
rs201156438 Chr.11: 133920773-133920773 on Build GRCh38
rs533134144 Chr.11: 133920939-133920939 on Build GRCh38
rs745569337 Chr.11: 133920849-133920849 on Build GRCh38
rs778276636 Chr.11: 133920838-133920838 on Build GRCh38
rs574419265 Chr.11: 133920754-133920754 on Build GRCh38
rs778188740 Chr.11: 133920889-133920889 on Build GRCh38
rs752727489 Chr.11: 133920878-133920878 on Build GRCh38
rs771206542 Chr.11: 133920844-133920844 on Build GRCh38
rs755027167 Chr.11: 133920803-133920803 on Build GRCh38
rs368677086 Chr.11: 133920876-133920876 on Build GRCh38
rs773607839 Chr.11: 133920751-133920751 on Build GRCh38
rs573512453 Chr.11: 133920865-133920865 on Build GRCh38
rs752645864 Chr.11: 133920858-133920858 on Build GRCh38
rs546570473 Chr.11: 133920855-133920855 on Build GRCh38
rs771896150 Chr.11: 133920910-133920910 on Build GRCh38
rs767563740 Chr.11: 133920877-133920877 on Build GRCh38
rs1047657878 Chr.11: 133920919-133920919 on Build GRCh38
rs997206437 Chr.11: 133920911-133920911 on Build GRCh38
rs372637704 Chr.11: 133920920-133920920 on Build GRCh38
rs373053404 Chr.11: 133920921-133920921 on Build GRCh38
rs760757168 Chr.11: 133920775-133920775 on Build GRCh38
rs775176299 Chr.11: 133920850-133920850 on Build GRCh38
rs753172962 Chr.11: 133920829-133920829 on Build GRCh38
rs763945705 Chr.11: 133920776-133920776 on Build GRCh38
rs375404016 Chr.11: 133920922-133920922 on Build GRCh38
rs374101848 Chr.11: 133920772-133920772 on Build GRCh38
rs551042626 Chr.11: 133920924-133920924 on Build GRCh38
rs199607489 Chr.11: 133920905-133920905 on Build GRCh38
rs770569301 Chr.11: 133920755-133920755 on Build GRCh38
rs762782479 Chr.11: 133920849-133920850 on Build GRCh38
rs745498636 Chr.11: 133920909-133920909 on Build GRCh38
rs756662862 Chr.11: 133920835-133920835 on Build GRCh38
rs776739531 Chr.11: 133920860-133920860 on Build GRCh38
rs971449601 Chr.11: 133920785-133920785 on Build GRCh38
rs773203344 Chr.11: 133920803-133920805 on Build GRCh38
rs763829457 Chr.11: 133920933-133920933 on Build GRCh38
rs201878845 Chr.11: 133920904-133920904 on Build GRCh38

View Full Product Details