Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13480922-13481177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND7
Entrez Gene ID
222389
Gene Name
BEN domain containing 7
Gene Aliases
C10orf30
UniGene
Hs.498740
Gene Chromosome Location
Chr.10: 13438484-13528772 on Build GRCh38
Transcripts
NM_001100912.1 NM_152751.2 XM_005252402.4 XM_011519390.2 XM_011519391.2 XM_011519392.2 XM_011519393.2 XM_011519394.2 XM_011519395.2 XM_011519396.2 XM_011519397.2 XM_011519398.2 XM_011519399.2 XM_011519400.2 XM_011519401.2 XM_011519402.2 XM_011519403.2 XM_011519404.2 XM_011519405.2 XM_011519406.2 XM_011519407.1 XM_011519408.2 XM_011519410.2 XM_011519411.2 XM_011519412.1 XM_011519413.1 XM_011519414.1 XM_017015915.1 XM_017015916.1 XM_017015917.1 XM_017015918.1 XM_017015919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3436046 c.897T>C p.S299S Chr.10: 13481065-13481065 on Build GRCh38
3436042 c.866C>T p.S289F Chr.10: 13480979-13480979 on Build GRCh38
6257104 c.762C>A p.R254R Chr.10: 13481044-13481044 on Build GRCh38
6988531 c.908G>A p.R303Q Chr.10: 13480937-13480937 on Build GRCh38
4665627 c.804_805insT p.D269fs*1 Chr.10: 13481001-13481002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757576138 Chr.10: 13481023-13481023 on Build GRCh38
rs766808051 Chr.10: 13481081-13481082 on Build GRCh38
rs781555829 Chr.10: 13481036-13481036 on Build GRCh38
rs997634739 Chr.10: 13481064-13481064 on Build GRCh38
rs768901330 Chr.10: 13481066-13481066 on Build GRCh38
rs753284452 Chr.10: 13480994-13480994 on Build GRCh38
rs374073381 Chr.10: 13480946-13480946 on Build GRCh38
rs765934614 Chr.10: 13481095-13481095 on Build GRCh38
rs758937817 Chr.10: 13481133-13481133 on Build GRCh38
rs560584324 Chr.10: 13481086-13481086 on Build GRCh38
rs773148014 Chr.10: 13481084-13481084 on Build GRCh38
rs774393502 Chr.10: 13481069-13481069 on Build GRCh38
rs999588350 Chr.10: 13481053-13481053 on Build GRCh38
rs1051813628 Chr.10: 13481132-13481132 on Build GRCh38
rs747553014 Chr.10: 13481014-13481014 on Build GRCh38
rs749309570 Chr.10: 13481055-13481055 on Build GRCh38
rs192505822 Chr.10: 13481142-13481142 on Build GRCh38
rs746041860 Chr.10: 13481046-13481046 on Build GRCh38
rs367691032 Chr.10: 13481050-13481050 on Build GRCh38
rs369077810 Chr.10: 13481113-13481113 on Build GRCh38
rs765848477 Chr.10: 13480959-13480959 on Build GRCh38
rs542722342 Chr.10: 13481092-13481092 on Build GRCh38
rs868449745 Chr.10: 13481026-13481026 on Build GRCh38
rs148207007 Chr.10: 13481075-13481075 on Build GRCh38
rs778229752 Chr.10: 13481007-13481007 on Build GRCh38
rs371441138 Chr.10: 13481052-13481052 on Build GRCh38
rs1041930214 Chr.10: 13481024-13481024 on Build GRCh38
rs761935473 Chr.10: 13481010-13481010 on Build GRCh38
rs767364240 Chr.10: 13481081-13481081 on Build GRCh38

View Full Product Details