Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36333763-36334006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0895
Entrez Gene ID
23366
Gene Name
KIAA0895
Gene Aliases
-
UniGene
Hs.6224
Gene Chromosome Location
Chr.7: 36324150-36390125 on Build GRCh38
Transcripts
NM_001100425.1 NM_001199706.1 NM_001199707.1 NM_001199708.1 NM_001300956.1 NM_015314.2 XM_005249689.3 XM_005249690.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1450542 c.1154A>G p.Q385R Chr.7: 36333969-36333969 on Build GRCh38
6177534 c.1227C>A p.G409G Chr.7: 36333935-36333935 on Build GRCh38
3674926 c.1118G>T p.R373L Chr.7: 36333891-36333891 on Build GRCh38
3228797 c.1246A>G p.N416D Chr.7: 36333916-36333916 on Build GRCh38
4186838 c.1256C>T p.T419I Chr.7: 36333867-36333867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746595334 Chr.7: 36333927-36333927 on Build GRCh38
rs371846023 Chr.7: 36333915-36333915 on Build GRCh38
rs766376129 Chr.7: 36333904-36333904 on Build GRCh38
rs777442652 Chr.7: 36333809-36333809 on Build GRCh38
rs964572099 Chr.7: 36333813-36333813 on Build GRCh38
rs778958068 Chr.7: 36333920-36333920 on Build GRCh38
rs769266123 Chr.7: 36333858-36333858 on Build GRCh38
rs769015417 Chr.7: 36333972-36333972 on Build GRCh38
rs561902520 Chr.7: 36333897-36333897 on Build GRCh38
rs74737039 Chr.7: 36333928-36333928 on Build GRCh38
rs375560072 Chr.7: 36333828-36333828 on Build GRCh38
rs771600853 Chr.7: 36333808-36333808 on Build GRCh38
rs754895603 Chr.7: 36333918-36333918 on Build GRCh38
rs556893719 Chr.7: 36333827-36333827 on Build GRCh38
rs760897071 Chr.7: 36333899-36333899 on Build GRCh38
rs774560788 Chr.7: 36333875-36333875 on Build GRCh38
rs781690509 Chr.7: 36333884-36333884 on Build GRCh38
rs201673199 Chr.7: 36333931-36333931 on Build GRCh38
rs146789655 Chr.7: 36333846-36333846 on Build GRCh38
rs767800476 Chr.7: 36333892-36333892 on Build GRCh38
rs763553070 Chr.7: 36333857-36333857 on Build GRCh38
rs762282551 Chr.7: 36333891-36333891 on Build GRCh38
rs745423634 Chr.7: 36333942-36333942 on Build GRCh38
rs758163959 Chr.7: 36333926-36333926 on Build GRCh38
rs752403478 Chr.7: 36333922-36333922 on Build GRCh38
rs542254419 Chr.7: 36333889-36333889 on Build GRCh38

View Full Product Details