Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77818924-77819163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK1
Entrez Gene ID
57143
Gene Name
aarF domain containing kinase 1
Gene Aliases
-
UniGene
Hs.413208
Gene Chromosome Location
Chr.14: 77800058-77933954 on Build GRCh38
Transcripts
NM_001142545.1 NM_020421.3 XM_005267885.3 XM_005267886.3 XM_011536987.2 XM_011536988.2 XM_017021474.1 XM_017021475.1 XM_017021476.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6594759 c.106G>A p.A36T Chr.14: 77819084-77819084 on Build GRCh38
4247824 c.135G>A p.T45T Chr.14: 77819113-77819113 on Build GRCh38
6594760 c.106G>A p.A36T Chr.14: 77819084-77819084 on Build GRCh38
4247830 c.135+10T>C p.? Chr.14: 77819123-77819123 on Build GRCh38
1562520 c.105C>T p.G35G Chr.14: 77819083-77819083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758826407 Chr.14: 77819080-77819080 on Build GRCh38
rs761748522 Chr.14: 77819017-77819017 on Build GRCh38
rs764960245 Chr.14: 77819107-77819107 on Build GRCh38
rs748556697 Chr.14: 77818985-77818985 on Build GRCh38
rs753191750 Chr.14: 77819066-77819066 on Build GRCh38
rs376359656 Chr.14: 77818965-77818965 on Build GRCh38
rs889089102 Chr.14: 77819117-77819117 on Build GRCh38
rs759664408 Chr.14: 77819060-77819060 on Build GRCh38
rs917002220 Chr.14: 77819067-77819067 on Build GRCh38
rs372662986 Chr.14: 77819030-77819030 on Build GRCh38
rs779086112 Chr.14: 77818955-77818956 on Build GRCh38
rs201496237 Chr.14: 77819034-77819034 on Build GRCh38
rs369119224 Chr.14: 77819021-77819021 on Build GRCh38
rs769350486 Chr.14: 77819092-77819092 on Build GRCh38
rs925886615 Chr.14: 77819103-77819103 on Build GRCh38
rs200680265 Chr.14: 77819038-77819038 on Build GRCh38
rs758900355 Chr.14: 77819095-77819095 on Build GRCh38
rs780877558 Chr.14: 77818989-77818989 on Build GRCh38
rs886578267 Chr.14: 77819073-77819073 on Build GRCh38
rs1040474780 Chr.14: 77819134-77819134 on Build GRCh38
rs753899637 Chr.14: 77819051-77819051 on Build GRCh38
rs945898824 Chr.14: 77819068-77819068 on Build GRCh38
rs748788482 Chr.14: 77819113-77819113 on Build GRCh38
rs199753100 Chr.14: 77819123-77819123 on Build GRCh38
rs377302968 Chr.14: 77819135-77819135 on Build GRCh38
rs774397600 Chr.14: 77819010-77819010 on Build GRCh38
rs778113031 Chr.14: 77819081-77819081 on Build GRCh38
rs370327529 Chr.14: 77819083-77819083 on Build GRCh38
rs1027802445 Chr.14: 77819138-77819138 on Build GRCh38
rs745513627 Chr.14: 77818991-77818991 on Build GRCh38
rs375951848 Chr.14: 77819018-77819018 on Build GRCh38
rs2287647 Chr.14: 77819131-77819131 on Build GRCh38
rs774873151 Chr.14: 77818992-77818992 on Build GRCh38
rs767605841 Chr.14: 77819119-77819119 on Build GRCh38
rs111982533 Chr.14: 77819002-77819002 on Build GRCh38
rs779356305 Chr.14: 77818984-77818984 on Build GRCh38
rs139573437 Chr.14: 77819008-77819008 on Build GRCh38
rs397711074 Chr.14: 77819123-77819123 on Build GRCh38
rs199563479 Chr.14: 77819112-77819112 on Build GRCh38
rs201490252 Chr.14: 77819007-77819007 on Build GRCh38
rs199703595 Chr.14: 77819118-77819118 on Build GRCh38
rs3214433 Chr.14: 77819119-77819119 on Build GRCh38
rs369655177 Chr.14: 77818975-77818975 on Build GRCh38
rs772080000 Chr.14: 77819137-77819137 on Build GRCh38
rs373839882 Chr.14: 77818976-77818976 on Build GRCh38
rs962145523 Chr.14: 77818972-77818972 on Build GRCh38
rs375700586 Chr.14: 77819043-77819043 on Build GRCh38

View Full Product Details