Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 20212011-20212268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1B2
Entrez Gene ID
526
Gene Name
ATPase H+ transporting V1 subunit B2
Gene Aliases
ATP6B1B2, ATP6B2, DOOD, HO57, VATB, VPP3, Vma2, ZLS2
UniGene
Hs.295917
Gene Chromosome Location
Chr.8: 20197193-20226852 on Build GRCh38
Transcripts
NM_001693.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1097888 c.748G>T p.E250* Chr.8: 20212144-20212144 on Build GRCh38
5063018 c.713T>C p.M238T Chr.8: 20212109-20212109 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778048934 Chr.8: 20212240-20212240 on Build GRCh38
rs779930328 Chr.8: 20212238-20212238 on Build GRCh38
rs377219665 Chr.8: 20212245-20212245 on Build GRCh38
rs769246654 Chr.8: 20212198-20212198 on Build GRCh38
rs751381654 Chr.8: 20212099-20212099 on Build GRCh38
rs762403969 Chr.8: 20212209-20212209 on Build GRCh38
rs372562556 Chr.8: 20212238-20212238 on Build GRCh38
rs369198184 Chr.8: 20212199-20212199 on Build GRCh38
rs771254645 Chr.8: 20212156-20212156 on Build GRCh38
rs779877448 Chr.8: 20212097-20212097 on Build GRCh38
rs756043862 Chr.8: 20212239-20212239 on Build GRCh38
rs754766322 Chr.8: 20212107-20212107 on Build GRCh38
rs999017654 Chr.8: 20212041-20212041 on Build GRCh38
rs762235475 Chr.8: 20212231-20212231 on Build GRCh38
rs756228385 Chr.8: 20212080-20212082 on Build GRCh38
rs369397805 Chr.8: 20212083-20212083 on Build GRCh38
rs753205501 Chr.8: 20212063-20212063 on Build GRCh38
rs775920350 Chr.8: 20212189-20212189 on Build GRCh38
rs772425223 Chr.8: 20212170-20212170 on Build GRCh38
rs749916258 Chr.8: 20212206-20212206 on Build GRCh38
rs368933775 Chr.8: 20212057-20212057 on Build GRCh38
rs780999092 Chr.8: 20212108-20212108 on Build GRCh38
rs760008719 Chr.8: 20212164-20212164 on Build GRCh38
rs146371934 Chr.8: 20212061-20212061 on Build GRCh38
rs17092160 Chr.8: 20212128-20212128 on Build GRCh38
rs777300174 Chr.8: 20212215-20212215 on Build GRCh38
rs761245266 Chr.8: 20212067-20212067 on Build GRCh38
rs764790997 Chr.8: 20212072-20212072 on Build GRCh38
rs758015407 Chr.8: 20212091-20212091 on Build GRCh38
rs376378032 Chr.8: 20212149-20212149 on Build GRCh38
rs754740759 Chr.8: 20212233-20212233 on Build GRCh38
rs373439304 Chr.8: 20212090-20212090 on Build GRCh38
rs748057717 Chr.8: 20212120-20212120 on Build GRCh38
rs774665238 Chr.8: 20212163-20212163 on Build GRCh38

View Full Product Details