Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 96370776-96371047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC82
Entrez Gene ID
79780
Gene Name
coiled-coil domain containing 82
Gene Aliases
HSPC048
UniGene
Hs.525088
Gene Chromosome Location
Chr.11: 96352765-96389940 on Build GRCh38
Transcripts
NM_001318736.1 NM_024725.3 XM_005274292.3 XM_006718911.3 XM_011542984.2 XM_011542987.2 XM_011542988.2 XM_017018305.1 XM_017018306.1 XM_017018307.1 XM_017018308.1 XM_017018309.1 XM_017018310.1 XM_017018311.1 XM_017018312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467615 c.1176G>T p.L392F Chr.11: 96371046-96371046 on Build GRCh38
5571373 c.1179A>G p.V393V Chr.11: 96371043-96371043 on Build GRCh38
1188373 c.1181C>G p.S394C Chr.11: 96371041-96371041 on Build GRCh38
2062010 c.1184G>T p.R395I Chr.11: 96371038-96371038 on Build GRCh38
4903097 c.1189C>T p.R397C Chr.11: 96371033-96371033 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773910055 Chr.11: 96370984-96370984 on Build GRCh38
rs544137257 Chr.11: 96370862-96370862 on Build GRCh38
rs185959891 Chr.11: 96370893-96370893 on Build GRCh38
rs1003124261 Chr.11: 96370798-96370798 on Build GRCh38
rs981981727 Chr.11: 96370854-96370854 on Build GRCh38
rs1033115373 Chr.11: 96370993-96370993 on Build GRCh38
rs775148310 Chr.11: 96371006-96371006 on Build GRCh38
rs769165002 Chr.11: 96370974-96370974 on Build GRCh38
rs969193242 Chr.11: 96370840-96370840 on Build GRCh38
rs560577220 Chr.11: 96370979-96370979 on Build GRCh38
rs934472910 Chr.11: 96371020-96371020 on Build GRCh38
rs765801197 Chr.11: 96370837-96370837 on Build GRCh38
rs564747118 Chr.11: 96370844-96370844 on Build GRCh38
rs545958079 Chr.11: 96370799-96370799 on Build GRCh38
rs531532018 Chr.11: 96370976-96370976 on Build GRCh38
rs7117427 Chr.11: 96370914-96370914 on Build GRCh38
rs373060828 Chr.11: 96370991-96370991 on Build GRCh38
rs375904117 Chr.11: 96370980-96370980 on Build GRCh38
rs770262215 Chr.11: 96370983-96370983 on Build GRCh38
rs200246505 Chr.11: 96371019-96371019 on Build GRCh38
rs531802516 Chr.11: 96370858-96370858 on Build GRCh38
rs547451649 Chr.11: 96370916-96370916 on Build GRCh38
rs565659149 Chr.11: 96370926-96370926 on Build GRCh38
rs763850284 Chr.11: 96371014-96371014 on Build GRCh38
rs900114721 Chr.11: 96370928-96370928 on Build GRCh38
rs780426490 Chr.11: 96370972-96370972 on Build GRCh38
rs190338280 Chr.11: 96370978-96370978 on Build GRCh38
rs375949005 Chr.11: 96370878-96370878 on Build GRCh38
rs998305070 Chr.11: 96370942-96370942 on Build GRCh38
rs373012169 Chr.11: 96370983-96370983 on Build GRCh38
rs995607854 Chr.11: 96370892-96370892 on Build GRCh38
rs781750027 Chr.11: 96370979-96370979 on Build GRCh38
rs1046815608 Chr.11: 96370895-96370895 on Build GRCh38
rs927691182 Chr.11: 96370881-96370881 on Build GRCh38
rs374507296 Chr.11: 96371001-96371001 on Build GRCh38
rs760426830 Chr.11: 96371008-96371008 on Build GRCh38
rs924448791 Chr.11: 96371009-96371009 on Build GRCh38
rs912559322 Chr.11: 96370945-96370945 on Build GRCh38
rs771657729 Chr.11: 96371002-96371002 on Build GRCh38

View Full Product Details