Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19132282-19132549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
UniGene
Hs.517407
Gene Chromosome Location
Chr.22: 19130279-19144726 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 NM_053006.4

Gene Symbol
TSSK2
Entrez Gene ID
23617
Gene Name
testis specific serine kinase 2
Gene Aliases
DGS-G, SPOGA2, STK22B, TSK2
UniGene
Hs.694070
Gene Chromosome Location
Chr.22: 19130808-19132623 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 NM_053006.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5981809 c.1066G>A p.A356T Chr.22: 19132465-19132465 on Build GRCh38
6810767 c.965G>A p.R322Q Chr.22: 19132364-19132364 on Build GRCh38
7395529 c.987C>A p.N329K Chr.22: 19132386-19132386 on Build GRCh38
5858894 c.1054G>T p.E352* Chr.22: 19132453-19132453 on Build GRCh38
1414854 c.1047C>T p.S349S Chr.22: 19132446-19132446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139396065 Chr.22: 19132317-19132317 on Build GRCh38
rs537157579 Chr.22: 19132442-19132442 on Build GRCh38
rs762070638 Chr.22: 19132316-19132327 on Build GRCh38
rs767669673 Chr.22: 19132328-19132339 on Build GRCh38
rs781106529 Chr.22: 19132421-19132421 on Build GRCh38
rs34029289 Chr.22: 19132487-19132488 on Build GRCh38
rs750317045 Chr.22: 19132508-19132508 on Build GRCh38
rs151015327 Chr.22: 19132455-19132455 on Build GRCh38
rs1033214507 Chr.22: 19132392-19132392 on Build GRCh38
rs779784685 Chr.22: 19132435-19132435 on Build GRCh38
rs542637553 Chr.22: 19132465-19132465 on Build GRCh38
rs777773500 Chr.22: 19132327-19132327 on Build GRCh38
rs146810069 Chr.22: 19132380-19132380 on Build GRCh38
rs752596217 Chr.22: 19132321-19132321 on Build GRCh38
rs778694870 Chr.22: 19132515-19132515 on Build GRCh38
rs750597559 Chr.22: 19132389-19132389 on Build GRCh38
rs200682587 Chr.22: 19132446-19132446 on Build GRCh38
rs370350369 Chr.22: 19132525-19132525 on Build GRCh38
rs140549220 Chr.22: 19132414-19132414 on Build GRCh38
rs757230340 Chr.22: 19132416-19132416 on Build GRCh38
rs769115156 Chr.22: 19132434-19132434 on Build GRCh38
rs772763295 Chr.22: 19132460-19132460 on Build GRCh38
rs373650594 Chr.22: 19132328-19132328 on Build GRCh38
rs759728904 Chr.22: 19132491-19132491 on Build GRCh38
rs745355051 Chr.22: 19132363-19132363 on Build GRCh38
rs758245828 Chr.22: 19132407-19132407 on Build GRCh38
rs767443405 Chr.22: 19132507-19132507 on Build GRCh38
rs189384018 Chr.22: 19132447-19132447 on Build GRCh38
rs573495157 Chr.22: 19132510-19132510 on Build GRCh38
rs754653100 Chr.22: 19132514-19132514 on Build GRCh38
rs750098431 Chr.22: 19132437-19132437 on Build GRCh38
rs757802014 Chr.22: 19132527-19132527 on Build GRCh38
rs760659207 Chr.22: 19132393-19132393 on Build GRCh38
rs766637203 Chr.22: 19132397-19132397 on Build GRCh38
rs573739758 Chr.22: 19132477-19132477 on Build GRCh38
rs776936568 Chr.22: 19132478-19132478 on Build GRCh38
rs753942362 Chr.22: 19132398-19132398 on Build GRCh38
rs775019033 Chr.22: 19132381-19132381 on Build GRCh38
rs768106552 Chr.22: 19132387-19132387 on Build GRCh38
rs185310637 Chr.22: 19132312-19132312 on Build GRCh38
rs201838544 Chr.22: 19132511-19132511 on Build GRCh38
rs989638459 Chr.22: 19132346-19132346 on Build GRCh38
rs1052773 Chr.22: 19132425-19132425 on Build GRCh38
rs757036434 Chr.22: 19132499-19132499 on Build GRCh38
rs748537405 Chr.22: 19132518-19132518 on Build GRCh38
rs769560398 Chr.22: 19132364-19132364 on Build GRCh38
rs375084328 Chr.22: 19132526-19132526 on Build GRCh38
rs777385407 Chr.22: 19132413-19132413 on Build GRCh38
rs150037057 Chr.22: 19132320-19132320 on Build GRCh38
rs17853776 Chr.22: 19132310-19132310 on Build GRCh38
rs549857607 Chr.22: 19132311-19132311 on Build GRCh38
rs773798571 Chr.22: 19132464-19132464 on Build GRCh38
rs376847235 Chr.22: 19132386-19132386 on Build GRCh38
rs140125132 Chr.22: 19132333-19132333 on Build GRCh38
rs772497881 Chr.22: 19132522-19132522 on Build GRCh38
rs750671644 Chr.22: 19132516-19132517 on Build GRCh38
rs770453644 Chr.22: 19132332-19132332 on Build GRCh38
rs143749090 Chr.22: 19132371-19132371 on Build GRCh38
rs373520876 Chr.22: 19132493-19132493 on Build GRCh38
rs748715816 Chr.22: 19132368-19132368 on Build GRCh38
rs756444543 Chr.22: 19132313-19132313 on Build GRCh38
rs375018282 Chr.22: 19132492-19132492 on Build GRCh38
rs145304535 Chr.22: 19132323-19132323 on Build GRCh38
rs906526136 Chr.22: 19132485-19132485 on Build GRCh38

View Full Product Details