Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219539475-219539716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPF
Entrez Gene ID
79586
Gene Name
chondroitin polymerizing factor
Gene Aliases
CHSY2, CSS2
UniGene
Hs.516711
Gene Chromosome Location
Chr.2: 219538947-219543765 on Build GRCh38
Transcripts
NM_001195731.1 NM_024536.5 XM_011511838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4593499 c.2072T>G p.L691R Chr.2: 219539639-219539639 on Build GRCh38
4591232 c.2077G>C p.A693P Chr.2: 219539634-219539634 on Build GRCh38
6628555 c.2226G>T p.E742D Chr.2: 219539485-219539485 on Build GRCh38
4672208 c.2161C>T p.R721W Chr.2: 219539550-219539550 on Build GRCh38
6311660 c.2165C>A p.A722E Chr.2: 219539546-219539546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199953235 Chr.2: 219539586-219539586 on Build GRCh38
rs559473455 Chr.2: 219539550-219539550 on Build GRCh38
rs757492342 Chr.2: 219539658-219539658 on Build GRCh38
rs372800920 Chr.2: 219539648-219539648 on Build GRCh38
rs767271435 Chr.2: 219539611-219539611 on Build GRCh38
rs148492233 Chr.2: 219539551-219539551 on Build GRCh38
rs201631130 Chr.2: 219539573-219539573 on Build GRCh38
rs746178580 Chr.2: 219539679-219539679 on Build GRCh38
rs149014949 Chr.2: 219539616-219539616 on Build GRCh38
rs545710569 Chr.2: 219539546-219539546 on Build GRCh38
rs760527513 Chr.2: 219539640-219539640 on Build GRCh38
rs368474650 Chr.2: 219539553-219539553 on Build GRCh38
rs756522803 Chr.2: 219539682-219539682 on Build GRCh38
rs770128957 Chr.2: 219539624-219539624 on Build GRCh38
rs576858390 Chr.2: 219539530-219539530 on Build GRCh38
rs746254529 Chr.2: 219539623-219539623 on Build GRCh38
rs778096039 Chr.2: 219539618-219539618 on Build GRCh38
rs137926358 Chr.2: 219539506-219539506 on Build GRCh38
rs369808731 Chr.2: 219539649-219539649 on Build GRCh38
rs755993200 Chr.2: 219539613-219539613 on Build GRCh38
rs746345380 Chr.2: 219539590-219539590 on Build GRCh38
rs751743548 Chr.2: 219539654-219539654 on Build GRCh38
rs764619083 Chr.2: 219539516-219539516 on Build GRCh38
rs148389410 Chr.2: 219539499-219539499 on Build GRCh38
rs372387019 Chr.2: 219539498-219539498 on Build GRCh38
rs767216542 Chr.2: 219539636-219539636 on Build GRCh38
rs76494878 Chr.2: 219539522-219539522 on Build GRCh38
rs143397847 Chr.2: 219539497-219539497 on Build GRCh38
rs750337404 Chr.2: 219539533-219539533 on Build GRCh38
rs759070913 Chr.2: 219539512-219539512 on Build GRCh38
rs114128742 Chr.2: 219539614-219539614 on Build GRCh38
rs774801501 Chr.2: 219539566-219539566 on Build GRCh38
rs769336758 Chr.2: 219539611-219539613 on Build GRCh38
rs375153120 Chr.2: 219539545-219539545 on Build GRCh38
rs761137866 Chr.2: 219539500-219539500 on Build GRCh38
rs374210342 Chr.2: 219539650-219539650 on Build GRCh38
rs751234187 Chr.2: 219539581-219539581 on Build GRCh38
rs766190405 Chr.2: 219539642-219539642 on Build GRCh38
rs768130754 Chr.2: 219539634-219539634 on Build GRCh38
rs750286550 Chr.2: 219539612-219539612 on Build GRCh38
rs777301999 Chr.2: 219539634-219539639 on Build GRCh38
rs778149340 Chr.2: 219539549-219539549 on Build GRCh38
rs751287769 Chr.2: 219539523-219539523 on Build GRCh38
rs371656584 Chr.2: 219539536-219539536 on Build GRCh38
rs149541521 Chr.2: 219539686-219539686 on Build GRCh38
rs777312208 Chr.2: 219539507-219539507 on Build GRCh38
rs781677817 Chr.2: 219539621-219539621 on Build GRCh38
rs769220554 Chr.2: 219539699-219539699 on Build GRCh38
rs757074924 Chr.2: 219539527-219539527 on Build GRCh38
rs143750392 Chr.2: 219539619-219539619 on Build GRCh38
rs749769051 Chr.2: 219539693-219539693 on Build GRCh38
rs145929912 Chr.2: 219539517-219539517 on Build GRCh38
rs142415191 Chr.2: 219539625-219539625 on Build GRCh38
rs1976616 Chr.2: 219539659-219539659 on Build GRCh38
rs376924121 Chr.2: 219539643-219539643 on Build GRCh38
rs773889943 Chr.2: 219539635-219539635 on Build GRCh38
rs145461090 Chr.2: 219539577-219539577 on Build GRCh38
rs150991113 Chr.2: 219539565-219539565 on Build GRCh38

View Full Product Details