Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115984481-115984754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LVRN
Entrez Gene ID
206338
Gene Name
laeverin
Gene Aliases
APQ, AQPEP, TAQPEP
UniGene
Hs.556880
Gene Chromosome Location
Chr.5: 115962440-116027602 on Build GRCh38
Transcripts
NM_173800.4 XM_011543263.2 XM_011543264.2 XM_017009215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1432608 c.875G>T p.W292L Chr.5: 115984606-115984606 on Build GRCh38
5693718 c.886A>G p.T296A Chr.5: 115984617-115984617 on Build GRCh38
1696065 c.875G>A p.W292* Chr.5: 115984606-115984606 on Build GRCh38
1059881 c.928T>A p.F310I Chr.5: 115984659-115984659 on Build GRCh38
1059880 c.924C>T p.V308V Chr.5: 115984655-115984655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376871499 Chr.5: 115984539-115984539 on Build GRCh38
rs760010960 Chr.5: 115984621-115984621 on Build GRCh38
rs536743494 Chr.5: 115984543-115984543 on Build GRCh38
rs760740981 Chr.5: 115984611-115984611 on Build GRCh38
rs773230435 Chr.5: 115984597-115984597 on Build GRCh38
rs895507633 Chr.5: 115984702-115984702 on Build GRCh38
rs78947008 Chr.5: 115984518-115984518 on Build GRCh38
rs752358149 Chr.5: 115984531-115984531 on Build GRCh38
rs763346153 Chr.5: 115984685-115984685 on Build GRCh38
rs533605617 Chr.5: 115984549-115984552 on Build GRCh38
rs369036593 Chr.5: 115984545-115984545 on Build GRCh38
rs576367558 Chr.5: 115984631-115984631 on Build GRCh38
rs746784058 Chr.5: 115984540-115984540 on Build GRCh38
rs377469881 Chr.5: 115984656-115984656 on Build GRCh38
rs773425570 Chr.5: 115984666-115984666 on Build GRCh38
rs760170484 Chr.5: 115984682-115984682 on Build GRCh38
rs776542896 Chr.5: 115984680-115984680 on Build GRCh38
rs137960407 Chr.5: 115984658-115984658 on Build GRCh38
rs750451334 Chr.5: 115984655-115984655 on Build GRCh38
rs767100762 Chr.5: 115984708-115984708 on Build GRCh38
rs372673653 Chr.5: 115984561-115984561 on Build GRCh38
rs879121173 Chr.5: 115984601-115984601 on Build GRCh38
rs763801056 Chr.5: 115984633-115984633 on Build GRCh38
rs765739446 Chr.5: 115984623-115984623 on Build GRCh38
rs769533542 Chr.5: 115984564-115984564 on Build GRCh38
rs756738334 Chr.5: 115984641-115984641 on Build GRCh38
rs763671464 Chr.5: 115984686-115984686 on Build GRCh38
rs762126747 Chr.5: 115984630-115984630 on Build GRCh38
rs761072323 Chr.5: 115984537-115984537 on Build GRCh38
rs781019454 Chr.5: 115984650-115984650 on Build GRCh38
rs909782260 Chr.5: 115984693-115984693 on Build GRCh38
rs553964940 Chr.5: 115984503-115984503 on Build GRCh38
rs149510341 Chr.5: 115984662-115984662 on Build GRCh38
rs747081515 Chr.5: 115984672-115984672 on Build GRCh38
rs373229835 Chr.5: 115984726-115984726 on Build GRCh38
rs769013775 Chr.5: 115984596-115984596 on Build GRCh38
rs751179688 Chr.5: 115984688-115984688 on Build GRCh38
rs983564219 Chr.5: 115984639-115984639 on Build GRCh38
rs749076159 Chr.5: 115984659-115984659 on Build GRCh38
rs776930188 Chr.5: 115984627-115984627 on Build GRCh38
rs865875031 Chr.5: 115984707-115984707 on Build GRCh38
rs775231940 Chr.5: 115984566-115984566 on Build GRCh38
rs17482536 Chr.5: 115984673-115984673 on Build GRCh38
rs758093708 Chr.5: 115984535-115984535 on Build GRCh38
rs756825113 Chr.5: 115984698-115984698 on Build GRCh38
rs766810003 Chr.5: 115984615-115984615 on Build GRCh38
rs779898106 Chr.5: 115984725-115984725 on Build GRCh38
rs146224958 Chr.5: 115984636-115984636 on Build GRCh38
rs564955632 Chr.5: 115984683-115984683 on Build GRCh38
rs572357148 Chr.5: 115984718-115984718 on Build GRCh38
rs373118440 Chr.5: 115984716-115984716 on Build GRCh38
rs965333283 Chr.5: 115984614-115984614 on Build GRCh38
rs762068329 Chr.5: 115984587-115984587 on Build GRCh38
rs778730062 Chr.5: 115984522-115984522 on Build GRCh38

View Full Product Details