Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75042366-75042621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5H
Entrez Gene ID
10476
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit D
Gene Aliases
ATPQ
UniGene
Hs.514465
Gene Chromosome Location
Chr.17: 75038860-75046979 on Build GRCh38
Transcripts
NM_001003785.1 NM_006356.2 NR_110835.1

Gene Symbol
KCTD2
Entrez Gene ID
23510
Gene Name
potassium channel tetramerization domain containing 2
Gene Aliases
-
UniGene
Hs.514468
Gene Chromosome Location
Chr.17: 75032567-75065889 on Build GRCh38
Transcripts
NM_001003785.1 NM_006356.2 NR_110835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1385906 c.99C>T p.S33S Chr.17: 75042552-75042552 on Build GRCh38
6607959 c.75G>C p.K25N Chr.17: 75042576-75042576 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746947469 Chr.17: 75042524-75042524 on Build GRCh38
rs987660541 Chr.17: 75042528-75042528 on Build GRCh38
rs373552679 Chr.17: 75042494-75042494 on Build GRCh38
rs951320226 Chr.17: 75042388-75042388 on Build GRCh38
rs902581826 Chr.17: 75042562-75042562 on Build GRCh38
rs1036045441 Chr.17: 75042588-75042588 on Build GRCh38
rs1055055790 Chr.17: 75042456-75042456 on Build GRCh38
rs968970953 Chr.17: 75042405-75042405 on Build GRCh38
rs543711750 Chr.17: 75042470-75042470 on Build GRCh38
rs574600761 Chr.17: 75042440-75042440 on Build GRCh38
rs755628833 Chr.17: 75042488-75042488 on Build GRCh38
rs767846385 Chr.17: 75042596-75042596 on Build GRCh38
rs746986507 Chr.17: 75042403-75042403 on Build GRCh38
rs139430113 Chr.17: 75042515-75042515 on Build GRCh38
rs112922015 Chr.17: 75042540-75042540 on Build GRCh38
rs978994318 Chr.17: 75042482-75042482 on Build GRCh38
rs139761287 Chr.17: 75042537-75042537 on Build GRCh38
rs557017001 Chr.17: 75042508-75042508 on Build GRCh38
rs768434668 Chr.17: 75042526-75042526 on Build GRCh38
rs749305952 Chr.17: 75042578-75042578 on Build GRCh38
rs558986152 Chr.17: 75042593-75042593 on Build GRCh38
rs757685789 Chr.17: 75042529-75042529 on Build GRCh38
rs916551971 Chr.17: 75042477-75042477 on Build GRCh38
rs1020683067 Chr.17: 75042389-75042389 on Build GRCh38
rs745666457 Chr.17: 75042510-75042510 on Build GRCh38
rs759950428 Chr.17: 75042594-75042594 on Build GRCh38
rs557831541 Chr.17: 75042464-75042464 on Build GRCh38
rs554827656 Chr.17: 75042435-75042435 on Build GRCh38
rs200410651 Chr.17: 75042572-75042572 on Build GRCh38
rs909843303 Chr.17: 75042427-75042427 on Build GRCh38
rs545514326 Chr.17: 75042592-75042592 on Build GRCh38
rs779917999 Chr.17: 75042520-75042520 on Build GRCh38

View Full Product Details