Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52646308-52646541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERO1A
Entrez Gene ID
30001
Gene Name
endoplasmic reticulum oxidoreductase 1 alpha
Gene Aliases
ERO1-L, ERO1-L-alpha, ERO1-alpha, ERO1L, ERO1LA, Ero1alpha
UniGene
Hs.525339
Gene Chromosome Location
Chr.14: 52639915-52695931 on Build GRCh38
Transcripts
NM_014584.2 XM_011536684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1370107 c.1163T>C p.I388T Chr.14: 52646424-52646424 on Build GRCh38
6656252 c.1135C>T p.R379* Chr.14: 52646452-52646452 on Build GRCh38
7297855 c.1162A>G p.I388V Chr.14: 52646425-52646425 on Build GRCh38
5916351 c.1126G>A p.E376K Chr.14: 52646461-52646461 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1048560505 Chr.14: 52646452-52646452 on Build GRCh38
rs772381008 Chr.14: 52646512-52646512 on Build GRCh38
rs780920634 Chr.14: 52646432-52646432 on Build GRCh38
rs372188484 Chr.14: 52646369-52646369 on Build GRCh38
rs774350848 Chr.14: 52646451-52646451 on Build GRCh38
rs749508867 Chr.14: 52646436-52646436 on Build GRCh38
rs749937959 Chr.14: 52646374-52646374 on Build GRCh38
rs895368671 Chr.14: 52646490-52646490 on Build GRCh38
rs756903858 Chr.14: 52646367-52646367 on Build GRCh38
rs772225040 Chr.14: 52646499-52646499 on Build GRCh38
rs369263722 Chr.14: 52646493-52646493 on Build GRCh38
rs758333053 Chr.14: 52646395-52646395 on Build GRCh38
rs369445375 Chr.14: 52646344-52646344 on Build GRCh38
rs1046380306 Chr.14: 52646508-52646508 on Build GRCh38
rs776166315 Chr.14: 52646500-52646500 on Build GRCh38
rs771159771 Chr.14: 52646440-52646440 on Build GRCh38
rs768461835 Chr.14: 52646418-52646418 on Build GRCh38
rs759943068 Chr.14: 52646365-52646365 on Build GRCh38
rs746890742 Chr.14: 52646409-52646409 on Build GRCh38
rs545816292 Chr.14: 52646355-52646355 on Build GRCh38
rs146578543 Chr.14: 52646402-52646402 on Build GRCh38
rs565402634 Chr.14: 52646469-52646469 on Build GRCh38
rs375367535 Chr.14: 52646511-52646511 on Build GRCh38

View Full Product Details