Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1375370-1375643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP1A
Entrez Gene ID
2280
Gene Name
FK506 binding protein 1A
Gene Aliases
FKBP-12, FKBP-1A, FKBP1, FKBP12, PKC12, PKCI2, PPIASE
UniGene
Hs.471933
Gene Chromosome Location
Chr.20: 1368977-1393172 on Build GRCh38
Transcripts
NM_000801.4 NM_001199786.1 NM_054014.3 NR_037661.1 NR_040047.1 NR_040048.1 NR_040049.1

Gene Symbol
FKBP1A-SDCBP2
Entrez Gene ID
100528031
Gene Name
FKBP1A-SDCBP2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.471933
Gene Chromosome Location
Chr.20: 1309909-1393172 on Build GRCh38
Transcripts
NM_000801.4 NM_001199786.1 NM_054014.3 NR_037661.1 NR_040047.1 NR_040048.1 NR_040049.1

Gene Symbol
SDCBP2-AS1
Entrez Gene ID
100507495
Gene Name
SDCBP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655785
Gene Chromosome Location
Chr.20: 1325343-1378735 on Build GRCh38
Transcripts
NM_000801.4 NM_001199786.1 NM_054014.3 NR_037661.1 NR_040047.1 NR_040048.1 NR_040049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
302241 c.111_112insTT p.D38fs*15 Chr.20: 1375577-1375578 on Build GRCh38
7097344 c.105G>T p.K35N Chr.20: 1375584-1375584 on Build GRCh38
6582495 c.141T>G p.F47L Chr.20: 1375548-1375548 on Build GRCh38
1565903 c.116C>G p.S39C Chr.20: 1375573-1375573 on Build GRCh38
6558501 c.121C>A p.R41R Chr.20: 1375568-1375568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056771956 Chr.20: 1375392-1375392 on Build GRCh38
rs772970172 Chr.20: 1375467-1375467 on Build GRCh38
rs771902148 Chr.20: 1375450-1375450 on Build GRCh38
rs753166355 Chr.20: 1375590-1375590 on Build GRCh38
rs1033273268 Chr.20: 1375423-1375423 on Build GRCh38
rs760364468 Chr.20: 1375472-1375472 on Build GRCh38
rs764552263 Chr.20: 1375500-1375500 on Build GRCh38
rs1022676887 Chr.20: 1375433-1375434 on Build GRCh38
rs763193816 Chr.20: 1375611-1375611 on Build GRCh38
rs572399308 Chr.20: 1375408-1375408 on Build GRCh38
rs765660953 Chr.20: 1375572-1375572 on Build GRCh38
rs6074549 Chr.20: 1375617-1375617 on Build GRCh38
rs1001813719 Chr.20: 1375418-1375418 on Build GRCh38
rs770587902 Chr.20: 1375474-1375474 on Build GRCh38
rs968850297 Chr.20: 1375449-1375449 on Build GRCh38
rs758888107 Chr.20: 1375415-1375415 on Build GRCh38
rs138988463 Chr.20: 1375491-1375491 on Build GRCh38
rs1000358406 Chr.20: 1375504-1375504 on Build GRCh38
rs1020333722 Chr.20: 1375602-1375602 on Build GRCh38
rs778119565 Chr.20: 1375557-1375557 on Build GRCh38
rs747880277 Chr.20: 1375442-1375442 on Build GRCh38
rs776227831 Chr.20: 1375477-1375477 on Build GRCh38
rs760096309 Chr.20: 1375568-1375568 on Build GRCh38
rs541423172 Chr.20: 1375424-1375424 on Build GRCh38

View Full Product Details