Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23359347-23359588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFS
Entrez Gene ID
10278
Gene Name
embryonal Fyn-associated substrate
Gene Aliases
CAS3, CASS3, EFS1, EFS2, HEFS, SIN
UniGene
Hs.24587
Gene Chromosome Location
Chr.14: 23356400-23365633 on Build GRCh38
Transcripts
NM_001277174.1 NM_005864.3 NM_032459.2 XM_005267256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6388458 c.1031T>C p.L344P Chr.14: 23359447-23359447 on Build GRCh38
7333982 c.1110T>C p.N370N Chr.14: 23359368-23359368 on Build GRCh38
1250992 c.1127C>T p.P376L Chr.14: 23359351-23359351 on Build GRCh38
6384850 c.994A>G p.I332V Chr.14: 23359484-23359484 on Build GRCh38
6400948 c.955T>C p.S319P Chr.14: 23359523-23359523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147068550 Chr.14: 23359475-23359475 on Build GRCh38
rs752295814 Chr.14: 23359438-23359438 on Build GRCh38
rs1051062711 Chr.14: 23359427-23359427 on Build GRCh38
rs113470967 Chr.14: 23359495-23359495 on Build GRCh38
rs758684896 Chr.14: 23359556-23359556 on Build GRCh38
rs774219426 Chr.14: 23359421-23359421 on Build GRCh38
rs779278865 Chr.14: 23359455-23359455 on Build GRCh38
rs978564499 Chr.14: 23359433-23359433 on Build GRCh38
rs760603122 Chr.14: 23359504-23359504 on Build GRCh38
rs759395794 Chr.14: 23359450-23359451 on Build GRCh38
rs766964133 Chr.14: 23359564-23359564 on Build GRCh38
rs763601003 Chr.14: 23359532-23359532 on Build GRCh38
rs750735299 Chr.14: 23359461-23359461 on Build GRCh38
rs776080727 Chr.14: 23359542-23359542 on Build GRCh38
rs773265649 Chr.14: 23359428-23359428 on Build GRCh38
rs866529859 Chr.14: 23359507-23359508 on Build GRCh38
rs533627167 Chr.14: 23359567-23359567 on Build GRCh38
rs764846330 Chr.14: 23359550-23359550 on Build GRCh38
rs567919656 Chr.14: 23359502-23359502 on Build GRCh38
rs770095379 Chr.14: 23359481-23359481 on Build GRCh38
rs200446928 Chr.14: 23359451-23359451 on Build GRCh38
rs763795252 Chr.14: 23359446-23359446 on Build GRCh38
rs750839916 Chr.14: 23359551-23359551 on Build GRCh38
rs768961635 Chr.14: 23359392-23359392 on Build GRCh38
rs772118374 Chr.14: 23359496-23359496 on Build GRCh38
rs776985217 Chr.14: 23359397-23359397 on Build GRCh38
rs759542910 Chr.14: 23359422-23359422 on Build GRCh38
rs747177013 Chr.14: 23359471-23359471 on Build GRCh38
rs780202633 Chr.14: 23359388-23359388 on Build GRCh38
rs770860058 Chr.14: 23359405-23359405 on Build GRCh38
rs762895299 Chr.14: 23359404-23359404 on Build GRCh38
rs779192810 Chr.14: 23359369-23359369 on Build GRCh38
rs891095397 Chr.14: 23359428-23359428 on Build GRCh38
rs531521647 Chr.14: 23359381-23359381 on Build GRCh38
rs761476214 Chr.14: 23359544-23359544 on Build GRCh38
rs753502762 Chr.14: 23359450-23359450 on Build GRCh38
rs1030254656 Chr.14: 23359382-23359382 on Build GRCh38
rs781450318 Chr.14: 23359568-23359568 on Build GRCh38
rs780393962 Chr.14: 23359465-23359465 on Build GRCh38
rs746379952 Chr.14: 23359379-23359379 on Build GRCh38
rs760103137 Chr.14: 23359441-23359441 on Build GRCh38
rs768600416 Chr.14: 23359474-23359474 on Build GRCh38
rs747036747 Chr.14: 23359390-23359390 on Build GRCh38
rs141615199 Chr.14: 23359432-23359432 on Build GRCh38

View Full Product Details