Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55726866-55727101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD63
Entrez Gene ID
967
Gene Name
CD63 molecule
Gene Aliases
LAMP-3, ME491, MLA1, OMA81H, TSPAN30
UniGene
Hs.445570
Gene Chromosome Location
Chr.12: 55725443-55729673 on Build GRCh38
Transcripts
NM_001257389.1 NM_001257390.1 NM_001257391.1 NM_001257392.1 NM_001257400.1 NM_001257401.1 NM_001267698.1 NM_001780.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7211727 c.330+9G>T p.? Chr.12: 55726881-55726881 on Build GRCh38
5521578 c.323G>A p.R108K Chr.12: 55726897-55726897 on Build GRCh38
6871268 c.261C>T p.A87A Chr.12: 55726959-55726959 on Build GRCh38
6623864 c.263T>A p.I88N Chr.12: 55726957-55726957 on Build GRCh38
4043370 c.298G>A p.A100T Chr.12: 55726922-55726922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769609977 Chr.12: 55726974-55726974 on Build GRCh38
rs772258 Chr.12: 55726984-55726984 on Build GRCh38
rs776564735 Chr.12: 55726960-55726960 on Build GRCh38
rs143228419 Chr.12: 55727025-55727026 on Build GRCh38
rs766792430 Chr.12: 55727037-55727037 on Build GRCh38
rs758273029 Chr.12: 55726945-55726946 on Build GRCh38
rs576524816 Chr.12: 55726924-55726924 on Build GRCh38
rs963772710 Chr.12: 55726914-55726914 on Build GRCh38
rs528587978 Chr.12: 55727054-55727054 on Build GRCh38
rs941450244 Chr.12: 55727072-55727072 on Build GRCh38
rs113373490 Chr.12: 55726939-55726939 on Build GRCh38
rs564617994 Chr.12: 55727031-55727031 on Build GRCh38
rs11542876 Chr.12: 55726901-55726901 on Build GRCh38
rs1017688965 Chr.12: 55726909-55726909 on Build GRCh38
rs888530842 Chr.12: 55727067-55727067 on Build GRCh38
rs747128338 Chr.12: 55726950-55726950 on Build GRCh38
rs531047095 Chr.12: 55727019-55727019 on Build GRCh38
rs372890146 Chr.12: 55727016-55727016 on Build GRCh38
rs779232780 Chr.12: 55726929-55726929 on Build GRCh38
rs756637406 Chr.12: 55726922-55726922 on Build GRCh38
rs751088242 Chr.12: 55726916-55726916 on Build GRCh38
rs780428841 Chr.12: 55726923-55726923 on Build GRCh38
rs942126683 Chr.12: 55727068-55727068 on Build GRCh38
rs201460404 Chr.12: 55726942-55726942 on Build GRCh38
rs148781286 Chr.12: 55726956-55726956 on Build GRCh38
rs374822776 Chr.12: 55726999-55726999 on Build GRCh38
rs755413075 Chr.12: 55726925-55726925 on Build GRCh38
rs922849685 Chr.12: 55727024-55727024 on Build GRCh38
rs752369698 Chr.12: 55726895-55726895 on Build GRCh38
rs369287045 Chr.12: 55727015-55727015 on Build GRCh38
rs547028559 Chr.12: 55727073-55727073 on Build GRCh38
rs967432248 Chr.12: 55726958-55726958 on Build GRCh38
rs759278073 Chr.12: 55726973-55726973 on Build GRCh38
rs377098109 Chr.12: 55727022-55727022 on Build GRCh38
rs757864288 Chr.12: 55726899-55726899 on Build GRCh38
rs911416050 Chr.12: 55727006-55727006 on Build GRCh38
rs542128516 Chr.12: 55726992-55726992 on Build GRCh38
rs772236354 Chr.12: 55726936-55726936 on Build GRCh38
rs763637330 Chr.12: 55726905-55726905 on Build GRCh38
rs748345489 Chr.12: 55726933-55726933 on Build GRCh38
rs762652591 Chr.12: 55726985-55726985 on Build GRCh38
rs990298314 Chr.12: 55726940-55726940 on Build GRCh38

View Full Product Details