Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17696044-17696309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L13
Entrez Gene ID
23786
Gene Name
BCL2 like 13
Gene Aliases
BCL-RAMBO, Bcl2-L-13, MIL1
UniGene
Hs.631672
Gene Chromosome Location
Chr.22: 17628855-17730855 on Build GRCh38
Transcripts
NM_001270726.1 NM_001270727.1 NM_001270728.1 NM_001270729.1 NM_001270730.1 NM_001270731.1 NM_001270732.1 NM_001270734.1 NM_001270735.1 NM_015367.3 NR_073068.1 NR_073069.1 XM_011546119.1 XM_011546120.1 XM_011546121.2 XM_011546123.1 XM_017028725.1 XM_017028726.1 XM_017028727.1 XM_017028728.1 XM_017028729.1 XM_017028730.1 XM_017028731.1 XM_017028732.1 XM_017028733.1 XM_017028734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5109438 c.401A>C p.Q134P Chr.22: 17696155-17696155 on Build GRCh38
725655 c.409C>T p.R137W Chr.22: 17696163-17696163 on Build GRCh38
5749273 c.451A>G p.N151D Chr.22: 17696205-17696205 on Build GRCh38
5780390 c.400C>G p.Q134E Chr.22: 17696154-17696154 on Build GRCh38
2152258 c.428C>T p.T143I Chr.22: 17696182-17696182 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773293223 Chr.22: 17696157-17696157 on Build GRCh38
rs538599427 Chr.22: 17696230-17696230 on Build GRCh38
rs746423100 Chr.22: 17696213-17696215 on Build GRCh38
rs924951072 Chr.22: 17696263-17696263 on Build GRCh38
rs758988570 Chr.22: 17696184-17696184 on Build GRCh38
rs772176474 Chr.22: 17696227-17696227 on Build GRCh38
rs772845337 Chr.22: 17696242-17696242 on Build GRCh38
rs761841738 Chr.22: 17696144-17696144 on Build GRCh38
rs111621210 Chr.22: 17696072-17696072 on Build GRCh38
rs755242901 Chr.22: 17696168-17696168 on Build GRCh38
rs771179424 Chr.22: 17696105-17696105 on Build GRCh38
rs954051851 Chr.22: 17696132-17696133 on Build GRCh38
rs774217870 Chr.22: 17696218-17696218 on Build GRCh38
rs762793127 Chr.22: 17696116-17696116 on Build GRCh38
rs777094382 Chr.22: 17696124-17696125 on Build GRCh38
rs754244583 Chr.22: 17696164-17696164 on Build GRCh38
rs775258647 Chr.22: 17696110-17696110 on Build GRCh38
rs779343604 Chr.22: 17696169-17696169 on Build GRCh38
rs766203253 Chr.22: 17696163-17696163 on Build GRCh38
rs141842486 Chr.22: 17696142-17696142 on Build GRCh38
rs561162060 Chr.22: 17696091-17696091 on Build GRCh38
rs140799351 Chr.22: 17696147-17696147 on Build GRCh38
rs376057310 Chr.22: 17696265-17696265 on Build GRCh38
rs552161262 Chr.22: 17696250-17696250 on Build GRCh38
rs577486545 Chr.22: 17696199-17696199 on Build GRCh38
rs746105939 Chr.22: 17696205-17696205 on Build GRCh38
rs2257083 Chr.22: 17696093-17696093 on Build GRCh38
rs753108863 Chr.22: 17696172-17696172 on Build GRCh38
rs781332681 Chr.22: 17696200-17696200 on Build GRCh38
rs1030186506 Chr.22: 17696112-17696112 on Build GRCh38
rs566747222 Chr.22: 17696215-17696215 on Build GRCh38
rs747156111 Chr.22: 17696198-17696198 on Build GRCh38
rs190897414 Chr.22: 17696070-17696070 on Build GRCh38
rs372300381 Chr.22: 17696134-17696134 on Build GRCh38
rs777795412 Chr.22: 17696191-17696191 on Build GRCh38
rs964639768 Chr.22: 17696074-17696074 on Build GRCh38
rs750003103 Chr.22: 17696151-17696151 on Build GRCh38
rs761633953 Chr.22: 17696224-17696224 on Build GRCh38

View Full Product Details