Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28758644-28758905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222B
Entrez Gene ID
55731
Gene Name
family with sequence similarity 222 member B
Gene Aliases
C17orf63
UniGene
Hs.564533
Gene Chromosome Location
Chr.17: 28755978-28855036 on Build GRCh38
Transcripts
NM_001077498.2 NM_001288631.1 NM_001288632.1 NM_001288633.1 NM_001288634.1 NM_001288635.1 NM_001288636.1 NM_001288637.1 NM_001288638.1 NM_001288639.1 NM_001288640.1 NM_018182.3 XM_011524986.2 XM_011524987.1 XM_011524988.2 XM_017024831.1 XM_017024832.1 XM_017024833.1 XM_017024834.1 XM_017024835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940688 c.1157C>T p.P386L Chr.17: 28758802-28758802 on Build GRCh38
6523396 c.1067A>T p.Y356F Chr.17: 28758892-28758892 on Build GRCh38
1381760 c.1297C>A p.P433T Chr.17: 28758662-28758662 on Build GRCh38
1381759 c.1297C>A p.P433T Chr.17: 28758662-28758662 on Build GRCh38
1381758 c.1300C>T p.P434S Chr.17: 28758659-28758659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755676003 Chr.17: 28758696-28758696 on Build GRCh38
rs932558506 Chr.17: 28758861-28758861 on Build GRCh38
rs770999851 Chr.17: 28758750-28758750 on Build GRCh38
rs375781000 Chr.17: 28758727-28758727 on Build GRCh38
rs781545036 Chr.17: 28758753-28758753 on Build GRCh38
rs1040067918 Chr.17: 28758784-28758784 on Build GRCh38
rs34269353 Chr.17: 28758821-28758821 on Build GRCh38
rs921222922 Chr.17: 28758855-28758855 on Build GRCh38
rs758234852 Chr.17: 28758726-28758726 on Build GRCh38
rs748141075 Chr.17: 28758722-28758722 on Build GRCh38
rs769944568 Chr.17: 28758836-28758836 on Build GRCh38
rs751956402 Chr.17: 28758683-28758683 on Build GRCh38
rs973773492 Chr.17: 28758741-28758741 on Build GRCh38
rs561518046 Chr.17: 28758776-28758776 on Build GRCh38
rs368465145 Chr.17: 28758842-28758842 on Build GRCh38
rs921911180 Chr.17: 28758801-28758801 on Build GRCh38
rs35911469 Chr.17: 28758815-28758816 on Build GRCh38
rs775945857 Chr.17: 28758772-28758772 on Build GRCh38
rs372776564 Chr.17: 28758672-28758672 on Build GRCh38
rs776035748 Chr.17: 28758669-28758669 on Build GRCh38
rs201771310 Chr.17: 28758661-28758661 on Build GRCh38
rs753217058 Chr.17: 28758709-28758709 on Build GRCh38
rs76905501 Chr.17: 28758708-28758708 on Build GRCh38
rs372142554 Chr.17: 28758884-28758884 on Build GRCh38
rs746142410 Chr.17: 28758825-28758825 on Build GRCh38
rs774643433 Chr.17: 28758780-28758780 on Build GRCh38
rs199606408 Chr.17: 28758739-28758739 on Build GRCh38
rs374755194 Chr.17: 28758770-28758770 on Build GRCh38
rs777864354 Chr.17: 28758814-28758814 on Build GRCh38
rs34697365 Chr.17: 28758810-28758810 on Build GRCh38
rs774902821 Chr.17: 28758869-28758869 on Build GRCh38
rs758943759 Chr.17: 28758793-28758793 on Build GRCh38
rs935445485 Chr.17: 28758871-28758871 on Build GRCh38
rs918350186 Chr.17: 28758744-28758744 on Build GRCh38
rs781248991 Chr.17: 28758824-28758824 on Build GRCh38
rs769966107 Chr.17: 28758771-28758771 on Build GRCh38
rs758359508 Chr.17: 28758811-28758811 on Build GRCh38
rs763413670 Chr.17: 28758773-28758773 on Build GRCh38
rs772532734 Chr.17: 28758885-28758885 on Build GRCh38
rs764486649 Chr.17: 28758673-28758673 on Build GRCh38
rs201236089 Chr.17: 28758837-28758837 on Build GRCh38
rs765940574 Chr.17: 28758782-28758782 on Build GRCh38
rs778976958 Chr.17: 28758715-28758715 on Build GRCh38
rs528103505 Chr.17: 28758790-28758790 on Build GRCh38
rs754580506 Chr.17: 28758710-28758710 on Build GRCh38

View Full Product Details