Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78110960-78111221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB2
Entrez Gene ID
10518
Gene Name
calcium and integrin binding family member 2
Gene Aliases
DFNB48, KIP2, USH1J
UniGene
Hs.129867
Gene Chromosome Location
Chr.15: 78104606-78131976 on Build GRCh38
Transcripts
NM_001271888.1 NM_001271889.1 NM_001301224.1 NM_006383.3 NR_125435.1 XM_005254126.3 XM_006720374.2 XM_011521161.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5460420 c.197G>A p.R66Q Chr.15: 78111166-78111166 on Build GRCh38
1201195 c.145A>G p.S49G Chr.15: 78111218-78111218 on Build GRCh38
3504272 c.153C>T p.I51I Chr.15: 78111210-78111210 on Build GRCh38
1374756 c.198+2T>C p.? Chr.15: 78111163-78111163 on Build GRCh38
4128428 c.192G>A p.E64E Chr.15: 78111171-78111171 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756326738 Chr.15: 78111147-78111147 on Build GRCh38
rs763437985 Chr.15: 78111117-78111117 on Build GRCh38
rs568524396 Chr.15: 78111047-78111047 on Build GRCh38
rs746831663 Chr.15: 78111164-78111164 on Build GRCh38
rs766159119 Chr.15: 78111134-78111134 on Build GRCh38
rs998218275 Chr.15: 78111065-78111065 on Build GRCh38
rs560859905 Chr.15: 78111197-78111197 on Build GRCh38
rs780951279 Chr.15: 78111077-78111077 on Build GRCh38
rs779074778 Chr.15: 78111190-78111190 on Build GRCh38
rs769256040 Chr.15: 78111195-78111195 on Build GRCh38
rs142130860 Chr.15: 78111141-78111141 on Build GRCh38
rs777662280 Chr.15: 78111162-78111162 on Build GRCh38
rs535456308 Chr.15: 78111060-78111060 on Build GRCh38
rs200530151 Chr.15: 78111140-78111140 on Build GRCh38
rs147151784 Chr.15: 78111019-78111019 on Build GRCh38
rs767935017 Chr.15: 78111199-78111199 on Build GRCh38
rs145609090 Chr.15: 78110985-78110985 on Build GRCh38
rs749299160 Chr.15: 78111192-78111192 on Build GRCh38
rs980306994 Chr.15: 78111170-78111170 on Build GRCh38
rs775705454 Chr.15: 78111189-78111189 on Build GRCh38
rs753473721 Chr.15: 78111135-78111135 on Build GRCh38
rs145631140 Chr.15: 78111166-78111166 on Build GRCh38
rs1025314673 Chr.15: 78111102-78111102 on Build GRCh38
rs1015786154 Chr.15: 78111097-78111097 on Build GRCh38
rs145415848 Chr.15: 78111171-78111171 on Build GRCh38
rs752139872 Chr.15: 78111157-78111157 on Build GRCh38
rs371015361 Chr.15: 78111198-78111198 on Build GRCh38
rs751512423 Chr.15: 78111035-78111035 on Build GRCh38
rs757007957 Chr.15: 78111165-78111165 on Build GRCh38
rs2304827 Chr.15: 78111015-78111015 on Build GRCh38
rs764856250 Chr.15: 78111154-78111154 on Build GRCh38
rs140284593 Chr.15: 78111061-78111061 on Build GRCh38
rs780168150 Chr.15: 78111167-78111167 on Build GRCh38
rs36078965 Chr.15: 78111166-78111167 on Build GRCh38
rs201062535 Chr.15: 78111203-78111203 on Build GRCh38

View Full Product Details