Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109610370-109610629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT2
Entrez Gene ID
2780
Gene Name
G protein subunit alpha transducin 2
Gene Aliases
ACHM4, GNATC
UniGene
Hs.36973
Gene Chromosome Location
Chr.1: 109603091-109619733 on Build GRCh38
Transcripts
NM_005272.3 XM_011541264.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
295722 c.150C>A p.V50V Chr.1: 109610476-109610476 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs140603860 Chr.1: 109610464-109610464 on Build GRCh38
rs146945932 Chr.1: 109610479-109610479 on Build GRCh38
rs759025436 Chr.1: 109610542-109610542 on Build GRCh38
rs754951371 Chr.1: 109610432-109610432 on Build GRCh38
rs752437712 Chr.1: 109610556-109610556 on Build GRCh38
rs761577743 Chr.1: 109610494-109610494 on Build GRCh38
rs371566014 Chr.1: 109610431-109610431 on Build GRCh38
rs6658641 Chr.1: 109610576-109610576 on Build GRCh38
rs538720887 Chr.1: 109610407-109610407 on Build GRCh38
rs200725841 Chr.1: 109610484-109610484 on Build GRCh38
rs552327370 Chr.1: 109610409-109610409 on Build GRCh38
rs764383106 Chr.1: 109610548-109610550 on Build GRCh38
rs982782821 Chr.1: 109610485-109610485 on Build GRCh38
rs757745881 Chr.1: 109610429-109610429 on Build GRCh38
rs3738765 Chr.1: 109610399-109610399 on Build GRCh38
rs867004827 Chr.1: 109610467-109610467 on Build GRCh38
rs781755062 Chr.1: 109610443-109610443 on Build GRCh38
rs749334089 Chr.1: 109610478-109610478 on Build GRCh38
rs764959263 Chr.1: 109610544-109610544 on Build GRCh38
rs770225385 Chr.1: 109610469-109610469 on Build GRCh38
rs146606352 Chr.1: 109610487-109610487 on Build GRCh38
rs1046618076 Chr.1: 109610595-109610595 on Build GRCh38
rs746346813 Chr.1: 109610460-109610460 on Build GRCh38
rs910315717 Chr.1: 109610546-109610546 on Build GRCh38
rs773010453 Chr.1: 109610523-109610523 on Build GRCh38
rs752831057 Chr.1: 109610488-109610488 on Build GRCh38
rs538415975 Chr.1: 109610534-109610534 on Build GRCh38
rs201659866 Chr.1: 109610481-109610481 on Build GRCh38
rs369052204 Chr.1: 109610533-109610533 on Build GRCh38
rs753381086 Chr.1: 109610539-109610539 on Build GRCh38
rs747363906 Chr.1: 109610417-109610417 on Build GRCh38
rs767343349 Chr.1: 109610506-109610506 on Build GRCh38
rs775584517 Chr.1: 109610472-109610472 on Build GRCh38

View Full Product Details