Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123647738-123648011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H3
Entrez Gene ID
2967
Gene Name
general transcription factor IIH subunit 3
Gene Aliases
BTF2, P34, TFB4, TFIIH
UniGene
Hs.355348
Gene Chromosome Location
Chr.12: 123633739-123662604 on Build GRCh38
Transcripts
NM_001271866.1 NM_001271867.1 NM_001271868.1 NM_001516.4 XM_017019228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4689158 c.202C>T p.R68* Chr.12: 123647964-123647964 on Build GRCh38
4840693 c.246C>T p.F82F Chr.12: 123648008-123648008 on Build GRCh38
7139758 c.203G>T p.R68L Chr.12: 123647965-123647965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs531549382 Chr.12: 123647981-123647981 on Build GRCh38
rs928452443 Chr.12: 123647802-123647802 on Build GRCh38
rs908419796 Chr.12: 123647859-123647859 on Build GRCh38
rs559527771 Chr.12: 123647820-123647820 on Build GRCh38
rs564587002 Chr.12: 123647976-123647976 on Build GRCh38
rs931117643 Chr.12: 123647840-123647840 on Build GRCh38
rs748516267 Chr.12: 123647919-123647919 on Build GRCh38
rs773908995 Chr.12: 123647927-123647927 on Build GRCh38
rs552214076 Chr.12: 123647904-123647904 on Build GRCh38
rs920020258 Chr.12: 123647806-123647806 on Build GRCh38
rs879110011 Chr.12: 123647972-123647972 on Build GRCh38
rs915600200 Chr.12: 123647775-123647775 on Build GRCh38
rs763358730 Chr.12: 123647941-123647942 on Build GRCh38
rs749339917 Chr.12: 123647986-123647986 on Build GRCh38
rs188662652 Chr.12: 123647984-123647984 on Build GRCh38
rs576258085 Chr.12: 123647776-123647776 on Build GRCh38
rs544901331 Chr.12: 123647954-123647954 on Build GRCh38
rs757083365 Chr.12: 123647967-123647967 on Build GRCh38
rs185459622 Chr.12: 123647819-123647819 on Build GRCh38
rs567824997 Chr.12: 123647939-123647939 on Build GRCh38
rs762468878 Chr.12: 123647947-123647947 on Build GRCh38
rs982647567 Chr.12: 123647856-123647856 on Build GRCh38
rs11572959 Chr.12: 123647787-123647787 on Build GRCh38
rs755068042 Chr.12: 123647987-123647987 on Build GRCh38
rs78528979 Chr.12: 123647785-123647785 on Build GRCh38
rs774570423 Chr.12: 123647918-123647918 on Build GRCh38
rs937371320 Chr.12: 123647942-123647942 on Build GRCh38
rs768761972 Chr.12: 123647915-123647915 on Build GRCh38
rs564255687 Chr.12: 123647941-123647941 on Build GRCh38
rs759077449 Chr.12: 123647964-123647964 on Build GRCh38
rs1008477237 Chr.12: 123647866-123647866 on Build GRCh38
rs1041327978 Chr.12: 123647879-123647879 on Build GRCh38
rs766842302 Chr.12: 123647989-123647990 on Build GRCh38
rs561862982 Chr.12: 123647786-123647786 on Build GRCh38
rs777384756 Chr.12: 123647767-123647767 on Build GRCh38
rs1037092491 Chr.12: 123647805-123647805 on Build GRCh38
rs772536675 Chr.12: 123647921-123647921 on Build GRCh38
rs939849725 Chr.12: 123647803-123647803 on Build GRCh38
rs761288946 Chr.12: 123647932-123647932 on Build GRCh38
rs774869120 Chr.12: 123647942-123647942 on Build GRCh38
rs767277783 Chr.12: 123647969-123647969 on Build GRCh38
rs527580660 Chr.12: 123647836-123647836 on Build GRCh38
rs770604930 Chr.12: 123647809-123647809 on Build GRCh38

View Full Product Details