Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203342284-203342557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMOD
Entrez Gene ID
2331
Gene Name
fibromodulin
Gene Aliases
FM, SLRR2E
UniGene
Hs.519168
Gene Chromosome Location
Chr.1: 203340621-203351429 on Build GRCh38
Transcripts
NM_002023.4 NR_103757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
902470 c.1017C>T p.V339V Chr.1: 203342457-203342457 on Build GRCh38
4685166 c.1091C>T p.A364V Chr.1: 203342383-203342383 on Build GRCh38
902469 c.1046G>A p.R349H Chr.1: 203342428-203342428 on Build GRCh38
902468 c.1061A>T p.E354V Chr.1: 203342413-203342413 on Build GRCh38
3481858 c.1055G>A p.G352E Chr.1: 203342419-203342419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145901742 Chr.1: 203342415-203342415 on Build GRCh38
rs143883652 Chr.1: 203342479-203342479 on Build GRCh38
rs777586224 Chr.1: 203342527-203342527 on Build GRCh38
rs753219251 Chr.1: 203342459-203342459 on Build GRCh38
rs750816904 Chr.1: 203342436-203342436 on Build GRCh38
rs537266188 Chr.1: 203342519-203342519 on Build GRCh38
rs754423384 Chr.1: 203342517-203342517 on Build GRCh38
rs775111384 Chr.1: 203342328-203342328 on Build GRCh38
rs373114828 Chr.1: 203342307-203342307 on Build GRCh38
rs751877296 Chr.1: 203342423-203342423 on Build GRCh38
rs536977170 Chr.1: 203342387-203342387 on Build GRCh38
rs764401923 Chr.1: 203342369-203342369 on Build GRCh38
rs377479219 Chr.1: 203342383-203342383 on Build GRCh38
rs139855707 Chr.1: 203342457-203342457 on Build GRCh38
rs558230108 Chr.1: 203342306-203342306 on Build GRCh38
rs34712154 Chr.1: 203342520-203342521 on Build GRCh38
rs750987880 Chr.1: 203342385-203342385 on Build GRCh38
rs370475053 Chr.1: 203342433-203342433 on Build GRCh38
rs776639176 Chr.1: 203342474-203342474 on Build GRCh38
rs747434529 Chr.1: 203342311-203342311 on Build GRCh38
rs200147359 Chr.1: 203342404-203342404 on Build GRCh38
rs762063968 Chr.1: 203342505-203342505 on Build GRCh38
rs768385270 Chr.1: 203342349-203342349 on Build GRCh38
rs1048276211 Chr.1: 203342485-203342485 on Build GRCh38
rs200474119 Chr.1: 203342382-203342382 on Build GRCh38
rs781663545 Chr.1: 203342384-203342384 on Build GRCh38
rs768134221 Chr.1: 203342397-203342397 on Build GRCh38
rs1044799289 Chr.1: 203342318-203342318 on Build GRCh38
rs748852635 Chr.1: 203342396-203342396 on Build GRCh38
rs376930920 Chr.1: 203342468-203342468 on Build GRCh38
rs759671087 Chr.1: 203342355-203342355 on Build GRCh38
rs142981560 Chr.1: 203342421-203342421 on Build GRCh38
rs141336017 Chr.1: 203342352-203342352 on Build GRCh38
rs756564558 Chr.1: 203342456-203342456 on Build GRCh38
rs758077408 Chr.1: 203342524-203342524 on Build GRCh38
rs763303009 Chr.1: 203342418-203342418 on Build GRCh38
rs201313469 Chr.1: 203342516-203342516 on Build GRCh38
rs371237431 Chr.1: 203342469-203342469 on Build GRCh38
rs761680300 Chr.1: 203342351-203342351 on Build GRCh38
rs770939371 Chr.1: 203342471-203342471 on Build GRCh38
rs767934884 Chr.1: 203342512-203342512 on Build GRCh38
rs774416390 Chr.1: 203342503-203342503 on Build GRCh38
rs752290430 Chr.1: 203342522-203342522 on Build GRCh38
rs763050638 Chr.1: 203342368-203342368 on Build GRCh38
rs374518065 Chr.1: 203342520-203342520 on Build GRCh38
rs369549642 Chr.1: 203342329-203342329 on Build GRCh38
rs148311417 Chr.1: 203342414-203342414 on Build GRCh38
rs772045623 Chr.1: 203342400-203342400 on Build GRCh38
rs781674354 Chr.1: 203342317-203342317 on Build GRCh38
rs149886220 Chr.1: 203342399-203342399 on Build GRCh38
rs768838898 Chr.1: 203342486-203342486 on Build GRCh38
rs546466318 Chr.1: 203342515-203342515 on Build GRCh38
rs376575413 Chr.1: 203342420-203342420 on Build GRCh38
rs542243510 Chr.1: 203342327-203342327 on Build GRCh38
rs747881241 Chr.1: 203342466-203342466 on Build GRCh38
rs769441362 Chr.1: 203342481-203342481 on Build GRCh38
rs762152396 Chr.1: 203342429-203342429 on Build GRCh38
rs373594801 Chr.1: 203342460-203342460 on Build GRCh38
rs757658881 Chr.1: 203342312-203342312 on Build GRCh38
rs779498869 Chr.1: 203342388-203342388 on Build GRCh38
rs752937396 Chr.1: 203342359-203342359 on Build GRCh38
rs374847025 Chr.1: 203342320-203342320 on Build GRCh38
rs907777319 Chr.1: 203342430-203342430 on Build GRCh38
rs765453522 Chr.1: 203342357-203342357 on Build GRCh38
rs767747763 Chr.1: 203342507-203342507 on Build GRCh38

View Full Product Details