Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151294995-151295238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR87
Entrez Gene ID
53836
Gene Name
G protein-coupled receptor 87
Gene Aliases
FKSG78, GPR95, KPG_002
UniGene
Hs.591292
Gene Chromosome Location
Chr.3: 151294086-151316848 on Build GRCh38
Transcripts
NM_023915.3 NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1

Gene Symbol
MED12L
Entrez Gene ID
116931
Gene Name
mediator complex subunit 12 like
Gene Aliases
NOPAR, TNRC11L, TRALP, TRALPUSH
UniGene
Hs.744234
Gene Chromosome Location
Chr.3: 151085665-151436677 on Build GRCh38
Transcripts
NM_023915.3 NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4688253 c.85G>T p.G29W Chr.3: 151295161-151295161 on Build GRCh38
1208747 c.139G>A p.V47M Chr.3: 151295107-151295107 on Build GRCh38
4879314 c.61C>A p.H21N Chr.3: 151295185-151295185 on Build GRCh38
5994703 c.84C>T p.D28D Chr.3: 151295162-151295162 on Build GRCh38
6990084 c.155T>A p.I52K Chr.3: 151295091-151295091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958642131 Chr.3: 151295090-151295090 on Build GRCh38
rs1049003229 Chr.3: 151295129-151295129 on Build GRCh38
rs778956908 Chr.3: 151295121-151295122 on Build GRCh38
rs761991856 Chr.3: 151295134-151295134 on Build GRCh38
rs756817481 Chr.3: 151295165-151295165 on Build GRCh38
rs775116512 Chr.3: 151295121-151295121 on Build GRCh38
rs531293531 Chr.3: 151295158-151295158 on Build GRCh38
rs368070708 Chr.3: 151295118-151295118 on Build GRCh38
rs145870688 Chr.3: 151295069-151295069 on Build GRCh38
rs370393342 Chr.3: 151295159-151295159 on Build GRCh38
rs755749601 Chr.3: 151295075-151295075 on Build GRCh38
rs910192789 Chr.3: 151295142-151295142 on Build GRCh38
rs1040064840 Chr.3: 151295148-151295148 on Build GRCh38
rs771902875 Chr.3: 151295119-151295119 on Build GRCh38
rs200700829 Chr.3: 151295044-151295044 on Build GRCh38
rs768433946 Chr.3: 151295207-151295207 on Build GRCh38
rs201724998 Chr.3: 151295079-151295079 on Build GRCh38
rs751031771 Chr.3: 151295147-151295147 on Build GRCh38
rs747889185 Chr.3: 151295070-151295070 on Build GRCh38
rs776872234 Chr.3: 151295099-151295099 on Build GRCh38
rs745845124 Chr.3: 151295197-151295197 on Build GRCh38
rs561644452 Chr.3: 151295161-151295161 on Build GRCh38
rs765406887 Chr.3: 151295137-151295137 on Build GRCh38
rs777293275 Chr.3: 151295162-151295162 on Build GRCh38
rs9819885 Chr.3: 151295108-151295108 on Build GRCh38
rs199820125 Chr.3: 151295210-151295210 on Build GRCh38
rs375588496 Chr.3: 151295087-151295087 on Build GRCh38
rs943089988 Chr.3: 151295146-151295146 on Build GRCh38
rs750896215 Chr.3: 151295056-151295056 on Build GRCh38
rs779993172 Chr.3: 151295201-151295201 on Build GRCh38
rs375263702 Chr.3: 151295057-151295057 on Build GRCh38
rs753895845 Chr.3: 151295042-151295042 on Build GRCh38
rs144397049 Chr.3: 151295109-151295109 on Build GRCh38
rs758903048 Chr.3: 151295059-151295059 on Build GRCh38
rs935518563 Chr.3: 151295153-151295153 on Build GRCh38
rs577139354 Chr.3: 151295092-151295092 on Build GRCh38
rs928737976 Chr.3: 151295034-151295034 on Build GRCh38
rs563106165 Chr.3: 151295125-151295125 on Build GRCh38
rs369844880 Chr.3: 151295206-151295206 on Build GRCh38
rs893332584 Chr.3: 151295170-151295170 on Build GRCh38
rs769074939 Chr.3: 151295052-151295052 on Build GRCh38
rs1012224195 Chr.3: 151295192-151295192 on Build GRCh38
rs776856960 Chr.3: 151295130-151295130 on Build GRCh38
rs778372409 Chr.3: 151295174-151295174 on Build GRCh38
rs773321339 Chr.3: 151295164-151295164 on Build GRCh38
rs146615024 Chr.3: 151295198-151295198 on Build GRCh38
rs191098627 Chr.3: 151295160-151295160 on Build GRCh38

View Full Product Details