Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109768522-109768785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222A
Entrez Gene ID
84915
Gene Name
family with sequence similarity 222 member A
Gene Aliases
C12orf34
UniGene
Hs.661785
Gene Chromosome Location
Chr.12: 109714382-109770509 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

Gene Symbol
FAM222A-AS1
Entrez Gene ID
84983
Gene Name
FAM222A antisense RNA 1
Gene Aliases
-
UniGene
Hs.675961
Gene Chromosome Location
Chr.12: 109734209-109773487 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
120030 c.621C>G p.N207K Chr.12: 109768550-109768550 on Build GRCh38
3667083 c.720C>A p.P240P Chr.12: 109768649-109768649 on Build GRCh38
3455895 c.730T>C p.Y244H Chr.12: 109768659-109768659 on Build GRCh38
4170364 c.807G>T p.L269F Chr.12: 109768736-109768736 on Build GRCh38
6431543 c.656C>T p.A219V Chr.12: 109768585-109768585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778158113 Chr.12: 109768618-109768618 on Build GRCh38
rs767534629 Chr.12: 109768668-109768668 on Build GRCh38
rs532999688 Chr.12: 109768764-109768764 on Build GRCh38
rs553887605 Chr.12: 109768594-109768594 on Build GRCh38
rs375805951 Chr.12: 109768763-109768763 on Build GRCh38
rs759692083 Chr.12: 109768762-109768775 on Build GRCh38
rs767676227 Chr.12: 109768760-109768760 on Build GRCh38
rs370934435 Chr.12: 109768575-109768575 on Build GRCh38
rs753269298 Chr.12: 109768613-109768613 on Build GRCh38
rs375860480 Chr.12: 109768646-109768646 on Build GRCh38
rs921904714 Chr.12: 109768636-109768636 on Build GRCh38
rs768696592 Chr.12: 109768642-109768642 on Build GRCh38
rs540424098 Chr.12: 109768649-109768649 on Build GRCh38
rs749603829 Chr.12: 109768620-109768620 on Build GRCh38
rs36083947 Chr.12: 109768571-109768571 on Build GRCh38
rs768717068 Chr.12: 109768722-109768722 on Build GRCh38
rs773932548 Chr.12: 109768541-109768541 on Build GRCh38
rs958445070 Chr.12: 109768556-109768556 on Build GRCh38
rs751665127 Chr.12: 109768758-109768758 on Build GRCh38
rs778364810 Chr.12: 109768628-109768628 on Build GRCh38
rs754952293 Chr.12: 109768591-109768591 on Build GRCh38
rs776650076 Chr.12: 109768729-109768729 on Build GRCh38
rs772052843 Chr.12: 109768549-109768549 on Build GRCh38
rs199593023 Chr.12: 109768689-109768689 on Build GRCh38
rs978647679 Chr.12: 109768581-109768581 on Build GRCh38
rs112811653 Chr.12: 109768673-109768673 on Build GRCh38
rs139699838 Chr.12: 109768567-109768567 on Build GRCh38
rs867053529 Chr.12: 109768596-109768596 on Build GRCh38
rs867676536 Chr.12: 109768587-109768587 on Build GRCh38
rs764543809 Chr.12: 109768677-109768677 on Build GRCh38
rs372654452 Chr.12: 109768745-109768745 on Build GRCh38
rs747781518 Chr.12: 109768710-109768710 on Build GRCh38
rs573612907 Chr.12: 109768674-109768674 on Build GRCh38
rs775233169 Chr.12: 109768551-109768551 on Build GRCh38
rs770649091 Chr.12: 109768624-109768624 on Build GRCh38
rs562758512 Chr.12: 109768740-109768740 on Build GRCh38
rs746472732 Chr.12: 109768716-109768716 on Build GRCh38
rs367783186 Chr.12: 109768690-109768690 on Build GRCh38
rs758043262 Chr.12: 109768711-109768711 on Build GRCh38
rs147633661 Chr.12: 109768629-109768629 on Build GRCh38
rs771541336 Chr.12: 109768631-109768631 on Build GRCh38
rs759726487 Chr.12: 109768746-109768746 on Build GRCh38
rs950019909 Chr.12: 109768617-109768617 on Build GRCh38
rs758931470 Chr.12: 109768542-109768542 on Build GRCh38
rs757508749 Chr.12: 109768687-109768687 on Build GRCh38
rs760681103 Chr.12: 109768553-109768553 on Build GRCh38
rs748099189 Chr.12: 109768733-109768733 on Build GRCh38
rs367798094 Chr.12: 109768598-109768598 on Build GRCh38
rs182420516 Chr.12: 109768682-109768682 on Build GRCh38
rs779761751 Chr.12: 109768713-109768713 on Build GRCh38
rs756581188 Chr.12: 109768616-109768616 on Build GRCh38
rs746986231 Chr.12: 109768633-109768633 on Build GRCh38
rs766634785 Chr.12: 109768654-109768654 on Build GRCh38
rs778549496 Chr.12: 109768664-109768664 on Build GRCh38
rs201105798 Chr.12: 109768656-109768656 on Build GRCh38
rs200837548 Chr.12: 109768572-109768572 on Build GRCh38
rs774491169 Chr.12: 109768663-109768663 on Build GRCh38

View Full Product Details