Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15644614-15644839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTD
Entrez Gene ID
686
Gene Name
biotinidase
Gene Aliases
-
UniGene
Hs.444197
Gene Chromosome Location
Chr.3: 15601352-15653709 on Build GRCh38
Transcripts
XM_006713314.3 XM_011534041.2 XM_017007088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
369437 c.893C>T p.A298V Chr.3: 15644749-15644749 on Build GRCh38
4750330 c.825G>T p.Q275H Chr.3: 15644681-15644681 on Build GRCh38
1040395 c.970G>T p.D324Y Chr.3: 15644826-15644826 on Build GRCh38
7357837 c.880A>G p.I294V Chr.3: 15644736-15644736 on Build GRCh38
7357836 c.910C>T p.P304S Chr.3: 15644766-15644766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771251410 Chr.3: 15644664-15644664 on Build GRCh38
rs764376897 Chr.3: 15644720-15644720 on Build GRCh38
rs148031701 Chr.3: 15644671-15644671 on Build GRCh38
rs145379955 Chr.3: 15644741-15644741 on Build GRCh38
rs754158925 Chr.3: 15644724-15644724 on Build GRCh38
rs549204682 Chr.3: 15644700-15644700 on Build GRCh38
rs144084212 Chr.3: 15644763-15644763 on Build GRCh38
rs769573111 Chr.3: 15644722-15644723 on Build GRCh38
rs529069219 Chr.3: 15644666-15644666 on Build GRCh38
rs751445487 Chr.3: 15644734-15644734 on Build GRCh38
rs552436982 Chr.3: 15644657-15644657 on Build GRCh38
rs397514394 Chr.3: 15644788-15644788 on Build GRCh38
rs372687866 Chr.3: 15644639-15644639 on Build GRCh38
rs397514386 Chr.3: 15644789-15644789 on Build GRCh38
rs397514395 Chr.3: 15644789-15644789 on Build GRCh38
rs946578169 Chr.3: 15644806-15644806 on Build GRCh38
rs915150723 Chr.3: 15644783-15644783 on Build GRCh38
rs778024234 Chr.3: 15644768-15644768 on Build GRCh38
rs397514334 Chr.3: 15644721-15644721 on Build GRCh38
rs398123142 Chr.3: 15644760-15644760 on Build GRCh38
rs377651057 Chr.3: 15644791-15644791 on Build GRCh38
rs980083786 Chr.3: 15644681-15644681 on Build GRCh38
rs398123141 Chr.3: 15644686-15644686 on Build GRCh38
rs35976361 Chr.3: 15644736-15644736 on Build GRCh38
rs748963622 Chr.3: 15644774-15644774 on Build GRCh38
rs397514390 Chr.3: 15644692-15644692 on Build GRCh38
rs769423080 Chr.3: 15644643-15644643 on Build GRCh38
rs397514391 Chr.3: 15644743-15644743 on Build GRCh38
rs397514389 Chr.3: 15644689-15644689 on Build GRCh38
rs532041656 Chr.3: 15644769-15644769 on Build GRCh38
rs760900130 Chr.3: 15644714-15644714 on Build GRCh38
rs772128470 Chr.3: 15644677-15644677 on Build GRCh38
rs185225945 Chr.3: 15644742-15644742 on Build GRCh38
rs397514396 Chr.3: 15644790-15644790 on Build GRCh38
rs144575084 Chr.3: 15644667-15644667 on Build GRCh38
rs397514388 Chr.3: 15644688-15644688 on Build GRCh38
rs777470168 Chr.3: 15644645-15644645 on Build GRCh38
rs368913302 Chr.3: 15644635-15644635 on Build GRCh38
rs775730599 Chr.3: 15644684-15644684 on Build GRCh38
rs587783006 Chr.3: 15644754-15644754 on Build GRCh38
rs747992825 Chr.3: 15644679-15644680 on Build GRCh38
rs757604137 Chr.3: 15644778-15644778 on Build GRCh38
rs397514385 Chr.3: 15644650-15644650 on Build GRCh38
rs397514387 Chr.3: 15644670-15644670 on Build GRCh38
rs397514392 Chr.3: 15644752-15644752 on Build GRCh38
rs397514393 Chr.3: 15644785-15644785 on Build GRCh38

View Full Product Details