Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 66878871-66879144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG5
Entrez Gene ID
27091
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 5
Gene Aliases
-
UniGene
Hs.278907
Gene Chromosome Location
Chr.17: 66877273-66885278 on Build GRCh38
Transcripts
NM_145811.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2797123 c.277G>A p.V93I Chr.17: 66879052-66879052 on Build GRCh38
2797119 c.211C>T p.R71C Chr.17: 66878986-66878986 on Build GRCh38
5387079 c.219C>T p.F73F Chr.17: 66878994-66878994 on Build GRCh38
5387080 c.219C>T p.F73F Chr.17: 66878994-66878994 on Build GRCh38
6523813 c.199G>A p.E67K Chr.17: 66878974-66878974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751848698 Chr.17: 66878998-66878998 on Build GRCh38
rs763442575 Chr.17: 66878987-66878987 on Build GRCh38
rs775226474 Chr.17: 66878962-66878962 on Build GRCh38
rs962028728 Chr.17: 66879103-66879103 on Build GRCh38
rs377716365 Chr.17: 66879037-66879037 on Build GRCh38
rs755009772 Chr.17: 66879008-66879008 on Build GRCh38
rs568669938 Chr.17: 66879072-66879072 on Build GRCh38
rs367823339 Chr.17: 66879074-66879074 on Build GRCh38
rs1054121464 Chr.17: 66878916-66878916 on Build GRCh38
rs747298451 Chr.17: 66879052-66879052 on Build GRCh38
rs373841089 Chr.17: 66879065-66879065 on Build GRCh38
rs146501310 Chr.17: 66879050-66879050 on Build GRCh38
rs370097357 Chr.17: 66879086-66879086 on Build GRCh38
rs752790776 Chr.17: 66879028-66879028 on Build GRCh38
rs374590687 Chr.17: 66879089-66879089 on Build GRCh38
rs752737597 Chr.17: 66879088-66879088 on Build GRCh38
rs746739229 Chr.17: 66879046-66879046 on Build GRCh38
rs753497890 Chr.17: 66878939-66878939 on Build GRCh38
rs750396913 Chr.17: 66879109-66879109 on Build GRCh38
rs759748575 Chr.17: 66879082-66879082 on Build GRCh38
rs771677811 Chr.17: 66878961-66878961 on Build GRCh38
rs201854089 Chr.17: 66878980-66878980 on Build GRCh38
rs1010144937 Chr.17: 66878930-66878930 on Build GRCh38
rs749315235 Chr.17: 66878929-66878929 on Build GRCh38
rs757034347 Chr.17: 66878935-66878935 on Build GRCh38
rs774424619 Chr.17: 66879077-66879077 on Build GRCh38
rs768250158 Chr.17: 66878972-66878972 on Build GRCh38
rs758258180 Chr.17: 66879044-66879044 on Build GRCh38
rs781760573 Chr.17: 66879119-66879119 on Build GRCh38
rs745671178 Chr.17: 66878940-66878940 on Build GRCh38
rs2286676 Chr.17: 66878917-66878917 on Build GRCh38
rs144101002 Chr.17: 66879045-66879045 on Build GRCh38
rs767695865 Chr.17: 66879015-66879015 on Build GRCh38
rs746476317 Chr.17: 66878968-66878968 on Build GRCh38
rs1038350340 Chr.17: 66879063-66879063 on Build GRCh38
rs972265344 Chr.17: 66879111-66879111 on Build GRCh38
rs368998295 Chr.17: 66878970-66878970 on Build GRCh38
rs773643479 Chr.17: 66878985-66878985 on Build GRCh38
rs758742270 Chr.17: 66878981-66878981 on Build GRCh38
rs117599246 Chr.17: 66879036-66879036 on Build GRCh38
rs907021611 Chr.17: 66879118-66879118 on Build GRCh38
rs766489114 Chr.17: 66878995-66878995 on Build GRCh38
rs747664553 Chr.17: 66879064-66879064 on Build GRCh38
rs765133515 Chr.17: 66879106-66879106 on Build GRCh38
rs918177388 Chr.17: 66879113-66879113 on Build GRCh38
rs777542010 Chr.17: 66878926-66878926 on Build GRCh38
rs773878220 Chr.17: 66879066-66879066 on Build GRCh38

View Full Product Details