Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42680497-42680724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR10
Entrez Gene ID
2826
Gene Name
C-C motif chemokine receptor 10
Gene Aliases
GPR2
UniGene
Hs.744542
Gene Chromosome Location
Chr.17: 42679402-42681827 on Build GRCh38
Transcripts
NM_016602.2 XM_017025238.1

Gene Symbol
CNTNAP1
Entrez Gene ID
8506
Gene Name
contactin associated protein 1
Gene Aliases
CASPR, CNTNAP, NRXN4, P190
UniGene
Hs.408730
Gene Chromosome Location
Chr.17: 42679963-42699993 on Build GRCh38
Transcripts
NM_016602.2 XM_017025238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3889705 c.49G>A p.D17N Chr.17: 42680593-42680593 on Build GRCh38
6016650 c.102C>G p.A34A Chr.17: 42680540-42680540 on Build GRCh38
118890 c.54A>G p.E18E Chr.17: 42680588-42680588 on Build GRCh38
1325065 c.45T>C p.S15S Chr.17: 42680597-42680597 on Build GRCh38
6574130 c.93C>T p.C31C Chr.17: 42680549-42680549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374150942 Chr.17: 42680667-42680667 on Build GRCh38
rs370149615 Chr.17: 42680562-42680562 on Build GRCh38
rs777287671 Chr.17: 42680622-42680622 on Build GRCh38
rs779820374 Chr.17: 42680574-42680574 on Build GRCh38
rs931540224 Chr.17: 42680635-42680635 on Build GRCh38
rs918287007 Chr.17: 42680540-42680540 on Build GRCh38
rs1025030301 Chr.17: 42680672-42680673 on Build GRCh38
rs1039748588 Chr.17: 42680591-42680591 on Build GRCh38
rs547726826 Chr.17: 42680568-42680568 on Build GRCh38
rs1001097202 Chr.17: 42680678-42680678 on Build GRCh38
rs778573053 Chr.17: 42680547-42680548 on Build GRCh38
rs542607084 Chr.17: 42680704-42680704 on Build GRCh38
rs777950767 Chr.17: 42680584-42680584 on Build GRCh38
rs748430257 Chr.17: 42680582-42680582 on Build GRCh38
rs527711725 Chr.17: 42680677-42680677 on Build GRCh38
rs747087633 Chr.17: 42680593-42680593 on Build GRCh38
rs749891490 Chr.17: 42680567-42680569 on Build GRCh38
rs772464309 Chr.17: 42680561-42680561 on Build GRCh38
rs374985926 Chr.17: 42680645-42680645 on Build GRCh38
rs770992008 Chr.17: 42680594-42680594 on Build GRCh38
rs751010049 Chr.17: 42680536-42680536 on Build GRCh38
rs776259745 Chr.17: 42680624-42680624 on Build GRCh38
rs770348958 Chr.17: 42680643-42680643 on Build GRCh38
rs757943527 Chr.17: 42680571-42680573 on Build GRCh38
rs757257890 Chr.17: 42680558-42680558 on Build GRCh38
rs887271197 Chr.17: 42680640-42680640 on Build GRCh38
rs763717175 Chr.17: 42680520-42680520 on Build GRCh38
rs533458025 Chr.17: 42680678-42680679 on Build GRCh38
rs1045169571 Chr.17: 42680676-42680676 on Build GRCh38
rs148991339 Chr.17: 42680668-42680668 on Build GRCh38
rs917656225 Chr.17: 42680613-42680613 on Build GRCh38
rs200193441 Chr.17: 42680648-42680648 on Build GRCh38
rs1047261332 Chr.17: 42680639-42680639 on Build GRCh38

View Full Product Details