Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 169689104-169689349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN3
Entrez Gene ID
1182
Gene Name
chloride voltage-gated channel 3
Gene Aliases
CLC3, ClC-3
UniGene
Hs.481186
Gene Chromosome Location
Chr.4: 169620521-169723187 on Build GRCh38
Transcripts
NM_001243372.1 NM_001243374.1 NM_001829.3 NM_173872.3 XM_005262726.2 XM_011531586.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1053183 c.587T>G p.L196* Chr.4: 169689211-169689211 on Build GRCh38
6301141 c.492G>A p.A164A Chr.4: 169689116-169689116 on Build GRCh38
1053182 c.557A>C p.K186T Chr.4: 169689181-169689181 on Build GRCh38
1201284 c.503A>G p.N168S Chr.4: 169689127-169689127 on Build GRCh38
3336466 c.508G>A p.E170K Chr.4: 169689132-169689132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373673523 Chr.4: 169689251-169689251 on Build GRCh38
rs768874623 Chr.4: 169689200-169689200 on Build GRCh38
rs372403360 Chr.4: 169689277-169689277 on Build GRCh38
rs369508350 Chr.4: 169689230-169689230 on Build GRCh38
rs61731290 Chr.4: 169689163-169689163 on Build GRCh38
rs181670822 Chr.4: 169689269-169689269 on Build GRCh38
rs953473166 Chr.4: 169689139-169689139 on Build GRCh38
rs755768006 Chr.4: 169689271-169689271 on Build GRCh38
rs771029683 Chr.4: 169689132-169689132 on Build GRCh38
rs762010145 Chr.4: 169689216-169689216 on Build GRCh38
rs199761618 Chr.4: 169689152-169689152 on Build GRCh38
rs752073451 Chr.4: 169689252-169689252 on Build GRCh38
rs774456990 Chr.4: 169689148-169689148 on Build GRCh38
rs754417159 Chr.4: 169689265-169689265 on Build GRCh38
rs747826742 Chr.4: 169689308-169689308 on Build GRCh38
rs986077983 Chr.4: 169689161-169689161 on Build GRCh38
rs79072182 Chr.4: 169689131-169689131 on Build GRCh38
rs1016259360 Chr.4: 169689198-169689198 on Build GRCh38
rs74366331 Chr.4: 169689130-169689130 on Build GRCh38
rs376136772 Chr.4: 169689222-169689222 on Build GRCh38
rs773500604 Chr.4: 169689227-169689227 on Build GRCh38
rs752381431 Chr.4: 169689270-169689270 on Build GRCh38
rs747033096 Chr.4: 169689190-169689190 on Build GRCh38
rs776931890 Chr.4: 169689215-169689215 on Build GRCh38
rs766832187 Chr.4: 169689245-169689245 on Build GRCh38

View Full Product Details