Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65584709-65584962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA9
Entrez Gene ID
81556
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C15orf44
UniGene
Hs.6686
Gene Chromosome Location
Chr.15: 65578757-65611289 on Build GRCh38
Transcripts
NM_001136043.2 NM_001207058.1 NM_001207059.1 NR_045104.1 NR_045105.2 NR_045106.2 NR_045901.1 XM_006720708.3 XM_006720709.3 XM_006720711.2 XM_011522101.2 XM_017022644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370230 c.1313A>G p.N438S Chr.15: 65584804-65584804 on Build GRCh38
5679497 c.1305_1308delTGCC p.A436fs*3 Chr.15: 65584809-65584812 on Build GRCh38
5558029 c.1009A>C p.K337Q Chr.15: 65584829-65584829 on Build GRCh38
5558028 c.1180A>C p.K394Q Chr.15: 65584829-65584829 on Build GRCh38
3503224 c.1243C>T p.P415S Chr.15: 65584874-65584874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970138011 Chr.15: 65584785-65584785 on Build GRCh38
rs749766948 Chr.15: 65584751-65584751 on Build GRCh38
rs75219055 Chr.15: 65584802-65584802 on Build GRCh38
rs767090427 Chr.15: 65584895-65584895 on Build GRCh38
rs755693764 Chr.15: 65584902-65584902 on Build GRCh38
rs752127914 Chr.15: 65584898-65584898 on Build GRCh38
rs181916225 Chr.15: 65584921-65584921 on Build GRCh38
rs779526425 Chr.15: 65584776-65584776 on Build GRCh38
rs746446919 Chr.15: 65584912-65584912 on Build GRCh38
rs779285193 Chr.15: 65584905-65584905 on Build GRCh38
rs776595935 Chr.15: 65584819-65584819 on Build GRCh38
rs768681172 Chr.15: 65584804-65584804 on Build GRCh38
rs139905468 Chr.15: 65584822-65584822 on Build GRCh38
rs974718186 Chr.15: 65584883-65584883 on Build GRCh38
rs747246228 Chr.15: 65584800-65584800 on Build GRCh38
rs773264549 Chr.15: 65584829-65584829 on Build GRCh38
rs150006676 Chr.15: 65584821-65584821 on Build GRCh38
rs200094084 Chr.15: 65584797-65584797 on Build GRCh38
rs943423251 Chr.15: 65584875-65584875 on Build GRCh38
rs549516107 Chr.15: 65584833-65584833 on Build GRCh38
rs911958695 Chr.15: 65584866-65584866 on Build GRCh38
rs75178492 Chr.15: 65584836-65584836 on Build GRCh38
rs780403124 Chr.15: 65584920-65584920 on Build GRCh38
rs757730102 Chr.15: 65584767-65584767 on Build GRCh38
rs772494546 Chr.15: 65584779-65584779 on Build GRCh38
rs541704325 Chr.15: 65584828-65584828 on Build GRCh38
rs758703970 Chr.15: 65584914-65584914 on Build GRCh38
rs754712737 Chr.15: 65584887-65584887 on Build GRCh38
rs145424731 Chr.15: 65584825-65584825 on Build GRCh38
rs780857694 Chr.15: 65584747-65584747 on Build GRCh38
rs759295047 Chr.15: 65584832-65584833 on Build GRCh38
rs767170805 Chr.15: 65584897-65584897 on Build GRCh38
rs368226294 Chr.15: 65584922-65584922 on Build GRCh38
rs951797137 Chr.15: 65584856-65584856 on Build GRCh38
rs751186831 Chr.15: 65584855-65584855 on Build GRCh38
rs746188420 Chr.15: 65584778-65584778 on Build GRCh38

View Full Product Details