Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 97972203-97972446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STPG2
Entrez Gene ID
285555
Gene Name
sperm tail PG-rich repeat containing 2
Gene Aliases
C4orf37
UniGene
Hs.680226
Gene Chromosome Location
Chr.4: 97441450-98143473 on Build GRCh38
Transcripts
NM_174952.2 XM_011531886.2 XM_017008049.1 XM_017008050.1 XM_017008051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4455785 c.897A>C p.K299N Chr.4: 97972316-97972316 on Build GRCh38
3607234 c.871C>T p.P291S Chr.4: 97972342-97972342 on Build GRCh38
3607233 c.892C>T p.Q298* Chr.4: 97972321-97972321 on Build GRCh38
5715962 c.779G>A p.G260E Chr.4: 97972434-97972434 on Build GRCh38
4918410 c.795G>A p.L265L Chr.4: 97972418-97972418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961732005 Chr.4: 97972316-97972316 on Build GRCh38
rs778845624 Chr.4: 97972332-97972332 on Build GRCh38
rs766774519 Chr.4: 97972405-97972405 on Build GRCh38
rs973592589 Chr.4: 97972324-97972324 on Build GRCh38
rs376239160 Chr.4: 97972241-97972241 on Build GRCh38
rs760345084 Chr.4: 97972269-97972269 on Build GRCh38
rs772708570 Chr.4: 97972252-97972252 on Build GRCh38
rs764725652 Chr.4: 97972295-97972295 on Build GRCh38
rs783960 Chr.4: 97972286-97972286 on Build GRCh38
rs374926948 Chr.4: 97972244-97972244 on Build GRCh38
rs777580428 Chr.4: 97972346-97972346 on Build GRCh38
rs768689530 Chr.4: 97972248-97972248 on Build GRCh38
rs768014726 Chr.4: 97972308-97972308 on Build GRCh38
rs752010884 Chr.4: 97972299-97972299 on Build GRCh38
rs750865624 Chr.4: 97972389-97972389 on Build GRCh38
rs982384106 Chr.4: 97972407-97972407 on Build GRCh38
rs774870889 Chr.4: 97972385-97972385 on Build GRCh38
rs1028808467 Chr.4: 97972281-97972281 on Build GRCh38
rs199755322 Chr.4: 97972275-97972275 on Build GRCh38
rs200606406 Chr.4: 97972402-97972402 on Build GRCh38
rs756444429 Chr.4: 97972322-97972322 on Build GRCh38
rs373513497 Chr.4: 97972384-97972384 on Build GRCh38
rs137988195 Chr.4: 97972326-97972326 on Build GRCh38
rs111720937 Chr.4: 97972338-97972338 on Build GRCh38
rs7654193 Chr.4: 97972377-97972377 on Build GRCh38
rs762498596 Chr.4: 97972392-97972395 on Build GRCh38
rs746637358 Chr.4: 97972354-97972354 on Build GRCh38
rs750632716 Chr.4: 97972310-97972310 on Build GRCh38
rs746099925 Chr.4: 97972238-97972238 on Build GRCh38
rs111697071 Chr.4: 97972245-97972245 on Build GRCh38
rs771770383 Chr.4: 97972266-97972266 on Build GRCh38
rs763351932 Chr.4: 97972292-97972292 on Build GRCh38
rs756211736 Chr.4: 97972239-97972239 on Build GRCh38
rs201716501 Chr.4: 97972300-97972300 on Build GRCh38
rs199644011 Chr.4: 97972339-97972339 on Build GRCh38
rs754013287 Chr.4: 97972408-97972408 on Build GRCh38
rs139143939 Chr.4: 97972255-97972255 on Build GRCh38
rs748052305 Chr.4: 97972337-97972337 on Build GRCh38
rs767963310 Chr.4: 97972387-97972387 on Build GRCh38
rs770625131 Chr.4: 97972356-97972356 on Build GRCh38
rs1008801208 Chr.4: 97972297-97972297 on Build GRCh38
rs201166171 Chr.4: 97972268-97972268 on Build GRCh38
rs965171943 Chr.4: 97972383-97972383 on Build GRCh38
rs762257000 Chr.4: 97972386-97972386 on Build GRCh38
rs783959 Chr.4: 97972325-97972325 on Build GRCh38

View Full Product Details