Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 102889342-102889575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL11A1
Entrez Gene ID
1301
Gene Name
collagen type XI alpha 1 chain
Gene Aliases
CO11A1, COLL6, STL2
UniGene
Hs.523446
Gene Chromosome Location
Chr.1: 102876467-103108580 on Build GRCh38
Transcripts
NM_001190709.1 NM_001854.3 NM_080629.2 NM_080630.3 NR_134980.1 XM_017000334.1 XM_017000335.1 XM_017000337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4454269 c.4489A>T p.K1497* Chr.1: 102889466-102889466 on Build GRCh38
4548085 c.4394G>A p.G1465E Chr.1: 102889561-102889561 on Build GRCh38
674664 c.4483G>T p.G1495* Chr.1: 102889472-102889472 on Build GRCh38
3470433 c.4380G>A p.L1460L Chr.1: 102889539-102889539 on Build GRCh38
4494915 c.4391C>T p.P1464L Chr.1: 102889528-102889528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970605834 Chr.1: 102889438-102889438 on Build GRCh38
rs781508026 Chr.1: 102889414-102889414 on Build GRCh38
rs917541931 Chr.1: 102889542-102889542 on Build GRCh38
rs762282588 Chr.1: 102889441-102889441 on Build GRCh38
rs376841140 Chr.1: 102889494-102889494 on Build GRCh38
rs768590245 Chr.1: 102889515-102889515 on Build GRCh38
rs369947899 Chr.1: 102889501-102889501 on Build GRCh38
rs748557200 Chr.1: 102889536-102889536 on Build GRCh38
rs202194245 Chr.1: 102889531-102889531 on Build GRCh38
rs758235552 Chr.1: 102889496-102889496 on Build GRCh38
rs773023575 Chr.1: 102889461-102889461 on Build GRCh38
rs180734278 Chr.1: 102889539-102889539 on Build GRCh38
rs764225064 Chr.1: 102889404-102889405 on Build GRCh38
rs753455066 Chr.1: 102889479-102889479 on Build GRCh38
rs773776234 Chr.1: 102889547-102889547 on Build GRCh38
rs772289485 Chr.1: 102889448-102889448 on Build GRCh38
rs71094579 Chr.1: 102889410-102889411 on Build GRCh38
rs956114369 Chr.1: 102889416-102889416 on Build GRCh38
rs753965353 Chr.1: 102889409-102889410 on Build GRCh38
rs867331843 Chr.1: 102889516-102889516 on Build GRCh38
rs866621880 Chr.1: 102889493-102889493 on Build GRCh38
rs780241838 Chr.1: 102889506-102889506 on Build GRCh38
rs765257116 Chr.1: 102889483-102889483 on Build GRCh38
rs113226085 Chr.1: 102889383-102889384 on Build GRCh38
rs758096197 Chr.1: 102889413-102889413 on Build GRCh38
rs774699566 Chr.1: 102889440-102889440 on Build GRCh38
rs55821405 Chr.1: 102889503-102889503 on Build GRCh38
rs749419834 Chr.1: 102889507-102889507 on Build GRCh38
rs766122412 Chr.1: 102889470-102889470 on Build GRCh38
rs765529267 Chr.1: 102889412-102889413 on Build GRCh38
rs777371253 Chr.1: 102889497-102889497 on Build GRCh38
rs777643087 Chr.1: 102889385-102889386 on Build GRCh38
rs759091446 Chr.1: 102889480-102889480 on Build GRCh38
rs780043168 Chr.1: 102889428-102889428 on Build GRCh38
rs769264112 Chr.1: 102889433-102889433 on Build GRCh38
rs769875026 Chr.1: 102889415-102889415 on Build GRCh38
rs778698174 Chr.1: 102889411-102889411 on Build GRCh38
rs866783525 Chr.1: 102889523-102889523 on Build GRCh38
rs760349942 Chr.1: 102889468-102889468 on Build GRCh38
rs1031753653 Chr.1: 102889405-102889405 on Build GRCh38
rs755030542 Chr.1: 102889404-102889404 on Build GRCh38

View Full Product Details