Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22094343-22094608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160B2
Entrez Gene ID
64760
Gene Name
family with sequence similarity 160 member B2
Gene Aliases
RAI16
UniGene
Hs.491223
Gene Chromosome Location
Chr.8: 22089150-22104380 on Build GRCh38
Transcripts
NM_022749.5 XM_005273602.3 XM_006716381.2 XM_006716385.3 XM_017013732.1 XM_017013733.1 XM_017013734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1097988 c.55A>G p.S19G Chr.8: 22094449-22094449 on Build GRCh38
5790147 c.102G>A p.T34T Chr.8: 22094496-22094496 on Build GRCh38
2785637 c.114C>T p.I38I Chr.8: 22094508-22094508 on Build GRCh38
2785638 c.124G>T p.D42Y Chr.8: 22094518-22094518 on Build GRCh38
7277829 c.124+8C>A p.? Chr.8: 22094526-22094526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182728799 Chr.8: 22094423-22094423 on Build GRCh38
rs754579255 Chr.8: 22094440-22094440 on Build GRCh38
rs748031119 Chr.8: 22094526-22094526 on Build GRCh38
rs201680362 Chr.8: 22094560-22094560 on Build GRCh38
rs374997784 Chr.8: 22094495-22094495 on Build GRCh38
rs765044215 Chr.8: 22094396-22094396 on Build GRCh38
rs764723609 Chr.8: 22094508-22094508 on Build GRCh38
rs781071257 Chr.8: 22094460-22094460 on Build GRCh38
rs770939639 Chr.8: 22094541-22094541 on Build GRCh38
rs751825062 Chr.8: 22094421-22094421 on Build GRCh38
rs571141421 Chr.8: 22094498-22094499 on Build GRCh38
rs767811700 Chr.8: 22094496-22094496 on Build GRCh38
rs925247432 Chr.8: 22094381-22094381 on Build GRCh38
rs367645450 Chr.8: 22094443-22094443 on Build GRCh38
rs371216333 Chr.8: 22094492-22094492 on Build GRCh38
rs769817102 Chr.8: 22094468-22094468 on Build GRCh38
rs118045682 Chr.8: 22094435-22094435 on Build GRCh38
rs370366168 Chr.8: 22094561-22094561 on Build GRCh38
rs373159977 Chr.8: 22094505-22094505 on Build GRCh38
rs771135996 Chr.8: 22094494-22094494 on Build GRCh38
rs979315886 Chr.8: 22094557-22094557 on Build GRCh38
rs761515276 Chr.8: 22094420-22094421 on Build GRCh38
rs749247106 Chr.8: 22094539-22094539 on Build GRCh38
rs776996124 Chr.8: 22094582-22094582 on Build GRCh38
rs773009471 Chr.8: 22094469-22094469 on Build GRCh38
rs371565241 Chr.8: 22094436-22094436 on Build GRCh38
rs759661056 Chr.8: 22094549-22094549 on Build GRCh38
rs757696592 Chr.8: 22094509-22094509 on Build GRCh38
rs768004338 Chr.8: 22094431-22094431 on Build GRCh38
rs375019626 Chr.8: 22094409-22094409 on Build GRCh38
rs763157367 Chr.8: 22094488-22094488 on Build GRCh38
rs997101867 Chr.8: 22094453-22094453 on Build GRCh38
rs766762370 Chr.8: 22094413-22094413 on Build GRCh38
rs140321569 Chr.8: 22094555-22094555 on Build GRCh38
rs773380808 Chr.8: 22094399-22094399 on Build GRCh38
rs375031656 Chr.8: 22094441-22094441 on Build GRCh38
rs957955054 Chr.8: 22094472-22094472 on Build GRCh38
rs370048349 Chr.8: 22094534-22094534 on Build GRCh38
rs145561106 Chr.8: 22094537-22094537 on Build GRCh38
rs754664753 Chr.8: 22094523-22094523 on Build GRCh38
rs750154326 Chr.8: 22094523-22094527 on Build GRCh38
rs71513864 Chr.8: 22094580-22094580 on Build GRCh38
rs767360021 Chr.8: 22094493-22094496 on Build GRCh38
rs764371910 Chr.8: 22094575-22094575 on Build GRCh38
rs369091522 Chr.8: 22094499-22094499 on Build GRCh38
rs75078590 Chr.8: 22094380-22094380 on Build GRCh38
rs930879664 Chr.8: 22094579-22094579 on Build GRCh38
rs768241018 Chr.8: 22094458-22094458 on Build GRCh38
rs765748849 Chr.8: 22094398-22094398 on Build GRCh38
rs774608054 Chr.8: 22094548-22094548 on Build GRCh38
rs779840070 Chr.8: 22094514-22094514 on Build GRCh38
rs550354111 Chr.8: 22094442-22094442 on Build GRCh38
rs568761716 Chr.8: 22094461-22094461 on Build GRCh38
rs953248108 Chr.8: 22094373-22094373 on Build GRCh38
rs376462849 Chr.8: 22094522-22094522 on Build GRCh38
rs779553944 Chr.8: 22094512-22094512 on Build GRCh38
rs867583478 Chr.8: 22094462-22094462 on Build GRCh38
rs200684768 Chr.8: 22094524-22094524 on Build GRCh38

View Full Product Details