Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54849829-54850076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML6
Entrez Gene ID
400954
Gene Name
echinoderm microtubule associated protein like 6
Gene Aliases
-
UniGene
Hs.656692
Gene Chromosome Location
Chr.2: 54723467-54972908 on Build GRCh38
Transcripts
NM_001039753.2 XM_017004098.1 XM_017004099.1 XM_017004100.1 XM_017004101.1 XM_017004102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3426539 c.1216G>T p.D406Y Chr.2: 54849990-54849990 on Build GRCh38
4600737 c.1225G>C p.E409Q Chr.2: 54849999-54849999 on Build GRCh38
1408588 c.1250C>A p.S417Y Chr.2: 54850024-54850024 on Build GRCh38
1021732 c.1237G>T p.E413* Chr.2: 54850011-54850011 on Build GRCh38
1408587 c.1203A>G p.V401V Chr.2: 54849977-54849977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974368167 Chr.2: 54850027-54850027 on Build GRCh38
rs897003655 Chr.2: 54849865-54849865 on Build GRCh38
rs12713278 Chr.2: 54849924-54849924 on Build GRCh38
rs775030539 Chr.2: 54849965-54849965 on Build GRCh38
rs768540491 Chr.2: 54849877-54849877 on Build GRCh38
rs955132368 Chr.2: 54849943-54849943 on Build GRCh38
rs796901636 Chr.2: 54849918-54849918 on Build GRCh38
rs538363819 Chr.2: 54850019-54850019 on Build GRCh38
rs1003512615 Chr.2: 54849896-54849896 on Build GRCh38
rs146306448 Chr.2: 54849999-54849999 on Build GRCh38
rs528466749 Chr.2: 54849885-54849885 on Build GRCh38
rs749456432 Chr.2: 54849986-54849986 on Build GRCh38
rs75416471 Chr.2: 54849906-54849906 on Build GRCh38
rs549704579 Chr.2: 54849957-54849957 on Build GRCh38
rs557300474 Chr.2: 54849863-54849863 on Build GRCh38
rs967249103 Chr.2: 54849925-54849925 on Build GRCh38
rs747386675 Chr.2: 54849939-54849939 on Build GRCh38
rs941269731 Chr.2: 54850015-54850015 on Build GRCh38
rs1048252840 Chr.2: 54849880-54849880 on Build GRCh38
rs760399521 Chr.2: 54850022-54850022 on Build GRCh38
rs985083055 Chr.2: 54849983-54849983 on Build GRCh38
rs774245505 Chr.2: 54849994-54849994 on Build GRCh38
rs147682109 Chr.2: 54849870-54849870 on Build GRCh38
rs181858905 Chr.2: 54849911-54849911 on Build GRCh38
rs1011600656 Chr.2: 54849919-54849919 on Build GRCh38
rs989836927 Chr.2: 54849967-54849967 on Build GRCh38
rs762482915 Chr.2: 54849972-54849972 on Build GRCh38
rs866848042 Chr.2: 54849993-54849993 on Build GRCh38
rs767294365 Chr.2: 54850005-54850005 on Build GRCh38
rs1046197717 Chr.2: 54850004-54850004 on Build GRCh38
rs906619643 Chr.2: 54849895-54849895 on Build GRCh38
rs928066414 Chr.2: 54850031-54850031 on Build GRCh38
rs200908895 Chr.2: 54849940-54849940 on Build GRCh38

View Full Product Details