Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154351554-154351827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
UniGene
Hs.195464
Gene Chromosome Location
Chr.X: 154348529-154374638 on Build GRCh38
Transcripts
NM_001110556.1 NM_001456.3 XM_011531127.1 XM_011531128.1 XM_011531129.1 XM_011531130.1 XM_011531131.1

SNP Detail

SNP ID Chromosome Location
rs782197194 Chr.X: 154351741-154351741 on Build GRCh38
rs782486602 Chr.X: 154351710-154351710 on Build GRCh38
rs374786760 Chr.X: 154351620-154351620 on Build GRCh38
rs782560073 Chr.X: 154351722-154351722 on Build GRCh38
rs781985045 Chr.X: 154351657-154351657 on Build GRCh38
rs782689584 Chr.X: 154351604-154351604 on Build GRCh38
rs11541266 Chr.X: 154351602-154351602 on Build GRCh38
rs186270494 Chr.X: 154351694-154351694 on Build GRCh38
rs782377203 Chr.X: 154351614-154351614 on Build GRCh38
rs1014739797 Chr.X: 154351754-154351754 on Build GRCh38
rs781933783 Chr.X: 154351624-154351624 on Build GRCh38
rs987524675 Chr.X: 154351791-154351791 on Build GRCh38
rs782661769 Chr.X: 154351617-154351617 on Build GRCh38
rs782815909 Chr.X: 154351712-154351712 on Build GRCh38
rs781984348 Chr.X: 154351607-154351607 on Build GRCh38
rs782245290 Chr.X: 154351747-154351747 on Build GRCh38
rs781876201 Chr.X: 154351775-154351775 on Build GRCh38
rs782614991 Chr.X: 154351744-154351744 on Build GRCh38
rs782557098 Chr.X: 154351738-154351738 on Build GRCh38
rs201203814 Chr.X: 154351668-154351668 on Build GRCh38
rs1137967 Chr.X: 154351582-154351582 on Build GRCh38
rs782356784 Chr.X: 154351653-154351653 on Build GRCh38
rs782770155 Chr.X: 154351603-154351603 on Build GRCh38
rs782084497 Chr.X: 154351640-154351640 on Build GRCh38
rs371689052 Chr.X: 154351613-154351613 on Build GRCh38
rs781878646 Chr.X: 154351591-154351591 on Build GRCh38
rs201153928 Chr.X: 154351611-154351611 on Build GRCh38
rs973473822 Chr.X: 154351762-154351762 on Build GRCh38
rs782799340 Chr.X: 154351755-154351755 on Build GRCh38
rs911889881 Chr.X: 154351800-154351800 on Build GRCh38
rs781833708 Chr.X: 154351702-154351702 on Build GRCh38
rs781910090 Chr.X: 154351689-154351689 on Build GRCh38
rs377274096 Chr.X: 154351606-154351606 on Build GRCh38
rs863223625 Chr.X: 154351637-154351637 on Build GRCh38
rs782472440 Chr.X: 154351743-154351743 on Build GRCh38
rs922037068 Chr.X: 154351773-154351773 on Build GRCh38
rs782718878 Chr.X: 154351700-154351700 on Build GRCh38
rs782777494 Chr.X: 154351571-154351571 on Build GRCh38
rs781910090 Chr.X: 154351689-154351689 on Build GRCh38
rs782310851 Chr.X: 154351621-154351621 on Build GRCh38
rs782129661 Chr.X: 154351667-154351667 on Build GRCh38
rs190732075 Chr.X: 154351721-154351721 on Build GRCh38

View Full Product Details