Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128523685-128523910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
UniGene
Hs.522418
Gene Chromosome Location
Chr.9: 128504692-128542301 on Build GRCh38
Transcripts
NM_001003722.1 NM_001499.2 XM_006717059.2 XM_006717060.2 XM_011518549.1 XM_011518550.1 XM_011518551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1314514 c.784C>A p.L262M Chr.9: 128523733-128523733 on Build GRCh38
3664124 c.880A>G p.I294V Chr.9: 128523829-128523829 on Build GRCh38
3327131 c.893C>T p.S298L Chr.9: 128523842-128523842 on Build GRCh38
3716222 c.865C>A p.L289I Chr.9: 128523814-128523814 on Build GRCh38
4413279 c.883C>T p.R295W Chr.9: 128523832-128523832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146800850 Chr.9: 128523772-128523772 on Build GRCh38
rs759899967 Chr.9: 128523760-128523760 on Build GRCh38
rs772830000 Chr.9: 128523874-128523874 on Build GRCh38
rs370507102 Chr.9: 128523883-128523883 on Build GRCh38
rs752667813 Chr.9: 128523822-128523822 on Build GRCh38
rs370164024 Chr.9: 128523769-128523769 on Build GRCh38
rs202111504 Chr.9: 128523771-128523771 on Build GRCh38
rs957041014 Chr.9: 128523733-128523733 on Build GRCh38
rs377055757 Chr.9: 128523747-128523747 on Build GRCh38
rs771213614 Chr.9: 128523738-128523738 on Build GRCh38
rs900585311 Chr.9: 128523717-128523717 on Build GRCh38
rs199815242 Chr.9: 128523713-128523713 on Build GRCh38
rs763373868 Chr.9: 128523870-128523870 on Build GRCh38
rs760361386 Chr.9: 128523881-128523881 on Build GRCh38
rs749866929 Chr.9: 128523783-128523783 on Build GRCh38
rs201347017 Chr.9: 128523862-128523862 on Build GRCh38
rs929252686 Chr.9: 128523868-128523868 on Build GRCh38
rs778765574 Chr.9: 128523815-128523815 on Build GRCh38
rs777152668 Chr.9: 128523741-128523741 on Build GRCh38
rs747030922 Chr.9: 128523850-128523850 on Build GRCh38
rs773572826 Chr.9: 128523718-128523731 on Build GRCh38
rs924303968 Chr.9: 128523721-128523721 on Build GRCh38
rs775924549 Chr.9: 128523867-128523867 on Build GRCh38
rs771303658 Chr.9: 128523857-128523857 on Build GRCh38
rs539582386 Chr.9: 128523850-128523851 on Build GRCh38
rs199679329 Chr.9: 128523877-128523877 on Build GRCh38
rs747231463 Chr.9: 128523730-128523730 on Build GRCh38
rs773628821 Chr.9: 128523720-128523720 on Build GRCh38
rs761284253 Chr.9: 128523782-128523782 on Build GRCh38
rs977069068 Chr.9: 128523715-128523715 on Build GRCh38
rs976358770 Chr.9: 128523801-128523801 on Build GRCh38
rs769069241 Chr.9: 128523873-128523873 on Build GRCh38
rs772171395 Chr.9: 128523712-128523712 on Build GRCh38
rs746263755 Chr.9: 128523863-128523863 on Build GRCh38
rs770178495 Chr.9: 128523866-128523866 on Build GRCh38
rs142336805 Chr.9: 128523814-128523814 on Build GRCh38
rs778007199 Chr.9: 128523842-128523842 on Build GRCh38
rs1040424743 Chr.9: 128523711-128523711 on Build GRCh38
rs989894540 Chr.9: 128523757-128523757 on Build GRCh38
rs758476654 Chr.9: 128523826-128523826 on Build GRCh38
rs755775223 Chr.9: 128523793-128523793 on Build GRCh38
rs148746665 Chr.9: 128523794-128523794 on Build GRCh38

View Full Product Details