Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40415885-40416122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD3
Entrez Gene ID
956
Gene Name
ectonucleoside triphosphate diphosphohydrolase 3
Gene Aliases
CD39L3, HB6, NTPDase-3
UniGene
Hs.441145
Gene Chromosome Location
Chr.3: 40387156-40428626 on Build GRCh38
Transcripts
NM_001248.3 NM_001291960.1 NM_001291961.1 XM_011534266.2 NR_040100.1

Gene Symbol
ENTPD3-AS1
Entrez Gene ID
285266
Gene Name
ENTPD3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.619663
Gene Chromosome Location
Chr.3: 40390951-40453308 on Build GRCh38
Transcripts
NM_001248.3 NM_001291960.1 NM_001291961.1 XM_011534266.2 NR_040100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1692630 c.643A>G p.T215A Chr.3: 40415885-40415885 on Build GRCh38
6891681 c.661G>A p.G221S Chr.3: 40415903-40415903 on Build GRCh38
1205356 c.810G>T p.K270N Chr.3: 40416052-40416052 on Build GRCh38
78935 c.694G>T p.G232* Chr.3: 40415936-40415936 on Build GRCh38
420015 c.702G>A p.K234K Chr.3: 40415944-40415944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757201619 Chr.3: 40416098-40416098 on Build GRCh38
rs753668285 Chr.3: 40415916-40415916 on Build GRCh38
rs1046363454 Chr.3: 40416005-40416005 on Build GRCh38
rs193921050 Chr.3: 40416054-40416054 on Build GRCh38
rs760176932 Chr.3: 40416051-40416051 on Build GRCh38
rs777937367 Chr.3: 40416027-40416027 on Build GRCh38
rs367889270 Chr.3: 40416015-40416015 on Build GRCh38
rs777474924 Chr.3: 40416084-40416084 on Build GRCh38
rs138605420 Chr.3: 40415996-40415996 on Build GRCh38
rs937834426 Chr.3: 40416009-40416009 on Build GRCh38
rs765213298 Chr.3: 40416007-40416007 on Build GRCh38
rs775077378 Chr.3: 40416050-40416050 on Build GRCh38
rs767529366 Chr.3: 40416087-40416087 on Build GRCh38
rs199627591 Chr.3: 40416049-40416049 on Build GRCh38
rs773994453 Chr.3: 40415972-40415972 on Build GRCh38
rs759175392 Chr.3: 40415979-40415979 on Build GRCh38
rs370656485 Chr.3: 40415944-40415944 on Build GRCh38
rs745342169 Chr.3: 40416047-40416047 on Build GRCh38
rs575537800 Chr.3: 40416025-40416025 on Build GRCh38
rs757229707 Chr.3: 40415918-40415918 on Build GRCh38
rs538265453 Chr.3: 40415992-40415992 on Build GRCh38
rs975098886 Chr.3: 40415981-40415981 on Build GRCh38
rs560602791 Chr.3: 40416088-40416088 on Build GRCh38
rs755739112 Chr.3: 40416045-40416047 on Build GRCh38
rs775323959 Chr.3: 40415993-40415993 on Build GRCh38
rs773650580 Chr.3: 40415962-40415962 on Build GRCh38
rs144743573 Chr.3: 40416032-40416032 on Build GRCh38
rs753635503 Chr.3: 40416085-40416085 on Build GRCh38
rs540350189 Chr.3: 40416085-40416085 on Build GRCh38
rs375689049 Chr.3: 40415927-40415927 on Build GRCh38
rs767258033 Chr.3: 40415984-40415984 on Build GRCh38
rs377177110 Chr.3: 40416098-40416098 on Build GRCh38
rs781278624 Chr.3: 40416021-40416021 on Build GRCh38
rs891689029 Chr.3: 40415968-40415968 on Build GRCh38
rs146864349 Chr.3: 40416059-40416059 on Build GRCh38
rs777736809 Chr.3: 40415910-40415910 on Build GRCh38
rs768848699 Chr.3: 40415929-40415929 on Build GRCh38
rs34266806 Chr.3: 40416033-40416033 on Build GRCh38
rs771476832 Chr.3: 40415970-40415970 on Build GRCh38
rs748511323 Chr.3: 40415939-40415939 on Build GRCh38
rs200719073 Chr.3: 40415982-40415982 on Build GRCh38
rs144633113 Chr.3: 40415930-40415930 on Build GRCh38
rs765123644 Chr.3: 40416080-40416080 on Build GRCh38
rs750318984 Chr.3: 40416084-40416084 on Build GRCh38
rs776276414 Chr.3: 40416075-40416075 on Build GRCh38
rs1053861908 Chr.3: 40416011-40416011 on Build GRCh38
rs988144742 Chr.3: 40415903-40415903 on Build GRCh38
rs761713139 Chr.3: 40416078-40416078 on Build GRCh38
rs748273050 Chr.3: 40416024-40416024 on Build GRCh38
rs767085943 Chr.3: 40416019-40416019 on Build GRCh38
rs755913625 Chr.3: 40415907-40415907 on Build GRCh38
rs780041185 Chr.3: 40415925-40415925 on Build GRCh38
rs546718980 Chr.3: 40415943-40415943 on Build GRCh38
rs763315016 Chr.3: 40415963-40415963 on Build GRCh38
rs751611255 Chr.3: 40416008-40416008 on Build GRCh38
rs149497356 Chr.3: 40416000-40416000 on Build GRCh38
rs757136136 Chr.3: 40416001-40416001 on Build GRCh38

View Full Product Details