Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36322832-36323105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVA1B
Entrez Gene ID
55194
Gene Name
eva-1 homolog B (C. elegans)
Gene Aliases
C1orf78, FAM176B
UniGene
Hs.87016
Gene Chromosome Location
Chr.1: 36322030-36324154 on Build GRCh38
Transcripts
NM_001304762.1 NM_018166.2 NR_130899.1 XM_017001627.1 XM_017002340.1 XM_017002341.1 XM_017002344.1 XM_017002345.1

Gene Symbol
SH3D21
Entrez Gene ID
79729
Gene Name
SH3 domain containing 21
Gene Aliases
C1orf113
UniGene
Hs.524496
Gene Chromosome Location
Chr.1: 36306348-36329221 on Build GRCh38
Transcripts
NM_001304762.1 NM_018166.2 NR_130899.1 XM_017001627.1 XM_017002340.1 XM_017002341.1 XM_017002344.1 XM_017002345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
680900 c.37A>C p.N13H Chr.1: 36323001-36323001 on Build GRCh38
3489056 c.25G>A p.E9K Chr.1: 36323013-36323013 on Build GRCh38
4682737 c.45G>C p.L15L Chr.1: 36322993-36322993 on Build GRCh38
4919463 c.64C>G p.R22G Chr.1: 36322974-36322974 on Build GRCh38
7121274 c.14G>A p.R5Q Chr.1: 36323024-36323024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748202520 Chr.1: 36322974-36322974 on Build GRCh38
rs773766772 Chr.1: 36322991-36322991 on Build GRCh38
rs929174770 Chr.1: 36322860-36322860 on Build GRCh38
rs759982436 Chr.1: 36323024-36323024 on Build GRCh38
rs200398356 Chr.1: 36323054-36323054 on Build GRCh38
rs763603454 Chr.1: 36322993-36322993 on Build GRCh38
rs201730881 Chr.1: 36323057-36323057 on Build GRCh38
rs781275812 Chr.1: 36322957-36322957 on Build GRCh38
rs750155690 Chr.1: 36323053-36323053 on Build GRCh38
rs377438130 Chr.1: 36323049-36323049 on Build GRCh38
rs778732234 Chr.1: 36322946-36322946 on Build GRCh38
rs765706320 Chr.1: 36322950-36322951 on Build GRCh38
rs763549976 Chr.1: 36323047-36323047 on Build GRCh38
rs754681450 Chr.1: 36322989-36322989 on Build GRCh38
rs199578110 Chr.1: 36322958-36322958 on Build GRCh38
rs1001333537 Chr.1: 36323084-36323084 on Build GRCh38
rs929013794 Chr.1: 36322878-36322878 on Build GRCh38
rs781311498 Chr.1: 36323017-36323017 on Build GRCh38
rs765194832 Chr.1: 36323051-36323051 on Build GRCh38
rs373158333 Chr.1: 36322921-36322921 on Build GRCh38
rs774816542 Chr.1: 36323019-36323019 on Build GRCh38
rs771585675 Chr.1: 36323001-36323001 on Build GRCh38
rs756195604 Chr.1: 36323060-36323060 on Build GRCh38
rs1056314736 Chr.1: 36322891-36322891 on Build GRCh38
rs752346350 Chr.1: 36323038-36323038 on Build GRCh38
rs561305377 Chr.1: 36323041-36323041 on Build GRCh38
rs767137555 Chr.1: 36323027-36323027 on Build GRCh38
rs565445600 Chr.1: 36323072-36323072 on Build GRCh38
rs377069767 Chr.1: 36322940-36322940 on Build GRCh38
rs1014702841 Chr.1: 36322920-36322922 on Build GRCh38
rs752181360 Chr.1: 36322930-36322930 on Build GRCh38
rs909029428 Chr.1: 36322917-36322917 on Build GRCh38
rs180865697 Chr.1: 36322902-36322902 on Build GRCh38
rs976193588 Chr.1: 36322861-36322861 on Build GRCh38
rs138179641 Chr.1: 36322973-36322973 on Build GRCh38
rs762236255 Chr.1: 36322934-36322934 on Build GRCh38
rs777751058 Chr.1: 36322937-36322937 on Build GRCh38
rs896445320 Chr.1: 36323046-36323046 on Build GRCh38
rs749410931 Chr.1: 36323061-36323061 on Build GRCh38
rs919006336 Chr.1: 36322872-36322872 on Build GRCh38
rs903447766 Chr.1: 36322960-36322960 on Build GRCh38
rs755588674 Chr.1: 36322931-36322931 on Build GRCh38
rs767105755 Chr.1: 36322928-36322928 on Build GRCh38
rs775008681 Chr.1: 36323076-36323076 on Build GRCh38
rs553993091 Chr.1: 36322870-36322870 on Build GRCh38
rs941440920 Chr.1: 36322888-36322888 on Build GRCh38
rs768830228 Chr.1: 36322951-36322951 on Build GRCh38
rs745689613 Chr.1: 36322950-36322950 on Build GRCh38
rs757018075 Chr.1: 36322945-36322945 on Build GRCh38
rs748387670 Chr.1: 36323058-36323058 on Build GRCh38
rs562247536 Chr.1: 36323007-36323007 on Build GRCh38
rs542640521 Chr.1: 36322906-36322906 on Build GRCh38

View Full Product Details