Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82062772-82063029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS1L
Entrez Gene ID
64118
Gene Name
dihydrouridine synthase 1 like
Gene Aliases
DUS1, PP3111
UniGene
Hs.514599
Gene Chromosome Location
Chr.17: 82057872-82065821 on Build GRCh38
Transcripts
NM_022156.4 XM_005256393.2 XM_005256394.2 XM_006722288.2 XM_006722289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7236362 c.435G>A p.T145T Chr.17: 82062936-82062936 on Build GRCh38
5458663 c.402G>A p.L134L Chr.17: 82062969-82062969 on Build GRCh38
5453605 c.510+5G>A p.? Chr.17: 82062856-82062856 on Build GRCh38
3523761 c.445C>T p.R149C Chr.17: 82062926-82062926 on Build GRCh38
6650256 c.411C>T p.H137H Chr.17: 82062960-82062960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752032535 Chr.17: 82062916-82062916 on Build GRCh38
rs114799231 Chr.17: 82062870-82062870 on Build GRCh38
rs11542332 Chr.17: 82062936-82062936 on Build GRCh38
rs138008936 Chr.17: 82062944-82062944 on Build GRCh38
rs200048736 Chr.17: 82062846-82062846 on Build GRCh38
rs748105874 Chr.17: 82062890-82062890 on Build GRCh38
rs951190676 Chr.17: 82062826-82062826 on Build GRCh38
rs747121126 Chr.17: 82062899-82062899 on Build GRCh38
rs1041946573 Chr.17: 82063007-82063007 on Build GRCh38
rs775667835 Chr.17: 82062994-82062994 on Build GRCh38
rs973912543 Chr.17: 82062879-82062879 on Build GRCh38
rs751958468 Chr.17: 82062937-82062937 on Build GRCh38
rs375155958 Chr.17: 82062964-82062964 on Build GRCh38
rs770057446 Chr.17: 82062906-82062906 on Build GRCh38
rs770111476 Chr.17: 82062993-82062993 on Build GRCh38
rs763383471 Chr.17: 82062859-82062859 on Build GRCh38
rs768468704 Chr.17: 82062824-82062824 on Build GRCh38
rs757295476 Chr.17: 82062968-82062968 on Build GRCh38
rs779743708 Chr.17: 82062818-82062818 on Build GRCh38
rs202183417 Chr.17: 82062891-82062891 on Build GRCh38
rs369553348 Chr.17: 82062812-82062812 on Build GRCh38
rs775989479 Chr.17: 82062908-82062908 on Build GRCh38
rs777827457 Chr.17: 82062895-82062895 on Build GRCh38
rs975292441 Chr.17: 82062856-82062856 on Build GRCh38
rs141386224 Chr.17: 82062869-82062869 on Build GRCh38
rs149239423 Chr.17: 82062915-82062915 on Build GRCh38
rs532728681 Chr.17: 82062926-82062926 on Build GRCh38
rs1003533498 Chr.17: 82062808-82062808 on Build GRCh38
rs760820386 Chr.17: 82062843-82062843 on Build GRCh38
rs768971022 Chr.17: 82063006-82063006 on Build GRCh38
rs748052791 Chr.17: 82062838-82062838 on Build GRCh38
rs780621822 Chr.17: 82062811-82062811 on Build GRCh38
rs903469255 Chr.17: 82062803-82062803 on Build GRCh38
rs778749440 Chr.17: 82062834-82062834 on Build GRCh38
rs753155840 Chr.17: 82062858-82062858 on Build GRCh38
rs200501649 Chr.17: 82062992-82062992 on Build GRCh38
rs781265707 Chr.17: 82062984-82062984 on Build GRCh38
rs772296605 Chr.17: 82062839-82062839 on Build GRCh38
rs771000522 Chr.17: 82062900-82062900 on Build GRCh38
rs765586476 Chr.17: 82062854-82062854 on Build GRCh38
rs766834526 Chr.17: 82062836-82062837 on Build GRCh38
rs200963097 Chr.17: 82062957-82062957 on Build GRCh38
rs754755127 Chr.17: 82062881-82062881 on Build GRCh38
rs746891949 Chr.17: 82062959-82062959 on Build GRCh38
rs753448203 Chr.17: 82062929-82062929 on Build GRCh38
rs778714865 Chr.17: 82062885-82062885 on Build GRCh38
rs530750993 Chr.17: 82062802-82062802 on Build GRCh38
rs749693862 Chr.17: 82063004-82063004 on Build GRCh38
rs759921904 Chr.17: 82062960-82062960 on Build GRCh38
rs749087821 Chr.17: 82062821-82062821 on Build GRCh38
rs776879303 Chr.17: 82062845-82062845 on Build GRCh38
rs771323776 Chr.17: 82062844-82062844 on Build GRCh38
rs372985546 Chr.17: 82062942-82062942 on Build GRCh38
rs760210278 Chr.17: 82062925-82062925 on Build GRCh38
rs759744727 Chr.17: 82062903-82062903 on Build GRCh38
rs755832361 Chr.17: 82062815-82062815 on Build GRCh38
rs138365909 Chr.17: 82062912-82062912 on Build GRCh38

View Full Product Details