Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21227776-21228001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNA17
Entrez Gene ID
3451
Gene Name
interferon alpha 17
Gene Aliases
IFN-alphaI, IFNA, INFA, LEIF2C1
UniGene
Hs.282276
Gene Chromosome Location
Chr.9: 21227243-21228222 on Build GRCh38
Transcripts
NM_021268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371806 c.352A>G p.N118D Chr.9: 21227822-21227822 on Build GRCh38
753774 c.268C>G p.L90V Chr.9: 21227906-21227906 on Build GRCh38
455771 c.369G>T p.V123V Chr.9: 21227805-21227805 on Build GRCh38
753775 c.348G>A p.L116L Chr.9: 21227826-21227826 on Build GRCh38
1107807 c.213C>T p.F71F Chr.9: 21227961-21227961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764371874 Chr.9: 21227856-21227856 on Build GRCh38
rs149908829 Chr.9: 21227971-21227971 on Build GRCh38
rs747924158 Chr.9: 21227887-21227887 on Build GRCh38
rs141849715 Chr.9: 21227822-21227822 on Build GRCh38
rs769441651 Chr.9: 21227938-21227938 on Build GRCh38
rs772003826 Chr.9: 21227837-21227837 on Build GRCh38
rs10117962 Chr.9: 21227943-21227943 on Build GRCh38
rs528004284 Chr.9: 21227902-21227902 on Build GRCh38
rs150705157 Chr.9: 21227954-21227954 on Build GRCh38
rs756182329 Chr.9: 21227871-21227871 on Build GRCh38
rs139808441 Chr.9: 21227964-21227964 on Build GRCh38
rs749484214 Chr.9: 21227874-21227874 on Build GRCh38
rs757728097 Chr.9: 21227865-21227865 on Build GRCh38
rs751562622 Chr.9: 21227979-21227981 on Build GRCh38
rs372605729 Chr.9: 21227974-21227974 on Build GRCh38
rs771948512 Chr.9: 21227892-21227892 on Build GRCh38
rs140273203 Chr.9: 21227969-21227969 on Build GRCh38
rs367746060 Chr.9: 21227881-21227881 on Build GRCh38
rs770580113 Chr.9: 21227918-21227918 on Build GRCh38
rs547643187 Chr.9: 21227928-21227928 on Build GRCh38
rs376464426 Chr.9: 21227810-21227810 on Build GRCh38
rs148562481 Chr.9: 21227926-21227926 on Build GRCh38
rs143136790 Chr.9: 21227846-21227846 on Build GRCh38
rs760235895 Chr.9: 21227841-21227841 on Build GRCh38
rs144977916 Chr.9: 21227975-21227975 on Build GRCh38
rs144769830 Chr.9: 21227866-21227866 on Build GRCh38
rs751914130 Chr.9: 21227857-21227857 on Build GRCh38
rs536442307 Chr.9: 21227951-21227951 on Build GRCh38
rs757718415 Chr.9: 21227801-21227801 on Build GRCh38
rs750915274 Chr.9: 21227952-21227952 on Build GRCh38
rs764303556 Chr.9: 21227951-21227955 on Build GRCh38
rs745899948 Chr.9: 21227820-21227820 on Build GRCh38
rs768320416 Chr.9: 21227824-21227824 on Build GRCh38
rs776217623 Chr.9: 21227936-21227936 on Build GRCh38
rs755158044 Chr.9: 21227879-21227879 on Build GRCh38
rs946737188 Chr.9: 21227836-21227836 on Build GRCh38
rs766449588 Chr.9: 21227962-21227962 on Build GRCh38
rs139755305 Chr.9: 21227967-21227967 on Build GRCh38
rs772867450 Chr.9: 21227838-21227838 on Build GRCh38
rs139173301 Chr.9: 21227835-21227835 on Build GRCh38
rs773281670 Chr.9: 21227895-21227895 on Build GRCh38
rs766205082 Chr.9: 21227844-21227844 on Build GRCh38
rs934448010 Chr.9: 21227894-21227894 on Build GRCh38
rs143201241 Chr.9: 21227965-21227965 on Build GRCh38
rs571521935 Chr.9: 21227868-21227868 on Build GRCh38
rs780311168 Chr.9: 21227873-21227873 on Build GRCh38
rs762207214 Chr.9: 21227950-21227950 on Build GRCh38
rs778602005 Chr.9: 21227885-21227885 on Build GRCh38

View Full Product Details