Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238125129-238125364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPNL
Entrez Gene ID
339768
Gene Name
espin-like
Gene Aliases
-
UniGene
Hs.127724
Gene Chromosome Location
Chr.2: 238100310-238133287 on Build GRCh38
Transcripts
NM_194312.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6852022 c.991C>A p.P331T Chr.2: 238125273-238125273 on Build GRCh38
5958755 c.1021delT p.F341fs*48 Chr.2: 238125303-238125303 on Build GRCh38
3364648 c.1020G>A p.P340P Chr.2: 238125302-238125302 on Build GRCh38
6426751 c.1073G>T p.R358I Chr.2: 238125355-238125355 on Build GRCh38
6350551 c.988-10T>C p.? Chr.2: 238125260-238125260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774990611 Chr.2: 238125253-238125253 on Build GRCh38
rs73100390 Chr.2: 238125333-238125333 on Build GRCh38
rs866375170 Chr.2: 238125300-238125300 on Build GRCh38
rs545926428 Chr.2: 238125310-238125310 on Build GRCh38
rs775548887 Chr.2: 238125185-238125185 on Build GRCh38
rs554091798 Chr.2: 238125289-238125289 on Build GRCh38
rs769665208 Chr.2: 238125252-238125252 on Build GRCh38
rs113133737 Chr.2: 238125180-238125180 on Build GRCh38
rs1018497599 Chr.2: 238125209-238125209 on Build GRCh38
rs371983774 Chr.2: 238125329-238125329 on Build GRCh38
rs976454377 Chr.2: 238125249-238125249 on Build GRCh38
rs752883260 Chr.2: 238125301-238125301 on Build GRCh38
rs745599520 Chr.2: 238125320-238125320 on Build GRCh38
rs10172440 Chr.2: 238125260-238125260 on Build GRCh38
rs773850284 Chr.2: 238125261-238125261 on Build GRCh38
rs201765703 Chr.2: 238125302-238125302 on Build GRCh38
rs751763204 Chr.2: 238125307-238125307 on Build GRCh38
rs575038995 Chr.2: 238125286-238125286 on Build GRCh38
rs766673526 Chr.2: 238125222-238125222 on Build GRCh38
rs754205734 Chr.2: 238125288-238125288 on Build GRCh38
rs146362529 Chr.2: 238125287-238125287 on Build GRCh38
rs778097852 Chr.2: 238125305-238125305 on Build GRCh38
rs965626612 Chr.2: 238125177-238125177 on Build GRCh38
rs535437952 Chr.2: 238125199-238125199 on Build GRCh38
rs745830757 Chr.2: 238125250-238125250 on Build GRCh38
rs1018967671 Chr.2: 238125154-238125154 on Build GRCh38
rs765772365 Chr.2: 238125295-238125295 on Build GRCh38
rs888321609 Chr.2: 238125184-238125184 on Build GRCh38
rs754497729 Chr.2: 238125301-238125302 on Build GRCh38
rs762361432 Chr.2: 238125255-238125255 on Build GRCh38
rs748875074 Chr.2: 238125334-238125334 on Build GRCh38
rs866223172 Chr.2: 238125280-238125280 on Build GRCh38
rs776372751 Chr.2: 238125235-238125235 on Build GRCh38
rs963876234 Chr.2: 238125240-238125240 on Build GRCh38
rs1005488408 Chr.2: 238125202-238125202 on Build GRCh38
rs868673070 Chr.2: 238125298-238125298 on Build GRCh38
rs866696350 Chr.2: 238125328-238125328 on Build GRCh38

View Full Product Details