Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 88128537-88128776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
UniGene
Hs.256398
Gene Chromosome Location
Chr.7: 87934143-88202889 on Build GRCh38
Transcripts
NM_001324417.1 NM_001324418.1 NM_001324419.1 NM_001324420.1 NM_001324421.1 NM_004194.4 NM_016351.5 NM_021721.4 NM_021722.5 NM_021723.4 XM_005250445.3 XM_006716028.3 XM_006716029.3 XM_011516318.1 XM_011516319.2 XM_011516320.2 XM_011516321.2 XM_011516322.1 XM_011516323.1 XM_011516324.1 XM_017012329.1 XM_017012330.1 XM_017012331.1 XM_017012332.1 XM_017012333.1 XM_017012334.1 XM_017012335.1 XM_017012336.1 XM_017012337.1 XM_017012338.1 XM_017012339.1 XM_017012341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6440613 c.749T>C p.L250P Chr.7: 88128672-88128672 on Build GRCh38
253306 c.679-1G>A p.? Chr.7: 88128601-88128601 on Build GRCh38
3929237 c.719A>G p.Y240C Chr.7: 88128642-88128642 on Build GRCh38
4587600 c.714C>T p.T238T Chr.7: 88128637-88128637 on Build GRCh38
1313373 c.729G>C p.L243L Chr.7: 88128652-88128652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553357216 Chr.7: 88128609-88128609 on Build GRCh38
rs753133590 Chr.7: 88128717-88128717 on Build GRCh38
rs375484942 Chr.7: 88128580-88128580 on Build GRCh38
rs529386106 Chr.7: 88128624-88128625 on Build GRCh38
rs778098154 Chr.7: 88128608-88128608 on Build GRCh38
rs755526707 Chr.7: 88128688-88128688 on Build GRCh38
rs762582581 Chr.7: 88128650-88128650 on Build GRCh38
rs755438965 Chr.7: 88128593-88128593 on Build GRCh38
rs765943904 Chr.7: 88128655-88128655 on Build GRCh38
rs200084120 Chr.7: 88128637-88128637 on Build GRCh38
rs760730972 Chr.7: 88128618-88128618 on Build GRCh38
rs35528131 Chr.7: 88128640-88128641 on Build GRCh38
rs576937341 Chr.7: 88128582-88128586 on Build GRCh38
rs750127025 Chr.7: 88128643-88128643 on Build GRCh38
rs751008329 Chr.7: 88128664-88128664 on Build GRCh38
rs1050184853 Chr.7: 88128743-88128743 on Build GRCh38
rs781625913 Chr.7: 88128594-88128594 on Build GRCh38
rs535039786 Chr.7: 88128605-88128605 on Build GRCh38
rs778652405 Chr.7: 88128590-88128590 on Build GRCh38
rs781507085 Chr.7: 88128689-88128689 on Build GRCh38
rs376280562 Chr.7: 88128617-88128617 on Build GRCh38
rs747573791 Chr.7: 88128581-88128581 on Build GRCh38
rs776499590 Chr.7: 88128625-88128625 on Build GRCh38
rs930981330 Chr.7: 88128586-88128586 on Build GRCh38
rs761475772 Chr.7: 88128636-88128636 on Build GRCh38
rs748408043 Chr.7: 88128599-88128599 on Build GRCh38
rs1036474407 Chr.7: 88128735-88128735 on Build GRCh38
rs772028598 Chr.7: 88128625-88128627 on Build GRCh38
rs373610086 Chr.7: 88128741-88128741 on Build GRCh38
rs771110652 Chr.7: 88128571-88128571 on Build GRCh38
rs770080720 Chr.7: 88128602-88128602 on Build GRCh38
rs371419225 Chr.7: 88128561-88128561 on Build GRCh38
rs915573351 Chr.7: 88128578-88128578 on Build GRCh38
rs940839290 Chr.7: 88128732-88128732 on Build GRCh38
rs746044102 Chr.7: 88128579-88128580 on Build GRCh38
rs757943920 Chr.7: 88128574-88128574 on Build GRCh38
rs768409996 Chr.7: 88128622-88128622 on Build GRCh38

View Full Product Details