Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41931623-41931882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BYSL
Entrez Gene ID
705
Gene Name
bystin like
Gene Aliases
BYSTIN
UniGene
Hs.106880
Gene Chromosome Location
Chr.6: 41921227-41933046 on Build GRCh38
Transcripts
NM_004053.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
221520 c.902C>T p.T301I Chr.6: 41931764-41931764 on Build GRCh38
741919 c.907C>T p.R303W Chr.6: 41931769-41931769 on Build GRCh38
4896711 c.886G>T p.E296* Chr.6: 41931748-41931748 on Build GRCh38
3718658 c.908G>A p.R303Q Chr.6: 41931770-41931770 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776714476 Chr.6: 41931758-41931758 on Build GRCh38
rs776499999 Chr.6: 41931673-41931673 on Build GRCh38
rs903628380 Chr.6: 41931819-41931819 on Build GRCh38
rs768416427 Chr.6: 41931667-41931667 on Build GRCh38
rs763726551 Chr.6: 41931713-41931713 on Build GRCh38
rs759434319 Chr.6: 41931761-41931761 on Build GRCh38
rs760921291 Chr.6: 41931661-41931661 on Build GRCh38
rs200658248 Chr.6: 41931804-41931804 on Build GRCh38
rs761399861 Chr.6: 41931772-41931772 on Build GRCh38
rs369562083 Chr.6: 41931744-41931744 on Build GRCh38
rs13198860 Chr.6: 41931816-41931816 on Build GRCh38
rs752554867 Chr.6: 41931699-41931699 on Build GRCh38
rs764063989 Chr.6: 41931769-41931769 on Build GRCh38
rs779179146 Chr.6: 41931807-41931807 on Build GRCh38
rs775314784 Chr.6: 41931765-41931765 on Build GRCh38
rs747035538 Chr.6: 41931748-41931748 on Build GRCh38
rs529128433 Chr.6: 41931834-41931834 on Build GRCh38
rs1039534633 Chr.6: 41931722-41931722 on Build GRCh38
rs754292572 Chr.6: 41931729-41931729 on Build GRCh38
rs373478537 Chr.6: 41931788-41931788 on Build GRCh38
rs762564640 Chr.6: 41931710-41931710 on Build GRCh38
rs573778463 Chr.6: 41931780-41931780 on Build GRCh38
rs140745860 Chr.6: 41931747-41931747 on Build GRCh38
rs769444505 Chr.6: 41931764-41931764 on Build GRCh38
rs1022347796 Chr.6: 41931800-41931800 on Build GRCh38
rs138775421 Chr.6: 41931770-41931770 on Build GRCh38
rs756982001 Chr.6: 41931714-41931714 on Build GRCh38
rs750026698 Chr.6: 41931781-41931781 on Build GRCh38
rs771148456 Chr.6: 41931750-41931750 on Build GRCh38
rs772531320 Chr.6: 41931745-41931745 on Build GRCh38
rs199704592 Chr.6: 41931688-41931688 on Build GRCh38
rs755413608 Chr.6: 41931801-41931801 on Build GRCh38
rs747160022 Chr.6: 41931835-41931835 on Build GRCh38
rs201550998 Chr.6: 41931730-41931730 on Build GRCh38
rs376511574 Chr.6: 41931696-41931696 on Build GRCh38
rs377518005 Chr.6: 41931845-41931845 on Build GRCh38
rs771095337 Chr.6: 41931672-41931672 on Build GRCh38
rs777798786 Chr.6: 41931689-41931689 on Build GRCh38
rs780922787 Chr.6: 41931721-41931721 on Build GRCh38
rs763005319 Chr.6: 41931766-41931766 on Build GRCh38
rs757315825 Chr.6: 41931840-41931840 on Build GRCh38
rs184513792 Chr.6: 41931831-41931831 on Build GRCh38

View Full Product Details