Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47498213-47498474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP6
Entrez Gene ID
54859
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
C3orf75, TMEM103
UniGene
Hs.311100
Gene Chromosome Location
Chr.3: 47495640-47513711 on Build GRCh38
Transcripts
NM_001031703.2 XM_005265241.3 XM_017006669.1 XM_017006670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5355356 c.625C>T p.R209W Chr.3: 47498333-47498333 on Build GRCh38
3775083 c.559G>C p.D187H Chr.3: 47498399-47498399 on Build GRCh38
7178621 c.600C>T p.L200L Chr.3: 47498358-47498358 on Build GRCh38
4005554 c.651C>G p.F217L Chr.3: 47498307-47498307 on Build GRCh38
1045627 c.626G>A p.R209Q Chr.3: 47498332-47498332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372166408 Chr.3: 47498434-47498434 on Build GRCh38
rs974758420 Chr.3: 47498354-47498354 on Build GRCh38
rs758777821 Chr.3: 47498428-47498428 on Build GRCh38
rs757685072 Chr.3: 47498408-47498408 on Build GRCh38
rs2170814 Chr.3: 47498261-47498261 on Build GRCh38
rs768267871 Chr.3: 47498322-47498322 on Build GRCh38
rs368490764 Chr.3: 47498248-47498248 on Build GRCh38
rs368871206 Chr.3: 47498394-47498394 on Build GRCh38
rs752801170 Chr.3: 47498262-47498262 on Build GRCh38
rs748003142 Chr.3: 47498333-47498333 on Build GRCh38
rs769676915 Chr.3: 47498360-47498360 on Build GRCh38
rs765727769 Chr.3: 47498409-47498409 on Build GRCh38
rs751844788 Chr.3: 47498255-47498255 on Build GRCh38
rs201760674 Chr.3: 47498301-47498301 on Build GRCh38
rs779117356 Chr.3: 47498292-47498292 on Build GRCh38
rs571566744 Chr.3: 47498264-47498265 on Build GRCh38
rs374866757 Chr.3: 47498265-47498266 on Build GRCh38
rs779812808 Chr.3: 47498316-47498316 on Build GRCh38
rs772901898 Chr.3: 47498371-47498371 on Build GRCh38
rs146074624 Chr.3: 47498416-47498416 on Build GRCh38
rs764935284 Chr.3: 47498249-47498249 on Build GRCh38
rs149565751 Chr.3: 47498380-47498380 on Build GRCh38
rs139283004 Chr.3: 47498321-47498321 on Build GRCh38
rs760187762 Chr.3: 47498424-47498426 on Build GRCh38
rs76414810 Chr.3: 47498277-47498277 on Build GRCh38
rs370492398 Chr.3: 47498332-47498332 on Build GRCh38
rs767727306 Chr.3: 47498383-47498383 on Build GRCh38
rs775052090 Chr.3: 47498421-47498422 on Build GRCh38
rs750785121 Chr.3: 47498426-47498426 on Build GRCh38
rs62617110 Chr.3: 47498395-47498395 on Build GRCh38
rs754333078 Chr.3: 47498281-47498281 on Build GRCh38
rs757710132 Chr.3: 47498291-47498291 on Build GRCh38
rs758010210 Chr.3: 47498306-47498306 on Build GRCh38
rs199767528 Chr.3: 47498393-47498393 on Build GRCh38

View Full Product Details