Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 67397466-67397739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETAA1
Entrez Gene ID
54465
Gene Name
Ewing tumor associated antigen 1
Gene Aliases
ETAA16
UniGene
Hs.353022
Gene Chromosome Location
Chr.2: 67397310-67410411 on Build GRCh38
Transcripts
NM_019002.3 XM_017004376.1 XM_017004377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
32287 c.150G>C p.E50D Chr.2: 67397598-67397598 on Build GRCh38
6267681 c.72G>T p.E24D Chr.2: 67397520-67397520 on Build GRCh38
6319407 c.137G>T p.C46F Chr.2: 67397585-67397585 on Build GRCh38
145380 c.50C>G p.P17R Chr.2: 67397498-67397498 on Build GRCh38
239750 c.64G>A p.A22T Chr.2: 67397512-67397512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753233460 Chr.2: 67397716-67397716 on Build GRCh38
rs748065408 Chr.2: 67397713-67397713 on Build GRCh38
rs766346734 Chr.2: 67397699-67397699 on Build GRCh38
rs1007828965 Chr.2: 67397561-67397561 on Build GRCh38
rs769954262 Chr.2: 67397633-67397633 on Build GRCh38
rs774073695 Chr.2: 67397565-67397565 on Build GRCh38
rs747070971 Chr.2: 67397554-67397554 on Build GRCh38
rs191028204 Chr.2: 67397582-67397582 on Build GRCh38
rs745974091 Chr.2: 67397632-67397632 on Build GRCh38
rs750001329 Chr.2: 67397587-67397587 on Build GRCh38
rs376765349 Chr.2: 67397567-67397567 on Build GRCh38
rs755608941 Chr.2: 67397486-67397486 on Build GRCh38
rs202103721 Chr.2: 67397533-67397533 on Build GRCh38
rs535180692 Chr.2: 67397532-67397532 on Build GRCh38
rs772652887 Chr.2: 67397542-67397542 on Build GRCh38
rs963316512 Chr.2: 67397549-67397549 on Build GRCh38
rs543961574 Chr.2: 67397630-67397630 on Build GRCh38
rs150473742 Chr.2: 67397496-67397496 on Build GRCh38
rs550653940 Chr.2: 67397593-67397593 on Build GRCh38
rs375479843 Chr.2: 67397537-67397537 on Build GRCh38
rs766029229 Chr.2: 67397551-67397552 on Build GRCh38
rs369204741 Chr.2: 67397585-67397585 on Build GRCh38
rs750398783 Chr.2: 67397581-67397581 on Build GRCh38
rs894532733 Chr.2: 67397485-67397485 on Build GRCh38
rs773880236 Chr.2: 67397544-67397544 on Build GRCh38
rs1043043780 Chr.2: 67397691-67397691 on Build GRCh38
rs947421649 Chr.2: 67397686-67397686 on Build GRCh38
rs559825297 Chr.2: 67397695-67397695 on Build GRCh38
rs201688163 Chr.2: 67397563-67397563 on Build GRCh38
rs186821529 Chr.2: 67397679-67397679 on Build GRCh38
rs755710786 Chr.2: 67397640-67397640 on Build GRCh38
rs758865669 Chr.2: 67397603-67397603 on Build GRCh38
rs200411178 Chr.2: 67397531-67397531 on Build GRCh38
rs182131786 Chr.2: 67397607-67397607 on Build GRCh38
rs779721315 Chr.2: 67397495-67397495 on Build GRCh38
rs528491231 Chr.2: 67397706-67397706 on Build GRCh38
rs767398483 Chr.2: 67397575-67397575 on Build GRCh38
rs761614625 Chr.2: 67397566-67397566 on Build GRCh38
rs369830504 Chr.2: 67397614-67397614 on Build GRCh38
rs778881204 Chr.2: 67397703-67397703 on Build GRCh38
rs866918651 Chr.2: 67397527-67397527 on Build GRCh38
rs770958792 Chr.2: 67397557-67397557 on Build GRCh38
rs368629531 Chr.2: 67397538-67397538 on Build GRCh38
rs753063311 Chr.2: 67397596-67397596 on Build GRCh38
rs771037294 Chr.2: 67397623-67397623 on Build GRCh38
rs764943266 Chr.2: 67397717-67397717 on Build GRCh38
rs954527703 Chr.2: 67397576-67397576 on Build GRCh38
rs761561082 Chr.2: 67397648-67397648 on Build GRCh38
rs114064178 Chr.2: 67397647-67397647 on Build GRCh38
rs770005627 Chr.2: 67397564-67397564 on Build GRCh38
rs903397923 Chr.2: 67397659-67397659 on Build GRCh38
rs868352821 Chr.2: 67397516-67397516 on Build GRCh38
rs373034985 Chr.2: 67397589-67397589 on Build GRCh38
rs76623705 Chr.2: 67397667-67397667 on Build GRCh38

View Full Product Details