Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1790666-1790891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFALS
Entrez Gene ID
3483
Gene Name
insulin like growth factor binding protein acid labile subunit
Gene Aliases
ACLSD, ALS
UniGene
Hs.839
Gene Chromosome Location
Chr.16: 1790413-1794908 on Build GRCh38
Transcripts
NM_001146006.1 NM_004970.2 NR_027389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
967676 c.1555C>T p.R519W Chr.16: 1790863-1790863 on Build GRCh38
1478564 c.1599G>A p.L533L Chr.16: 1790819-1790819 on Build GRCh38
4985400 c.1642C>T p.R548W Chr.16: 1790776-1790776 on Build GRCh38
5363337 c.1722C>T p.P574P Chr.16: 1790696-1790696 on Build GRCh38
4783888 c.1833G>A p.P611P Chr.16: 1790699-1790699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753103491 Chr.16: 1790806-1790806 on Build GRCh38
rs780935774 Chr.16: 1790817-1790817 on Build GRCh38
rs144587967 Chr.16: 1790813-1790814 on Build GRCh38
rs367736547 Chr.16: 1790830-1790830 on Build GRCh38
rs1028585073 Chr.16: 1790862-1790862 on Build GRCh38
rs146150726 Chr.16: 1790768-1790768 on Build GRCh38
rs761055308 Chr.16: 1790718-1790718 on Build GRCh38
rs140029438 Chr.16: 1790841-1790841 on Build GRCh38
rs758601429 Chr.16: 1790810-1790810 on Build GRCh38
rs763375409 Chr.16: 1790853-1790853 on Build GRCh38
rs143070371 Chr.16: 1790710-1790710 on Build GRCh38
rs866554435 Chr.16: 1790782-1790782 on Build GRCh38
rs781257809 Chr.16: 1790758-1790758 on Build GRCh38
rs555598374 Chr.16: 1790744-1790744 on Build GRCh38
rs371573897 Chr.16: 1790837-1790837 on Build GRCh38
rs200236817 Chr.16: 1790709-1790709 on Build GRCh38
rs202125685 Chr.16: 1790781-1790781 on Build GRCh38
rs373935300 Chr.16: 1790772-1790772 on Build GRCh38
rs370380271 Chr.16: 1790739-1790739 on Build GRCh38
rs138453673 Chr.16: 1790693-1790693 on Build GRCh38
rs776951879 Chr.16: 1790848-1790848 on Build GRCh38
rs897024097 Chr.16: 1790699-1790699 on Build GRCh38
rs190570821 Chr.16: 1790734-1790734 on Build GRCh38
rs886051775 Chr.16: 1790724-1790724 on Build GRCh38
rs766787656 Chr.16: 1790780-1790780 on Build GRCh38
rs1034101201 Chr.16: 1790746-1790746 on Build GRCh38
rs771622025 Chr.16: 1790845-1790845 on Build GRCh38
rs764235104 Chr.16: 1790858-1790858 on Build GRCh38
rs985185906 Chr.16: 1790795-1790795 on Build GRCh38
rs776272184 Chr.16: 1790695-1790695 on Build GRCh38
rs775287440 Chr.16: 1790708-1790708 on Build GRCh38
rs918166269 Chr.16: 1790790-1790790 on Build GRCh38
rs895114283 Chr.16: 1790760-1790760 on Build GRCh38
rs121909247 Chr.16: 1790800-1790800 on Build GRCh38
rs760061243 Chr.16: 1790849-1790849 on Build GRCh38
rs753982847 Chr.16: 1790767-1790767 on Build GRCh38
rs768462499 Chr.16: 1790774-1790774 on Build GRCh38
rs765624618 Chr.16: 1790792-1790792 on Build GRCh38
rs955315443 Chr.16: 1790786-1790786 on Build GRCh38
rs764361975 Chr.16: 1790813-1790813 on Build GRCh38
rs755932415 Chr.16: 1790823-1790823 on Build GRCh38
rs576561125 Chr.16: 1790815-1790815 on Build GRCh38
rs779961927 Chr.16: 1790825-1790825 on Build GRCh38
rs770760966 Chr.16: 1790756-1790756 on Build GRCh38
rs201091628 Chr.16: 1790696-1790696 on Build GRCh38
rs1032892017 Chr.16: 1790773-1790773 on Build GRCh38
rs758435187 Chr.16: 1790741-1790741 on Build GRCh38
rs763718123 Chr.16: 1790711-1790711 on Build GRCh38
rs533252393 Chr.16: 1790700-1790700 on Build GRCh38
rs757236289 Chr.16: 1790869-1790869 on Build GRCh38
rs9282731 Chr.16: 1790776-1790776 on Build GRCh38
rs888881437 Chr.16: 1790863-1790863 on Build GRCh38
rs759757640 Chr.16: 1790791-1790791 on Build GRCh38
rs773976260 Chr.16: 1790775-1790775 on Build GRCh38
rs749143663 Chr.16: 1790827-1790827 on Build GRCh38
rs745841900 Chr.16: 1790819-1790819 on Build GRCh38
rs747166662 Chr.16: 1790755-1790755 on Build GRCh38
rs770945305 Chr.16: 1790694-1790694 on Build GRCh38
rs751822089 Chr.16: 1790859-1790859 on Build GRCh38
rs766957939 Chr.16: 1790733-1790733 on Build GRCh38
rs2230053 Chr.16: 1790852-1790852 on Build GRCh38
rs376068218 Chr.16: 1790713-1790713 on Build GRCh38
rs774115726 Chr.16: 1790840-1790840 on Build GRCh38
rs765784345 Chr.16: 1790850-1790850 on Build GRCh38
rs200313587 Chr.16: 1790740-1790740 on Build GRCh38
rs755433702 Chr.16: 1790735-1790735 on Build GRCh38
rs745585593 Chr.16: 1790759-1790759 on Build GRCh38

View Full Product Details