Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98034796-98035063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR3
Entrez Gene ID
200959
Gene Name
gamma-aminobutyric acid type A receptor rho3 subunit (gene/pseudogene)
Gene Aliases
-
UniGene
Hs.534578
Gene Chromosome Location
Chr.3: 97986683-98035304 on Build GRCh38
Transcripts
NM_001105580.2 NR_047685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5351670 c.107C>A p.S36Y Chr.3: 98034881-98034881 on Build GRCh38
3767604 c.98G>A p.R33Q Chr.3: 98034890-98034890 on Build GRCh38
4920837 c.7C>T p.L3L Chr.3: 98034981-98034981 on Build GRCh38
3598217 c.30C>T p.F10F Chr.3: 98034958-98034958 on Build GRCh38
3945763 c.60T>C p.N20N Chr.3: 98034928-98034928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945319333 Chr.3: 98034920-98034920 on Build GRCh38
rs774292906 Chr.3: 98034836-98034836 on Build GRCh38
rs770772282 Chr.3: 98035000-98035000 on Build GRCh38
rs760390933 Chr.3: 98034845-98034845 on Build GRCh38
rs76438318 Chr.3: 98034841-98034841 on Build GRCh38
rs770738816 Chr.3: 98035004-98035004 on Build GRCh38
rs762387385 Chr.3: 98035028-98035028 on Build GRCh38
rs982115382 Chr.3: 98034903-98034903 on Build GRCh38
rs774794406 Chr.3: 98035030-98035030 on Build GRCh38
rs749322852 Chr.3: 98034988-98034988 on Build GRCh38
rs766071244 Chr.3: 98034846-98034846 on Build GRCh38
rs777576773 Chr.3: 98034919-98034919 on Build GRCh38
rs554842303 Chr.3: 98034971-98034971 on Build GRCh38
rs916694125 Chr.3: 98034894-98034894 on Build GRCh38
rs764531458 Chr.3: 98034942-98034942 on Build GRCh38
rs984154147 Chr.3: 98034892-98034892 on Build GRCh38
rs759045445 Chr.3: 98034848-98034848 on Build GRCh38
rs780165542 Chr.3: 98034983-98034983 on Build GRCh38
rs772194418 Chr.3: 98034918-98034918 on Build GRCh38
rs143792990 Chr.3: 98034928-98034928 on Build GRCh38
rs776299973 Chr.3: 98034934-98034934 on Build GRCh38
rs756353831 Chr.3: 98034982-98034982 on Build GRCh38
rs773295629 Chr.3: 98034921-98034921 on Build GRCh38
rs547104764 Chr.3: 98034860-98034860 on Build GRCh38
rs375217741 Chr.3: 98034828-98034828 on Build GRCh38
rs374030321 Chr.3: 98035029-98035029 on Build GRCh38
rs762344579 Chr.3: 98035034-98035034 on Build GRCh38
rs369303504 Chr.3: 98034939-98034939 on Build GRCh38
rs777811241 Chr.3: 98034999-98034999 on Build GRCh38
rs751983055 Chr.3: 98034884-98034884 on Build GRCh38
rs750624008 Chr.3: 98034979-98034979 on Build GRCh38
rs752069215 Chr.3: 98034962-98034962 on Build GRCh38
rs541308361 Chr.3: 98034899-98034899 on Build GRCh38
rs781412057 Chr.3: 98034891-98034891 on Build GRCh38
rs373708730 Chr.3: 98035013-98035013 on Build GRCh38
rs568690772 Chr.3: 98035023-98035023 on Build GRCh38
rs546066170 Chr.3: 98034999-98035000 on Build GRCh38
rs532906761 Chr.3: 98034890-98034890 on Build GRCh38
rs763442013 Chr.3: 98034940-98034940 on Build GRCh38
rs769391248 Chr.3: 98035026-98035026 on Build GRCh38
rs759162357 Chr.3: 98034938-98034938 on Build GRCh38
rs755112434 Chr.3: 98034990-98034990 on Build GRCh38
rs936701923 Chr.3: 98034958-98034958 on Build GRCh38
rs746963619 Chr.3: 98035001-98035001 on Build GRCh38
rs1028060152 Chr.3: 98034902-98034902 on Build GRCh38
rs1015263205 Chr.3: 98034870-98034870 on Build GRCh38
rs746732585 Chr.3: 98034924-98034924 on Build GRCh38
rs527275820 Chr.3: 98034856-98034856 on Build GRCh38
rs986616384 Chr.3: 98034912-98034912 on Build GRCh38
rs746111039 Chr.3: 98034897-98034897 on Build GRCh38
rs767818851 Chr.3: 98034972-98034972 on Build GRCh38
rs560983160 Chr.3: 98034905-98034905 on Build GRCh38
rs376910907 Chr.3: 98035021-98035021 on Build GRCh38
rs748089706 Chr.3: 98034911-98034911 on Build GRCh38
rs764677878 Chr.3: 98034853-98034853 on Build GRCh38
rs1019876778 Chr.3: 98034839-98034839 on Build GRCh38
rs374780759 Chr.3: 98034877-98034877 on Build GRCh38
rs369269296 Chr.3: 98035027-98035027 on Build GRCh38
rs770916263 Chr.3: 98034977-98034977 on Build GRCh38
rs776331926 Chr.3: 98035005-98035005 on Build GRCh38
rs761701493 Chr.3: 98034838-98034838 on Build GRCh38

View Full Product Details