Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75279898-75280157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOS
Entrez Gene ID
2353
Gene Name
Fos proto-oncogene, AP-1 transcription factor subunit
Gene Aliases
AP-1, C-FOS, p55
UniGene
Hs.25647
Gene Chromosome Location
Chr.14: 75278778-75282234 on Build GRCh38
Transcripts
NM_005252.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6862022 c.244G>A p.V82I Chr.14: 75279979-75279979 on Build GRCh38
6889003 c.301C>T p.P101S Chr.14: 75280036-75280036 on Build GRCh38
6202270 c.217C>T p.L73L Chr.14: 75279952-75279952 on Build GRCh38
4485595 c.294C>T p.V98V Chr.14: 75280029-75280029 on Build GRCh38
5030848 c.289G>A p.G97R Chr.14: 75280024-75280024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753551210 Chr.14: 75280005-75280005 on Build GRCh38
rs111677793 Chr.14: 75280129-75280129 on Build GRCh38
rs1047557832 Chr.14: 75280083-75280083 on Build GRCh38
rs374460026 Chr.14: 75279922-75279922 on Build GRCh38
rs769109044 Chr.14: 75280091-75280091 on Build GRCh38
rs749787178 Chr.14: 75280085-75280085 on Build GRCh38
rs74685695 Chr.14: 75279965-75279965 on Build GRCh38
rs764698199 Chr.14: 75280017-75280017 on Build GRCh38
rs200018787 Chr.14: 75280028-75280028 on Build GRCh38
rs373077681 Chr.14: 75280007-75280007 on Build GRCh38
rs76195634 Chr.14: 75279924-75279924 on Build GRCh38
rs764792930 Chr.14: 75280120-75280120 on Build GRCh38
rs1008791119 Chr.14: 75280113-75280113 on Build GRCh38
rs148786753 Chr.14: 75279946-75279946 on Build GRCh38
rs759555917 Chr.14: 75279967-75279967 on Build GRCh38
rs781343060 Chr.14: 75280067-75280067 on Build GRCh38
rs150427794 Chr.14: 75280102-75280102 on Build GRCh38
rs752219516 Chr.14: 75280136-75280136 on Build GRCh38
rs766059225 Chr.14: 75279999-75279999 on Build GRCh38
rs983768983 Chr.14: 75280103-75280103 on Build GRCh38
rs769885316 Chr.14: 75279968-75279968 on Build GRCh38
rs539178116 Chr.14: 75280037-75280037 on Build GRCh38
rs370389688 Chr.14: 75280097-75280097 on Build GRCh38
rs1000193997 Chr.14: 75279947-75279947 on Build GRCh38
rs760397156 Chr.14: 75279993-75279993 on Build GRCh38
rs750005065 Chr.14: 75280092-75280092 on Build GRCh38
rs1044633931 Chr.14: 75279938-75279938 on Build GRCh38
rs762206686 Chr.14: 75280095-75280095 on Build GRCh38
rs757058965 Chr.14: 75280048-75280048 on Build GRCh38
rs865962345 Chr.14: 75279978-75279978 on Build GRCh38
rs774634258 Chr.14: 75279988-75279988 on Build GRCh38
rs547215123 Chr.14: 75280016-75280016 on Build GRCh38
rs752661669 Chr.14: 75280027-75280027 on Build GRCh38
rs1046117 Chr.14: 75279987-75279987 on Build GRCh38
rs746120492 Chr.14: 75280076-75280076 on Build GRCh38
rs759315463 Chr.14: 75280111-75280111 on Build GRCh38
rs746741558 Chr.14: 75280041-75280041 on Build GRCh38
rs771873149 Chr.14: 75279923-75279923 on Build GRCh38
rs377565128 Chr.14: 75279963-75279963 on Build GRCh38
rs774924017 Chr.14: 75280093-75280093 on Build GRCh38
rs869124947 Chr.14: 75280112-75280113 on Build GRCh38
rs763409064 Chr.14: 75279975-75279975 on Build GRCh38
rs747221475 Chr.14: 75279936-75279936 on Build GRCh38
rs141780846 Chr.14: 75280078-75280078 on Build GRCh38
rs151293218 Chr.14: 75280077-75280077 on Build GRCh38
rs200973067 Chr.14: 75279973-75279973 on Build GRCh38
rs1020715103 Chr.14: 75280057-75280057 on Build GRCh38

View Full Product Details