Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230710309-230710568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGT
Entrez Gene ID
183
Gene Name
angiotensinogen
Gene Aliases
ANHU, SERPINA8
UniGene
Hs.19383
Gene Chromosome Location
Chr.1: 230702523-230714590 on Build GRCh38
Transcripts
NM_000029.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3485003 c.346C>T p.R116C Chr.1: 230710505-230710505 on Build GRCh38
6596482 c.472C>T p.L158L Chr.1: 230710379-230710379 on Build GRCh38
1639750 c.284T>C p.L95P Chr.1: 230710567-230710567 on Build GRCh38
6596477 c.473T>C p.L158P Chr.1: 230710378-230710378 on Build GRCh38
6596478 c.484C>A p.L162M Chr.1: 230710367-230710367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766006056 Chr.1: 230710532-230710532 on Build GRCh38
rs143232180 Chr.1: 230710473-230710473 on Build GRCh38
rs746282361 Chr.1: 230710493-230710493 on Build GRCh38
rs755823462 Chr.1: 230710410-230710410 on Build GRCh38
rs535619442 Chr.1: 230710347-230710347 on Build GRCh38
rs138660113 Chr.1: 230710421-230710421 on Build GRCh38
rs34829218 Chr.1: 230710441-230710441 on Build GRCh38
rs766201490 Chr.1: 230710452-230710452 on Build GRCh38
rs5040 Chr.1: 230710521-230710521 on Build GRCh38
rs915949844 Chr.1: 230710513-230710513 on Build GRCh38
rs933260912 Chr.1: 230710537-230710537 on Build GRCh38
rs201189192 Chr.1: 230710455-230710455 on Build GRCh38
rs200223350 Chr.1: 230710546-230710546 on Build GRCh38
rs11557885 Chr.1: 230710449-230710449 on Build GRCh38
rs201406560 Chr.1: 230710541-230710541 on Build GRCh38
rs61762539 Chr.1: 230710439-230710439 on Build GRCh38
rs376049907 Chr.1: 230710451-230710451 on Build GRCh38
rs780044023 Chr.1: 230710427-230710427 on Build GRCh38
rs1035501663 Chr.1: 230710475-230710475 on Build GRCh38
rs771357989 Chr.1: 230710426-230710426 on Build GRCh38
rs760098220 Chr.1: 230710434-230710434 on Build GRCh38
rs747055984 Chr.1: 230710488-230710488 on Build GRCh38
rs756744141 Chr.1: 230710349-230710349 on Build GRCh38
rs374118996 Chr.1: 230710514-230710514 on Build GRCh38
rs369425934 Chr.1: 230710472-230710472 on Build GRCh38
rs909004117 Chr.1: 230710474-230710474 on Build GRCh38
rs138340265 Chr.1: 230710442-230710442 on Build GRCh38
rs1021962354 Chr.1: 230710466-230710466 on Build GRCh38
rs759296166 Chr.1: 230710534-230710534 on Build GRCh38
rs11557881 Chr.1: 230710522-230710522 on Build GRCh38
rs199864970 Chr.1: 230710501-230710501 on Build GRCh38
rs778188336 Chr.1: 230710413-230710413 on Build GRCh38
rs779179920 Chr.1: 230710486-230710486 on Build GRCh38
rs748298261 Chr.1: 230710403-230710403 on Build GRCh38
rs149236456 Chr.1: 230710484-230710484 on Build GRCh38
rs759917126 Chr.1: 230710333-230710333 on Build GRCh38
rs756283153 Chr.1: 230710465-230710465 on Build GRCh38
rs143437550 Chr.1: 230710387-230710387 on Build GRCh38
rs750049082 Chr.1: 230710370-230710370 on Build GRCh38
rs11557886 Chr.1: 230710446-230710446 on Build GRCh38
rs563597943 Chr.1: 230710440-230710440 on Build GRCh38
rs150161533 Chr.1: 230710399-230710399 on Build GRCh38
rs763661780 Chr.1: 230710335-230710335 on Build GRCh38
rs764938517 Chr.1: 230710361-230710361 on Build GRCh38
rs1038351739 Chr.1: 230710343-230710343 on Build GRCh38
rs147355405 Chr.1: 230710505-230710505 on Build GRCh38
rs752658346 Chr.1: 230710461-230710461 on Build GRCh38
rs781347977 Chr.1: 230710460-230710460 on Build GRCh38
rs139513443 Chr.1: 230710541-230710541 on Build GRCh38
rs2229389 Chr.1: 230710511-230710511 on Build GRCh38
rs750256750 Chr.1: 230710390-230710390 on Build GRCh38
rs767370325 Chr.1: 230710538-230710538 on Build GRCh38
rs751222492 Chr.1: 230710533-230710533 on Build GRCh38
rs756049025 Chr.1: 230710542-230710542 on Build GRCh38
rs564074355 Chr.1: 230710330-230710330 on Build GRCh38
rs886046082 Chr.1: 230710524-230710524 on Build GRCh38
rs777960151 Chr.1: 230710469-230710469 on Build GRCh38
rs771231332 Chr.1: 230710539-230710539 on Build GRCh38
rs776665337 Chr.1: 230710545-230710545 on Build GRCh38
rs559327874 Chr.1: 230710454-230710454 on Build GRCh38
rs753920149 Chr.1: 230710543-230710543 on Build GRCh38
rs61757173 Chr.1: 230710384-230710384 on Build GRCh38
rs144347709 Chr.1: 230710436-230710436 on Build GRCh38
rs555684009 Chr.1: 230710411-230710411 on Build GRCh38
rs772453722 Chr.1: 230710504-230710504 on Build GRCh38
rs774944406 Chr.1: 230710331-230710331 on Build GRCh38

View Full Product Details