Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41343157-41343410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX3X
Entrez Gene ID
1654
Gene Name
DEAD-box helicase 3, X-linked
Gene Aliases
CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102
UniGene
Hs.728563
Gene Chromosome Location
Chr.X: 41333308-41364472 on Build GRCh38
Transcripts
NM_001193416.2 NM_001193417.2 NM_001356.4 NR_126093.1 XM_011543892.2 XM_017029313.1 XM_017029314.1

SNP Detail

SNP ID Chromosome Location
rs886039719 Chr.X: 41343307-41343307 on Build GRCh38
rs869312692 Chr.X: 41343291-41343291 on Build GRCh38
rs1000898735 Chr.X: 41343308-41343308 on Build GRCh38
rs959358916 Chr.X: 41343366-41343366 on Build GRCh38
rs767609622 Chr.X: 41343204-41343204 on Build GRCh38
rs751416043 Chr.X: 41343365-41343365 on Build GRCh38
rs200078632 Chr.X: 41343363-41343363 on Build GRCh38
rs754913084 Chr.X: 41343197-41343197 on Build GRCh38
rs777274761 Chr.X: 41343272-41343272 on Build GRCh38
rs373063765 Chr.X: 41343196-41343196 on Build GRCh38
rs752955693 Chr.X: 41343365-41343369 on Build GRCh38
rs796052237 Chr.X: 41343271-41343272 on Build GRCh38
rs762956543 Chr.X: 41343199-41343199 on Build GRCh38
rs758794511 Chr.X: 41343366-41343371 on Build GRCh38
rs1032666321 Chr.X: 41343359-41343359 on Build GRCh38
rs375996245 Chr.X: 41343258-41343258 on Build GRCh38
rs875989803 Chr.X: 41343249-41343249 on Build GRCh38
rs755750366 Chr.X: 41343268-41343268 on Build GRCh38
rs1021070719 Chr.X: 41343208-41343208 on Build GRCh38

View Full Product Details